imPULSE Single-Use Mixers (S.U.M.s)
The imPULSE Single-Use Mixer (S.U.M.) can be utilized for many bioprocess mixing applications. The imPULSE design features include innovative disc mixing technology, configurable high-end controls, and monitors to fit specific process requirements. These features all enable uniform, superior mixing—scalable from 30 L to 5,000 L.
The touchscreen console for the imPULSE S.U.M. provides superior integrated sensor monitoring, and pump control for pH and saline titration and for automatic fill and harvest of the S.U.M.
Innovative mixing technology
  • Available in 30, 100, 250, 500, 1,000, 2,000, 3,000 and 5,000 L
  • Touchscreen Console provides ease of use with multi-functional capabilities to monitor and control mixing parameters
  • Integrated rolling diaphragm that provides the pumping action to the mixing disc; the diaphragm will not abrade the surfaces or produce particulate
  • Mixing tank jacket and insulation
  • Weighing systems that utilize load cells enable accurate batch weight monitoring
  • Auto inflate and vent control options
  • Adjustable powder port for 1,000 L and higher that fits 1, 5, and 25 kg Powdertainer BPCs
  • Open cart frame for easier cleaning
Areas of applications:
  • Media preparation
  • Final formulation steps
  • Buffer preparation
  • Harvest vessels
  • Large-volume mixing
  • Pooling and transfer
  • Resuspension
  • Mixing and storing multiple batches
  • Viral inactivation
The disc is an integrated part of the single-use BPC and is designed with slots and silicone flaps. The moving disc creates a pulsing action that has two effects:
1. On the upstroke, the flaps open as the disc moves up from the bottom of the mixing bag and fluid flows through the slots of the disc.
2. On the downstroke, the flaps close as the disc moves down and energy is directed down and channeled through the space between the mixing disc profile and the vessel side walls.
As a result, the velocity of the displaced liquid provides robust mixing.
Rolling diaphragm: Provides an interface between the BPC and the movable shaft with the mixing disc. There are no contact surfaces between the side walls and the diaphragm; therefore, the system does not generate particulates.
Linear scalability: The imPULSE mixing disc diameter increases proportionally as the size of the vessel increases. The ratio of the displaced liquid volume to the vessel volume is the same from all sizes ranging from 30 L to 5,000 L. This enables consistent and reproducible results, as processes are seamlessly scaled up across all sizes.
Graphical diagram of the mixing disc technology
  • Available in 30, 100, 250, 500, 1,000, 2,000, 3,000 and 5,000 L
  • Touchscreen Console provides ease of use with multi-functional capabilities to monitor and control mixing parameters
  • Integrated rolling diaphragm that provides the pumping action to the mixing disc; the diaphragm will not abrade the surfaces or produce particulate
  • Mixing tank jacket and insulation
  • Weighing systems that utilize load cells enable accurate batch weight monitoring
  • Auto inflate and vent control options
  • Adjustable powder port for 1,000 L and higher that fits 1, 5, and 25 kg Powdertainer BPCs
  • Open cart frame for easier cleaning
Areas of applications:
  • Media preparation
  • Final formulation steps
  • Buffer preparation
  • Harvest vessels
  • Large-volume mixing
  • Pooling and transfer
  • Resuspension
  • Mixing and storing multiple batches
  • Viral inactivation
The disc is an integrated part of the single-use BPC and is designed with slots and silicone flaps. The moving disc creates a pulsing action that has two effects:
1. On the upstroke, the flaps open as the disc moves up from the bottom of the mixing bag and fluid flows through the slots of the disc.
2. On the downstroke, the flaps close as the disc moves down and energy is directed down and channeled through the space between the mixing disc profile and the vessel side walls.
As a result, the velocity of the displaced liquid provides robust mixing.
Rolling diaphragm: Provides an interface between the BPC and the movable shaft with the mixing disc. There are no contact surfaces between the side walls and the diaphragm; therefore, the system does not generate particulates.
Linear scalability: The imPULSE mixing disc diameter increases proportionally as the size of the vessel increases. The ratio of the displaced liquid volume to the vessel volume is the same from all sizes ranging from 30 L to 5,000 L. This enables consistent and reproducible results, as processes are seamlessly scaled up across all sizes.
Graphical diagram of the mixing disc technology
imPULSE S.U.M. featured animation
The imPULSE single-use mixing system is ideal for a wide range of liquid-to-liquid or powder-to-liquid mixing applications, utilizing an innovative mixing disc technology and customizable BPCs.
Cytovance and Thermo Fisher Scientific imPULSE Mixer Collaboration
This video highlights the success Cytovance has had solving complex issues with our Thermo Scientific imPULSE Single-Use Mixers (S.U.M.s) and BioProcess Containers (BPCs).
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextCytovance and Thermo Fisher Scientific imPULSE Mixer Collaboration
          This video highlights the success Cytovance has had solving complex issues with our Thermo Scientific imPULSE Single-Use Mixers (S.U.M.s) and BioProcess Containers (BPCs).
        2. Now Playing
          Up NextimPULSE Single-Use Mixer (S.U.M.) with Touchscreen
          Watch this video to learn how the Thermo Scientific imPULSE Single-Use Mixer (S.U.M.) is an ideal mixing solution for liquid-to-liquid or liquid-to-powder mixing applications with capabilities from basic mixing to fully customized applications.
      Related products
      Standard and custom BPCs and fluid transfer solutions covering a range of applications, sizes, and connection systems.
      Manufactured at three efficient, cGMP-compliant manufacturing sites: two in North America—Logan, UT, and Millersburg, PA—and one in Cramlington, UK.
      Ordering information
      Bioprocessing resources
      Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: