For owners of legacy EVOS imaging systems, we have conveniently archived all supporting technical resources, including installation and user guides, specification sheets, application notes, videos, and more.
decorative image
decorative image
User guides and documentation
Demonstration videos
Software updates
decorative image
User guides and documentation
Compatible accessories
Application notes
Demonstration videos
Software updates
decorative image
decorative image
decorative image
Video Player is loading.
Current Time 0:00
Duration 5:00
Loaded: 3.29%
Stream Type LIVE
Remaining Time 5:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Specifications
    AttributeDetails
    Compatible vesselsWell plates, 35 mm dishes, T25 flasks
    Temperature rangeAmbient to 40°C
    CO % range0 to 20%
    O2 % range0 to 21% (ambient)
    Humidity range>80% relative humidity at 37°C
    Dimensions25 x 19 x 3.7 cm (Environment chamber)
    37 x 16 x 20 cm (Control unit)
    Weight1.5 kg (Environment chamber)
    10 kg (Control unit)
    Video Player is loading.
    Current Time 0:00
    Duration 0:04
    Loaded: 100.00%
    Stream Type LIVE
    Remaining Time 0:04