Varioskan LUX Multimode Microplate Readers and Invitrogen EVOS cell imaging systems
Why compromise on performance, ease of use, or flexibility?  
For plate reading and imaging applications, Invitrogen EVOS Cell Imaging Systems, Thermo Scientific High-Content Platforms, and Thermo Scientific Varioskan LUX Multimode Microplate Readers work together to bring you results—quickly and easily. Using the right combination of analysis platforms helps enable experiments that would otherwise be limited by a competing need for instrumentation in your lab. Follow time-lapse imaging of your biofilm development using the EVOS FL Auto 2 Imaging System with the EVOS Onstage Incubator. Even a multiday imaging experiment will not interfere with your ability to measure calcium flux or gene expression using luminescence on the Varioskan LUX multimode reader.
Request a joint quote   Request a joint demo
See an optimized solution to quantify and visualize cells
Video Player is loading.
Current Time 0:00
Duration 3:33
Loaded: 4.65%
Stream Type LIVE
Remaining Time 3:33
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Analysis of biofilm matrix
    Fluorescence-based imaging and quantitation of biofilm matrix using wheat germ agglutinin
    Fluorescence-based imaging and quantitation of biofilm matrix using wheat germ agglutinin (WGA). (A) Invitrogen WGA Alexa Fluor 488 fluorescent conjugate–based imaging and (B) quantification of poly-N-acetylglucosamine (PNAG) is shown in the biofilm matrix of S. aureus using an EVOS FL Cell Imaging System and Varioskan LUX Multimode Microplate Reader, respectively. See the complete microbial biofilms application note.
    Cell imaging systems
    Cellular imaging with Thermo Fisher Scientific draws on the combined strengths of Invitrogen EVOS cell imaging systems and Thermo Scientific high-content platforms and analysis tools. Select an all-in-one transmitted light system to fit in your cell culture hood, or an automated fluorescence system with an onstage incubator, plate scanning, image stitching, and integrated analysis.
     Learn more about
    With solutions for simple transmitted light microscopy and multiplexed confocal imaging, Thermo Fisher Scientific systems are designed to fit your cellular imaging needs.
    Varioskan LUX multimode reader
    Varioskan LUX multimode reader
    The Varioskan LUX Multimode Microplate Reader comes equipped with a flexible range of measuring technologies including absorbance, fluorescence intensity, luminescence, AlphaScreen technology, and time-resolved fluorescence. The instrument simplifies measurement setup with automatic dynamic range selection, flexible wavelength selection, integrated gas control, and smart safety controls.
     Learn more about
    The Thermo Scientific Varioskan LUX Multimode Microplate Reader is designed specifically for bioscience researchers with a wide variety of needs and assay requirements.
    Environmental control
    These imaging instruments and microplate readers have the option for environmental control features for extended experimental protocols using live cells—together they can tell you a more complete biological story.
    For Research Use Only. Not for use in diagnostic procedures.