The media bottle that makes lab work less work
Designed to provide better ergonomic features, ease of use, and reduced chance of contamination. The compact design of the Gibco media bottle allows for easier handling and storage.
The Gibco bottle is now available…empty. Now you can enjoy the benefits of the Gibco bottle whether you are buying liquid Gibco media and reagents or powder ingredients to make your own formulations. The Gibco bottle is sterile-packed and triple-bagged.
Cartoons of 4 bottles from 1962 to present
product photo
Purposeful design
The Gibco bottle was developed using anthropological research methods in cell culture laboratories spanning many geographies. Cell culture researchers provided candid input and feedback throughout the process, resulting in a new media bottle that can improve both your laboratory experience and your research results.
  • Angled neck makes pipetting and pouring easier
  • Larger footprint and squared profile improves stability on the bench and under the hood
  • Wider mouth reduces the chance that pipette will touch the sides and contaminate the contents
  • Increased headspace provides more room for addition of supplements
  • Narrower shape is more comfortable for small hands and takes up less storage space
product photo
Reproducible results
The more you control the variables in your research process, the more you can count on reproducing your results each time. The Gibco bottle is more comfortable, making repetitive tasks less painful to replicate precisely. The wider mouth of the Gibco bottle reduces the risk of contamination, which can jeopardize both your research and the work of others in your lab.
product photo
Ergonomic optimization
Not all media bottles are created equally. The Gibco bottle was designed to improve the comfort level of researchers who perform cell culture under the hood on a regular basis. The angle of the neck allows you to adopt a more natural position while pipetting, thereby reducing shoulder strain and the opportunity to develop repetitive strain injuries associated with laboratory work. The larger cap with more pronounced ridges allows easier one-handed opening and closing.
product photo
Environmental responsibility
Thermo Fisher Scientific is committed to environmental sustainability and to providing you with the best options to help you make responsible choices when purchasing recyclable products for use within your lab. The Gibco bottle is made from recyclable PET plastic, the most sought after class of plastic for use in garments, carpets, containers, and many other recycled products. The cap of the Gibco bottle is HDPE, also a valued recyclable plastic.  
Ordering information
Catalog # Name Size Price (USD) Qty
10339001 Each
1,496.00
10340001 Each
1,492.00
10341001 Each
978.00
GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Resources and support
Resources
Gibco Media Selection Tool
Find the right Gibco media for your application.  Use this interactive tool to select media based upon culture type, components, additives, and format. 
Cell Culture Select Tool
Search your cell line of interest to find product recommendations and resources to help enable a successful cell culture.
Gibco Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and the latest advancements in the field.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.