Highly sensitive enrichment of methylated DNA
MethylMiner™ Methylated DNA Enrichment Kit enables superior enrichment and differential fractionation of double-stranded DNA based on CpG methylation density, with increased sensitivity over antibody-based methods. Fractionation permits important comparisons between samples and enables researchers to focus analysis on only the methylation densities of interest.
Advantages of MethylMiner™ Methylated DNA Enrichment Kit include:
  • Partitioning with high affinity binding—at least 4-fold greater sensitivity than antibody-based methods
  • Fractionation based on CpG methylation density— ds DNA capture is achieved with MBD2 protein and facilitates ligation of double-stranded adaptors for next-generation sequencing
  • Rapid and easy elution with salt eliminates the need for proteinase K treatment and phenol:chloroform extraction
  • Precise answers—fractionated DNA permits distinctions to be made regarding methylation status and density
  • Fast protocol—completed in less than 4 hours
  • Easy—simple handling with Dynabeads® - the gold standard in magnetic beads
Figure 1— MBD-biotin based CPG methylated DNA affinity Capture
The Methyl Miner™ Methylated DNA Enrichment Kit allows for a diversity of elution strategies, depending on you preferred work flow and downstream application. It supports: 1) a single elution using undiluted high salt elution buffer; 2) a single elution using Proteinase K ; 3) a series of step-wise elutions with buffers containing successively greater NaCl concentrations; or 4) step-wise fractional elutions followed by a final Proteinase K treatment. This method overcomes challenges associated with denatured DNA using antibody-based methods. The dsDNA is more compatible with downstream epigenetic analysis applications—including next-generation sequencing.
Figure 2 – MethylMiner™ Methylated DNA Enrichment Kit captures more heavily methylated DNA compared to an antibody-based method. 
Relative capture and recovery of qPCR amplicon 1 DNA by MethylMiner™ Methylated DNA Enrichment Kit and anti-5mC from two different vendors.   In parallel, equal moles of anti-5mC antibody molecules were coupled to M-280 Protein G Dynabeads and MethylMiner MBD-biotin molecules were coupled to M-280 Streptavidin Dynabeads.  After coupling, triplicate aliquots of 10 µL of antibody-beads were incubated with 0.5 µg of fragmented, heat-denatured MCF-7 ssDNA and triplicate aliquots of 10 µL of MethylMiner beads were incubated with 0.5 µg of fragmented, non-denatured dsDNA.  After mixing at 22ºC for one hour, the supernatants were recovered, the beads were washed and then serially eluted with buffers of increasing NaCl concentration.  As a final step, all beads were treated with 20 µg of proteinase K at 56ºC for two hours.  The proteinase K digest supernatants were collected and extracted with phenol:chloroform:isoamyl alcohol (25:24:1).  All fractions were ethanol precipitated, redissolved in water, and assayed with amplicon 1 specific primers by qPCR using a SYBR® GreenER mastermix.  Recovery of amplifiable DNA was measured relative to a standard curve of input fragmented genomic DNA.  As shown, ~100% of the  heavily methylated DNA sequence was captured by the MethylMiner beads and ~80% could be eluted with 1 M NaCl.  In contrast, the anti-5mC antibody from two different vendors could only capture ~10-20% of this heavily methylated DNA and elution could only be achieved with proteinase K treatment.
Ordering Information
Catalog # Name Size Price (USD) Qty
ME10025 MethylMiner™ Methylated DNA Enrichment Kit Each
1,238.00
Epigenetics Learning Center
Our Learning Center provides an overview of epigenetic regulation of gene expression as well as considerations underpinning the research workflow. Learn about:
microRNAs
DNA methylation