Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator and collected in the CFC-Free Refrigerated Vapor Trap. Solvent vapors condense in a glass condensation flask preventing the vapors from reaching the oil pump and causing degradation.
Thermo Scientific™ Savant™ Refrigerated Vapor Traps are available in two temperature ranges. The Savant RVT450 refrigerated vapor trap ranges from ambient temperature to -50°C and the Savant RVT5105 refrigerated vapor trap ranges from ambient temperature to -105°C.
Features of both models include:
The RVT5105 also features:
![]() Refrigerated Vapor Trap: RVT450 | ![]() Ultra-low temp Refrigerated Vapor Trap: RVT5105 | |
---|---|---|
Weight | 71 lbs / 32kg | 150 lbs / 68kg |
Dimensions | 14x24x12 in (35x31x60cm) | 24x24x18.3 in (61x61x47cm) |
Capacity | 4L | 4L |
Temperature | -50°C | -105°C |
Solvent types | Water, non-aggressive organic solvents | Organic solvents, highly volatile solvents (methylene chloride, acetone, hexane, chloroform), acids, water, and both low and high boiling point solvents |
Available options |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.