Genetic selection has become an important tool for livestock breeders. With the use of high- to low-density Applied Biosystems Axiom arrays you can select genetically superior sires, replace heifers, and make timely breeding decisions.  Let us help you with your genomic testing challenges for livestock.
Customer testimonials
Improving Herd Performance: Thermo Fisher's Crucial Role in Ireland's Genotyping Program
Reducing greenhouse emissions in Ireland through cow genomic testing
Personalized herd management in sheep with Axiom genotyping platform
Genomics and Increased Selection Accuracy in Aquaculture
Products
axiom-expert-catalog
Axiom expert catalog genotyping arrays
Our catalog array-based genotyping solutions are ideal for the identification, validation, and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays.
These low-, medium-, and high-density arrays offer the power and resolution for a wide range of applications in plant and animal breeding and genomics. These applications include studying marker-trait association, evaluating elite lines, and identifying multi-line reference populations, as well as research applications for genome wide analysis and selective sweep analysis studies.
axiom-mydesign-note
Axiom myDesign custom genotyping arrays
Our catalog array-based genotyping solutions are ideal for the identification, validation, and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays.
Axiom myDesign custom Arrays give you the power to develop an effective array for genome-wide association, replication, fine mapping, and candidate gene studies in any species.
With Axiom myDesign custom genotyping arrays, fully- or semi-customized arrays containing 300 to 2.6 million markers can be designed. Use variants you provide, or choose SNPs from our Axiom Genomic Database, the world's largest resource of genotype-tested SNPs.
Multiple-species arrays are supported without any restriction on number of markers per species.
Resources
Agrigenomics Genotyping Arrays
The Importance of Microarrays in Agrigenomics
Reducing greenhouse gases with microarrays
Highly reliable and cost-effective SNP selection for parentage and other important traits product bulletin
Axiom-buffalo-genotyping-array
Axiom Porcine Genotyping Array