TaqMan real-time PCR master mixes
Applied Biosystems TaqMan master mixes have been optimized for use across a wide variety of research and discovery-stage biopharmaceutical applications. Choosing the right master mix depends on your target (RNA or DNA), application, test format, and multiplexing needs. For most qPCR research applications, we recommend using either TaqMan Fast Advanced Master Mix or TaqMan Fast Virus 1-Step Master Mixes
Which TaqMan Master Mix is right for you?
TaqMan Fast Advanced Master Mix
TaqMan Fast Virus 1-Step Master Mixes
Check price or order now
Check price or order now
Key applications
  • Gene expression analysis (2-step)
  • DNA detection
  • miRNA analysis
  • Gene expression analysis (1-step)
  • RNA virus detection
Target
DNA, including cDNA
RNA
Concentration
2X
4X
Multiplexing and passive reference dye
With ROX:  up to 2 targets
Available in 2 versions:
With ROX: up to 3 targets
No ROX: 3 or more targets 
Cycling mode/run time
Fast:  ~40 min
Standard: ~50 min
Fast: ~60 min
Standard: ~90 min
Carryover contamination control
Available in 2 versions:
With UNG
No UNG
No UNG
Reverse transcriptase
Purchase SuperScript IV VILO Master Mix separately and use before qPCR
Thermostable MMLV RT included in master mix
Assay recommendations
TaqMan Gene Expression assays
TaqMan Gene Expression assays
Request a sample
Request a sample