Obtain high yields of functional neuronal protein from all neuronal cell compartments
by Scott Meier, M.S.; Hai-Yan Wu, Ph.D.; Kay Opperman, Ph.D.; Suzanne Smith, M.S.; Barbara Kaboord, Ph.D. - 03/05/12
Results and Discussion
Neuronal tissue and primary cells are often used in neuroscience research to study proteins involved in learning, behavior and neurodegenerative disease. The Thermo Scientific N-PER Neuronal Protein Extraction Reagent is a specialized formulation for extracting total protein from neuronal tissue while maintaining protein function for downstream assays. The N-PER* Reagent typically produces better protein yield and extraction efficiency than other reagents commonly used for protein extraction.
Neurons have a unique morphology and lipid content that make total protein extraction from all cellular compartments more difficult than other cell types. Dendrites branch near the neuronal cell body and act as signaling sensors; axons extend away from the cell body and transduce signals to the synapse. Axons and dendrites are different in structure and function and isolation of proteins present in these projections can be difficult. Furthermore, the axon is coated with an insulative myelin sheath rich in glycolipid, sphingomyelin, and cholesterol, which adds to the difficulty of extracting protein from this structure. Gentle methods of total neuronal protein extraction are often ineffective. Although denaturing detergents improve total neuronal protein extraction, such harsh conditions negatively affect protein function.
The N-PER Reagent enables total protein isolation, including membrane proteins, from neuronal tissue or primary cultured cells, without adversely affecting protein function. We evaluated extraction by quantifying total protein yield and analyzing proteins from different neuronal compartments by Western blot. We also evaluated reagent compatibility with several downstream activity assays related to neuronal protein function.
Approximately 25-50% more total protein is extracted per milligram of tissue with the N-PER Reagent than other common detergent-based buffers and three-fold more compared to phosphate-buffered saline (PBS; Figure 1). The average yield from one hemisphere of fresh mouse brain was 18.3mg of total protein; the average yield from one million cultured primary neuronal cells was 0.35mg total protein (Table 1). Additionally, protein was extracted from multiple cellular compartments, including membranes. N-PER Reagent was more effective at extracting complex integral membrane proteins (e.g., N-methyl-D-aspartate receptor type 2B) and membrane-associated proteins (e.g., flotillin-1 and postsynaptic density protein 95) compared to other reagents (Figure 2). Interestingly, lysates prepared with N-PER Reagent extracted less glial fibrillary acidic protein (GFAP), a protein marker for astrocytes, than the other reagents, suggesting that N-PER Reagent preferentially extracts protein from neuronal cells (Figure 2).
A12n02-Fig1-87792-N-PER
Figure 1. Thermo Scientific N-PER Reagent extracts more protein from mouse brain tissue than other reagents. Tissue was processed with a Dounce homogenizer, and the protein was quantitated using the Thermo Scientific Pierce BCA Protein Assay (Part No. 23225).
Table 1. Average total protein yields from neuronal tissue and primary cells
SourceSample SizeReagent† VolumeAverage Total ProteinProtein Concentration
Fresh Mouse Brain1 brain hemisphere2 mL18.3±0.4 mg9.31±0.2 mg/mL
Primary Neuronal Cells106 cells0.5 mL0.35±0.01 mg0.69±0.02 mg/mL
†Thermo Scientific N-PER Neuronal Protein Extraction Reagent
A12n02-Fig2-87792-N-PER
Figure 2. Extraction of specific neuronal proteins. Lysates (10mL per lane) were separated by SDS-PAGE, transferred to nitrocellulose, and probed for specific neuronal proteins, including membrane-bound associated proteins (NMDAR2B, AMPA, PSD-95, synaptophysin, flotillin-1), cytoplasmic protein (tyrosine hydroxylase, MAPK), and GFAP, a specific marker for astrocytes.  Lane 1: N-PER Reagent; Lane 2: T-PER Reagent, Lane 3: Phosphate-buffered saline; Lane 4: Phosphate-buffered saline with 0.1% Triton* X-100.
To assess protein function after extraction with N-PER Reagent, we performed several activity assays related to neuronal function. The Rho GTPases, including Rho, Rac, and Cdc42, regulate actin and microtubule dynamics with Rac1 promoting neurite formation and Rho serving as an inhibitor. Both GTPases are located in the cell body. We evaluated the ability of N-PER Reagent to extract native Rho and Rac GTPases from fresh mouse brain. Lysates were supplemented with 5mM MgCl2 and either 10mM GTPgs or 10mM GDP to keep the native Rho or Rac protein in active and inactive forms, respectively. Active Rho or Rac was isolated from the treated lysates using the Thermo Scientific Active Rho Pull-down and Detection Kit (Part No. 16116) or Active Rac Pull-down and Detection Kit (Part No. 16118). Both proteins were able to bind GTPyS or GDP in their respective nucleotide binding sites, and the GTP-bound form was capable of interacting with its downstream effector in the pull-down assay (Figure 3), indicating that the function of Rho and Rac1 was retained.
A12n02-Fig3-87792-N-PER
Figure 3. GTPase activity in neuronal tissue is retained after lysis and extraction. Brain tissue lysates (1mg) supplemented with 5mM MgCl2 and treated with GTPγS or GDP were incubated with the indicated GST-PBD and glutathione resin. Half of the eluted sample volumes were analyzed by western blot and probed using small GTPase-specific antibodies.
To determine if protein phosphatases retained activity upon extraction with the N-PER Reagent, we measured the fluorescence of fluorescein diphosphate (FDP), a substrate that fluoresces after phosphate cleavage by protein phosphatases. Phosphorylation of molecules in the brain is important in neurotransmitter release, long-term potentiation and neuronal differentiation. In particular, protein tyrosine phosphatases (PTP1B, SHP-2, PTEN and LAR) and serine/threonine phosphatases (PP1 and PP2A) are involved in cell signaling and are frequent targets for therapeutic drug development and inhibitor screening assays. Enzymatic activity was similarly preserved in samples prepared with N-PER Reagent and Thermo Scientific T-PER Tissue Protein Extraction Reagent (Figure 4). Also, ATPase/kinase activity was retained (Figure 5), indicating that the N-PER Reagent could be used for screening small molecules that affect kinase activity.
A12n02-Fig4-87792-N-PER
Figure 4. Protein phosphatase activity is retained after lysis and extraction. Brain tissue lysate (10mg) was incubated with a fluorogenic phosphatase substrate for 1 hour at 37°C. Subsequent hydrolysis of the substrate by phosphatases increases fluorescence.
A12n02-Fig5-87792-N-PER
Figure 5. Kinase/ATPase retains activity after lysis and extraction. ATPase activity was measured as a function of luciferase chemiluminescence. Active kinase/ATPases hydrolyze ATP, decreasing the luminescence of luciferase.
We also evaluated activity preservation of an enzymatic target involved in a disease model. Beta-secretase (BACE1), a transmembrane protease, is important in the progression of Alzheimer’s disease through the increased generation of beta-amyloid peptide insoluble aggregates. Beta-secretase is upstream of beta-amyloid peptide and is responsible for cleaving amyloid precursor protein (APP). We used a FRET-based system to evaluate preservation of beta secretase enzymatic activity in lysates. The N-PER Reagent solubilized beta secretase from the membrane while retaining activity (Figure 6). Brain tissue processed with the N-PER Reagent could be useful for researchers that use animal models to study Alzheimer’s disease and want to preserve protein function.
A12n02-Fig6-87792-N-PER
Figure 6. β-Secretase retains activity in neuronal cell lysates prepared with Thermo Scientific N-PER Reagent. Activity was evaluated using a FRET-based assay system (EMD Millipore). Increasing concentrations of brain tissue lysate were added to wells containing a fluorogenic peptide substrate and incubated for 1 hour at 37°C. Fluorescence was measured using a Safire* Plate Reader (Tecan Ltd.).
Conclusions
The N-PER Neuronal Protein Extraction Reagent extracts protein from neuronal tissue more efficiently than other extraction reagents tested. The N-PER Reagent is optimized for the extraction of proteins from neuronal tissue and primary cells. Extracts prepared using the N-PER Neuronal Protein Extraction Reagent had better protein yields than other reagents and retained protein function for downstream applications.
Methods
Protein extraction from neuronal tissue: Brain was harvested from 8-10 week old C57/BL6 mice. Brains were divided in half, weighed, and suspended in 10 volumes of buffer, containing protease inhibitors, per gram of tissue. Tissue was disrupted using Dounce homogenizers (7mL glass Dounce homogenizer) and incubated on ice for 10 minutes. Lysates were cleared by centrifugation at 10,000 × g for 10 minutes, and protein concentration of the supernatant was measured with the Pierce BCA Protein Assay Kit (Part No. 23225).
Primary neuronal cell culture and lysis: Brain tissue from embryonic mice (17 days) was dissected and dissociated enzymatically. Cells were grown on six-well plates at 106 cells/well for 3 weeks. Before harvest, media was aspirated and cells were washed with ice-cold PBS. N-PER Reagent was added to each well (0.5mL per well) and allowed to contact cells for 5 minutes on ice. Cells were scraped from the plate, transferred to a 1.5mL tube, and centrifuged for 10 minutes at 10,000 × g. Protein concentration was determined using the Pierce BCA Protein Assay Kit.
Western blot: Brain tissue lysates (10µL) were separated by SDS-PAGE, transferred to a nitrocellulose membrane and blocked for 1 hour. Blots were incubated overnight in primary antibody (Anti-NMDAR2B, Anti-AMPA, Anti-tyrosine hydroxylase, Anti-MAPK and Anti-GFAP from Cell Signal Technology; Anti-synaptophysin from Sigma Aldrich; Anti-PSD95 from Abcam; Anti-flotillin 1 from BD Bioscience) diluted 1:1000 in blocking buffer, washed in TBS-Tween*-20 (Part No. 28360), and incubated in either goat anti-mouse HRP (Part No. 32430) or goat anti-rabbit HRP (Part No. 32460) secondary antibodies diluted 1:20,000 in 5% non-fat milk in TBS-Tween-20. Blots were developed with Thermo Scientific SuperSignal West Pico Substrate (Part No. 34080) and imaged using a CCD camera with a 10 second exposure.
Active Rho/Rac pull-down: Lysates were prepared as previously described with N-PER Reagent containing 5mM MgCl2. Lysates (1mg) were incubated with 10mM GTPyS to activate GTPases, or 10mM GDP to inactivate GTPases for 15 minutes at 37°C and then quenched with EDTA and MgCl2. Active Rho and Rac were isolated according to the instructions for Active Rho Pull-down and Detection Kit (Part No. 16116) and Active Rac Pull-down and Detection Kit (Part No. 16118).
Protein phosphatase activity assay: A fluorescent protein phosphatase assay (Anaspec) was used to evaluate protein phosphatase activity. Briefly, 10µg of total protein was incubated in assay buffer with FDP substrate for 1 hour. The change in fluorescence from substrate hydrolysis was measured (excitation/emission: 485/520nm) using the Thermo Scientific Varioskan Flash Multimode Plate Reader.
ATPase/Kinase activity assay: ATP hydrolysis was measured as a function of luminescence of luciferase with the ATPlite Luminescence Assay Kit (Perkin Elmer) and a modified protocol. Briefly, brain tissue lysates (10µg) were spiked with 5mM ATP, final concentration. A portion of the lysate was inactivated as a negative control, according to the kit instructions. Lysates were added to an equal volume of reaction mix and luminescence was measured with a Varioskan Flash Multimode Plate Reader at 5 minute intervals for 1 hour. Values were normalized to initial luminescence values.
Beta-Secretase activity assay: Activity of beta-secretase was evaluated using a FRET-based assay system (EMD Millipore). Increasing concentrations of brain tissue lysate were added to wells containing a fluorogenic peptide substrate and incubated for 1 hour at 37°C. Fluorescence was measured (excitation/emission: 345/495nm) using a Safire Plate Reader (Tecan Ltd.).
General References
Saito, N. and Shirai, Y. (2002). Protein kinase C gamma (PKC gamma): Function of neuron specific isotype. J Biochem 132(5):683-7.
Want, C., et al. (2009). Protein phosphatase 2a regulates self-renewal of Drosophila neural stem cells. Development 136(13):2287-96.
Yan, R., et al. (1999). Membrane-anchored aspartyl protease with Alzheimer’s disease beta-secretase activity. Nature 402:533-7.
Neuronal Protein Extraction Reagent
Thermo Scientific N-PER Neuronal Protein Extraction Reagent is a proprietary cell lysis reagent optimized for efficient extraction of proteins from all cellular compartments of neuronal tissue and primary cultured neurons.
Features of N-PER Neuronal Protein Extraction Reagent:
  • Optimized—efficient total protein extraction, including membrane proteins, from brain tissue or primary cultured neurons
  • Gentle—preserves protein function without compromising yield
  • Versatile—can be supplemented with protease inhibitors, reducing or chelating agents or required cofactors
  • Compatible—extracts are suitable for use with total protein, enzymatic and immunological assays and protein purification methods
Learn more about Thermo Scientific N-PER Neuronal Protein Extraction Reagent
Related products
Catalog # Name Size Price (USD) Qty
16118 Active Rac1 Pull-Down and Detection Kit Each
979.65
Online Exclusive
1,128.00
Save 148.35 (13%)
16116 Active Rho Pull-Down and Detection Kit Each
1,028.65
Online Exclusive
1,128.00
Save 99.35 (9%)
32430 Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP 2 mL
171.00
32460 Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP 2 mL
194.00
28360 Pierce™ 20X TBS Tween™ 20 Buffer, 500 mL Each
116.65
Online Exclusive
129.00
Save 12.35 (10%)
23225 Pierce™ BCA Protein Assay Kit, 1 Kit (1 L) Each
253.65
Online Exclusive
293.00
Save 39.35 (13%)
For Research Use Only. Not for use in diagnostic procedures.