Particle Analysis for Battery Manufacturing Quality Assurance
Automated particle analysis in battery manufacturing
As we progress towards a greener future, lithium-ion batteries have emerged as a crucial component in electric vehicles and renewable energy storage solutions due to their high energy density and long life cycle. However, the manufacturing process of these batteries is not without its challenges. One of the major hurdles is the presence of impurity particles that can compromise the safety and durability of the batteries. This article will explore how automated particle analysis using scanning electron microscopy (SEM) is revolutionizing the quality control process in lithium-ion battery manufacturing.
Impurities such as copper (Cu), iron (Fe), chromium (Cr), and zinc (Zn) introduced during the battery production process significantly impact the electrochemical performance and material stability of the batteries. More alarmingly, they can even cause internal short circuits, leading to battery failure. A prime example is copper, a common impurity that can dissolve on the cathode side, diffuse to the anode side, and cause micro short circuits. Therefore, the identification and analysis of these impurities are crucial for enhancing battery safety and performance. This is where SEM comes in, offering a precise and efficient method for impurity analysis.
Battery manufacturing
The analysis of metal impurity particles, particularly in cathode powder material, is attracting attention from battery manufacturers globally. China, a leading manufacturer of lithium-ion batteries, has implemented the GB/T 41704-2022 standard for battery impurity particle testing to enhance safety and standardize the testing process. Traditionally, collecting and analyzing these impurity particles has been labor intensive and time consuming, often involving the use of a filter membrane or transferring the particles to carbon tape for manual analysis. However, with the advent of automated particle analysis technologies, these processes have been significantly streamlined. These advanced tools can reduce the analysis time per particle to less than one second, quickly providing results and generating detailed reports with conclusions.
Automated particle analysis software integrates with SEM to facilitate the entire particle analysis process. Users can define specific criteria for analyzing particles of interest, such as size parameters (diameter, aspect ratio, etc.) and particle categorization based on their composition and morphology. This level of detail aids in tracing the source of impurities, providing valuable insights for improving production process control.
Particle analysis also offers advanced reporting capabilities. It generates customized reports presenting impurity particle data through various charts and graphs, providing a visual assessment of whether the analysis outcome aligns with quality control standards. This feature is particularly beneficial for manufacturers who need to adhere to stringent quality control regulations.
Automated particle analysis using SEM is crucial for the quality control process in lithium-ion battery manufacturing. Automating the impurity particle analysis process enhances measurement efficiency, data reliability, and overall safety of the batteries. As the demand for lithium-ion batteries continues to grow worldwide, adopting such advanced analysis solutions will be instrumental in ensuring their reliability and performance, ultimately contributing to the broader goal of sustainable energy solutions.
Battery webinar
In this webinar, we discuss how automated particle analysis can help manufacturers obtain fast, reliable, and reproducible data about battery quality. Watch on demand to learn how particle analysis can help improve battery quality and to see a demonstration of Thermo Scientific Perception Software’s flexible reporting tools.
Articles on particle analysis
Read our industry-related articles to learn more about how our solutions can help solve your specific challenges.
Understanding Particulate Matter in Air Pollution: Significance, Measurement, and Advanced Analysis Techniques
Additive Manufacturing Powder Quality Control: Ensuring Excellence in Production
How to Control Your Cleanroom Cleanliness by Monitoring Airborne Particles
Enhancing Steel Quality through Scanning Electron Microscopy
Understanding Technical Cleanliness: Meeting ISO 16232 and VDA 19 Cleanliness Standards in the Automotive Industry
SEM EDS for Gunshot Residue Analysis
Particle Analysis eBooks and webinars
Quality control: Turn data into answers
Learn how Thermo Scientific Perception Software can support your work with standardized, recipe-based workflows
Perception Software introduction webinar
This webinar introduces our all-in- one software package that can turn your scanning electron microscope of choice into a dedicated solution for your particle analysis needs.
Perception Software demonstration webinar
In this webinar, we demonstrate how Thermo Scientific Perception Software on the Thermo Scientific Axia ChemiSEM Scanning Electron Microscope can be used to automatically acquire morphological and compositional data on particles, powders, and inclusions.
Thermo Scientific particle analysis instruments and software
Want to know more about Thermo Scientific ParticleX Desktop SEM?
Want to know more about Perception Software on the Axia ChemiSEM System?