Monitoring cleanroom particles
Cleanrooms and associated controlled spaces serve as environments that maintain critical levels of cleanliness for clean production, used in various industries, including pharmaceuticals, food, semiconductors, and microelectronics. Airborne particles in cleanrooms are defined as the microscopic particles suspended in the air. Their concentration defines the contamination level, which can significantly affect product quality and overall performance. The deposition rate of airborne particles onto the surface of manufactured sensitive parts is a vital measurement for production performance.
Cleanroom Phenom
Sources and impact of cleanroom particles
Airborne particles usually originate from human activities, equipment operation, and materials handling in cleanrooms. The size and type of airborne particles are of important consideration. Sub-micron particles, or ultrafine particles, can remain suspended in the air for extended periods, while larger particles settle quickly and pose a higher risk of surface contamination. In particular, particles larger than 5 μm are crucial in assessing contamination risk and surface cleanliness. Effective control measures are necessary to prevent these particles from depositing on vulnerable parts.
ISO cleanroom standards
ISO 14644 is an international standard that provides guidelines for cleanroom classification, design, operation, and monitoring. According to ISO 14644-1:2015, cleanrooms are classified based on the maximum allowable particle concentration per cubic meter of air, with Class 1 being the strictest and Class 9 the least strict. While monitoring airborne particles is critical, surface cleanliness, as addressed in ISO 14644-9:2012, and particle deposition rates, covered in ISO 14644-3:2019, are also essential. The recent ISO 14644-17:2021 offers practical guidance on particle deposition rate applications in cleanrooms.
Particle analysis techniques
Microscopes, such as optical microscopes and scanning electron microscopes (SEM), can be used to count and visualize deposited particles. SEM can provide additional information on particle composition through energy-dispersive X-ray spectroscopy (EDS). SEM- EDS offers several advantages:
High detail and resolution—SEM provides a high level of detail and resolution, allowing for the visualization and characterization of particles at the microscopic level, which are crucial for understanding contamination sources.
Elemental composition identification—The EDS detector allows for determination of the elemental composition of particles. This information can be used to pinpoint the possible sources of contamination and to implement targeted control measures.
Automated quantitative analysis—Integrated software enables accurate counting and sizing of particles. Automated software can be employed to process the SEM-EDS data, providing essential statistical data on particle concentration.
Cleanroom cleanliness management and quality control
Cleanrooms are pivotal in industries requiring stringent environmental control to ensure product quality and reliability. Monitoring and controlling airborne particle contamination are essential for maintaining surface cleanliness. Other methods for measuring surface cleanliness may struggle with accuracy, secondary contamination, or damage to fragile parts. In contrast, SEM-EDS combined with gravitational samplers provides a high-resolution, detailed approach to particle analysis, making it an effective tool for cleanroom management and quality control. Adhering to ISO protocols and standards helps assess contamination levels and implement effective control measures.
Automated SEM-EDS enhances surface cleanliness management in cleanroom manufacturing
Discover how Perception Software provides automated image acquisition and processing of SEM-EDS data for continuous monitoring of surface cleanliness in cleanroom manufacturing.
Articles on particle analysis
Read our industry-related articles to learn more about how our solutions can help solve your specific challenges.
Understanding Particulate Matter in Air Pollution: Significance, Measurement, and Advanced Analysis Techniques
Additive Manufacturing Powder Quality Control: Ensuring Excellence in Production
Quality Assurance in Battery Production: The Power of Automated Particle Analysis
Enhancing Steel Quality through Scanning Electron Microscopy
Understanding Technical Cleanliness: Meeting ISO 16232 and VDA 19 Cleanliness Standards in the Automotive Industry
SEM EDS for Gunshot Residue Analysis
Particle Analysis eBooks, App notes, and webinars
Quality control: Turn data into answers
Learn how Thermo Scientific Perception Software can support your work with standardized, recipe-based workflows.
Perception Software demonstration webinar
In this webinar, we demonstrate how Thermo Scientific Perception Software on the Thermo Scientific Axia ChemiSEM Scanning Electron Microscope can be used to automatically acquire morphological and compositional data on particles, powders, and inclusions.
Perception Software introduction webinar
This webinar introduces our all-in- one software package that can turn your scanning electron microscope of choice into a dedicated solution for your particle analysis needs.
Thermo Scientific particle analysis instruments and software
For Research Use Only. Not for use in diagnostic procedures.
Want to know more about Thermo Scientific ParticleX Desktop SEM?
Want to know more about Perception Software on the Axia ChemiSEM System?
1x1 image pixel for data collection