Mouse intestine cryosection showing basement membranes.
Mouse intestine cryosection showing basement membranes labeled with our chicken IgY anti-fibronectin antibody (Cat. No. A21316) and the Alexa Fluor® 488 Goat Anti–Chicken IgG (Cat. No. A11039, green). Goblet cells and crypt cells were labeled with Alexa Fluor® 594 wheat germ agglutinin (Cat. No. W11262, red). The microvillar brush border and smooth muscle layers were visualized with Alexa Fluor® 680 phalloidin (Cat. No. A22286, pseudocolored purple). The section was counterstained with DAPI (Cat. No. D1306, D3571, D21490, blue).
Mouse intestine cryosection showing basement membranes labeled with our chicken IgY anti-fibronectin antibody (Cat. No. A21316) and the Alexa Fluor® 488 Goat Anti–Chicken IgG (Cat. No. A11039, green). Goblet cells and crypt cells were labeled with Alexa Fluor® 594 wheat germ agglutinin (Cat. No. W11262, red). The microvillar brush border and smooth muscle layers were visualized with Alexa Fluor® 680 phalloidin (Cat. No. A22286, pseudocolored purple). The section was counterstained with DAPI (Cat. No. D1306, D3571, D21490, blue).
Related Products
Catalog # Name Size Price (USD) Qty
A22286 Alexa Fluor™ 680 Phalloidin, Near-infrared, 679/702 nm Each
628.65
Online Exclusive
656.00
Save 27.35 (4%)
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D21490 DAPI, FluoroPure™ grade, 10 mg, 10 mg Each
225.65
Online Exclusive
231.00
Save 5.35 (2%)
D3571 DAPI (4',6-diamidino-2-phenylindole, dilactate), 10 mg, 10 mg Each
151.65
Online Exclusive
156.00
Save 4.35 (3%)
W11262 Wheat Germ Agglutinin, Alexa Fluor™ 594 Conjugate, 5 mg 5 mg
473.65
Online Exclusive
487.00
Save 13.35 (3%)
Related Images