Brain of a Xenopus laevis embryo.
A whole mount of the embryonic brain of Xenopus laevis that has been double-labeled with our lysine-fixable 10,000 MW fluorescein dextran (fluoro-emerald, Cat. No. D1820) and lysine-fixable 10,000 MW tetramethylrhodamine dextran (fluoro-ruby, Cat. No. D1817). The tetramethylrhodamine dextran was used to label the neurons projecting from the retina, whereas the fluorescein dextran was applied to the transected spinal cord, thus allowing the detailed evaluation of the topological relationship of these two populations of neurons. Image contributed by Martina Manns and Bernd Fritzsch, Creighton University.
A whole mount of the embryonic brain of Xenopus laevis that has been double-labeled with our lysine-fixable 10,000 MW fluorescein dextran (fluoro-emerald, Cat. No. D1820) and lysine-fixable 10,000 MW tetramethylrhodamine dextran (fluoro-ruby, Cat. No. D1817). The tetramethylrhodamine dextran was used to label the neurons projecting from the retina, whereas the fluorescein dextran was applied to the transected spinal cord, thus allowing the detailed evaluation of the topological relationship of these two populations of neurons. Image contributed by Martina Manns and Bernd Fritzsch, Creighton University.
Related Products
Catalog # Name Size Price (USD) Qty
D1817 Dextran, Tetramethylrhodamine, 10,000 MW, Lysine Fixable (fluoro-Ruby) Each
366.00
D1820 Dextran, Fluorescein, 10,000 MW, Anionic, Lysine Fixable (Fluoro-Emerald) Each
362.65
Online Exclusive
373.00
Save 10.35 (3%)
Related Images