DVC in Avizo Software for material deformation analysis
Using digital volume correlation to compute 3D displacement and strains from volume images
On-demand demo webinar
Thermo Scientific Avizo Software is an all-in-one image analysis digital lab that helps you quantify 3D full-field displacement and strain maps from volume images acquired before and after material deformation. The software can both visualize and analyze the changes to quantify deformation-induced microstructural changes, such as strain transfer in multiphase materials, pore growth and coalescence related to fractures, and crack opening displacement (COD).
In our previous webinar—which is also available on-demand—we provided an introduction to digital volume correlation. In this webinar, field experts showcased Avizo Software’s DVC capabilities.
Watch now to learn:
  • How to design a DVC experiment
  • The difference between global and local approaches to DVC
  • How DVC is used with large datasets 
  • The advantage of mechanical regularization
  • How DVC can help inform and validate numerical simulations
About the speaker(s)
Kamel Madi, CEO, 3Dmagination Ltd
Kamel Madi
CEO, 3Dmagination Ltd 
Kamel has more than 15 years of academic and industrial research experience. He has acquired solid methods in 4D imaging combined with digital volume correlation to capture dynamic processes of materials in service or during the manufacturing process across diverse areas such as the glass industry and healthcare. He now uses his expertise and skills to help enhance software performance, particularly focusing on optimizing user experience.
Loic Courtois, CSO, 3Dmagination Ltd
Loic Courtois
CSO, 3Dmagination Ltd 
Loic has worked in 4D imaging for many years and has a deep understanding of the complete workflow, from experimental design to analysis and data extraction. Drawing from his experience on a wide range of materials, Loic has been heavily involved in the development of the digital volume correlation interface in Avizo Software.
Luigi Raspolini, Thermo Fisher Scientific
Luigi Raspolini
Product Marketing Manager, Thermo Fisher Scientific 
Luigi is a process engineer working in materials science and characterization. With a background in spectroscopy and electron microscopy, he helps customers find the right tools to convert datasets into actionable data, track improvements, and identify imperfections in manufacturing or research processes.
Submit the form below to watch.
I would like to:
Email Permissions
To comply with your preferences, we need to confirm permission to send you communications by email. Personal information provided will be used in accordance with our Privacy Policy.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
1x1 image pixel for data collection