Search Thermo Fisher Scientific
Visitors who stopped by our booth during the Craft Brewers Conference & BrewExpo America discovered our latest analytical technologies to support their beer analysis and applications.
Have questions about your beer analysis? Fill out our form below to speak with an expert. You can also join our community for invitations to our events and information about the solutions we offer.
Gallery Plus Beermaster Discrete Analyzer for Beer Applications
Dedicated for beer, malt, and wort analysis, the Gallery Plus Beermaster discrete analyzer automates labor-intensive and time-consuming tests and delivers reliable analyses for multiple parameters. Increased efficiency in controlling the beer quality reduces costs and improves productivity in breweries. The discrete analyzer can accommodate 108 samples and 42 reagents in its separated sample and reagent disks, with the capacity to perform up to 350 tests/hour, and approximately 8 bitterness tests/hour in parallel with the BCM.
Read fully-integrated multiparameter analysis.
GENESYS Vis/UV-Vis Spectrophotometers
Meet your quality control and other beverage analysis needs with Thermo Scientific GENESYS Vis/UV-Vis Spectrophotometers.
Non-alcoholic and alcoholic drinks require reliable beverage analysis methods to assess ingredients and other definitive characteristics. Typically, this is accomplished with colorimetric measurements in the visible range. UV-Visible spectrophotometers offer quick, inexpensive, easy-to-use analysis to verify whether product quality for beverages like beer and wine is within specification.
The size, simplicity, and cost-effective operation of our GENESYS 50/150 spectrophotometer and the variety of tests available make it an ideal instrument for analyzing beer and evaluating brewing ingredients. Thermo Scientific BeerCraft Software makes it simple to ensure the consistency of each unique brew without complicated calculations. Pre-programmed methods enable quick and easy measurements of over twenty beer attributes at the touch of a button to maintain your beer’s identity and the success of your brand.
Read how pFriem Family Brewers use UV-Vis for quality control.
Our experts can guide you to the perfect solution for your needs. Contact us today!
ISQ 7610 Single Quadrupole GC-MS System
Maintain product consistency and stay on top of consumer demand with Thermo Scientific gas chromatograph (GC) and gas chromatography-mass spectrometer (GC-MS) systems.
Combine powerful GC-MS instruments with productivity-enhancing hardware and software to create complete, user-friendly solutions for your most pressing analytical needs.
Financial and leasing solutions
Acquiring equipment doesn’t have to be complicated. Let us show you how our financial solutions can help you obtain the equipment you need easily and affordably. Read more
Get credit towards a new Gallery discrete analyzer
Trade in your discrete or continuous flow analyzer system for credit towards the purchase of a Thermo Scientific Gallery or Gallery Plus Discrete Analyzer. Read more
Webinar, on demand: Control the beer process
Ensure a consistent product with effective quality monitoring throughout beer making stages to improve productivity with Gallery discrete analyzers. Register and view
Gallery Discrete Analyzer Beer Applications Compendium
Discrete analyzers offer flexibility and ease of use, as well as, a unique opportunity to automate time-consuming and labor-intensive wet chemical analysis. When conducting multiparameter analysis for malt, beer, and wine the discrete analyzer technology ensures high product quality and throughput while reducing cost, waste, and hands-on sample time. Discover more about the Thermo Scientific Gallery Discrete Analyzers in this applications compendium.
BeerCraft Software for GENESYS 50 and GENESYS 150 Spectrophotometers
Learn more about GENESYS instruments and how UV-Vis spectroscopy and BeerCraft software are changing beer brewing for the better.
Find out about our GENESYS 50 Free Trial Program
Try the Thermo Scientific GENESYS 50 UV-Vis Spectrophotometer for one week, then box it up and return it to us using the pre-paid freight label. There’s no cost to you! Request a trial unit today!
* Program available in US and Canada only