High-Throughput Screening Libraries for Small- Molecule Drug Discovery
Increase the odds of screening success while optimizing efficiency
High-throughput screening involves testing thousands or even tens of thousands of compounds to identify “hits” that bind to the target. The process is like trying many keys to find one (or more) that fit a lock.
The Thermo Scientific Maybridge Screening Collection comprises over 50,000 synthetically derived organic compounds that are structurally and functionally diverse and demonstrate suitable pharmacokinetic properties while excluding potentially problematic structures. These individually designed compounds have been manufactured by Thermo Fisher Scientific using innovative synthetic techniques, based on over 50 years of experience in heterocyclic chemistry. 
The choice of screening library can make or break a small-molecule drug discovery program in its early phases. Our screening libraries are designed to increase the probability of positive results while optimizing cost, time, and effort spent per project. 
Pre-plated high-throughput screening libraries
We offer three pre-plated libraries for high-throughput screening HTS that represent the diversity of the human pharmacophore. The compounds in all three libraries conform to Lipinski Rule of 5 (Ro5) guidelines for “drug-like” properties. They are also designed to reduce false positives by excluding reactive compounds that exhibit covalent bonding, toxicity, insolubility, or oversimplicity.
For libraries focused on specific types of targets, like antibacterial or kinase, see Focused screening libraries below.
Maybridge HTS library comparison table
Focused HTS screening libraries
In close collaboration with an external partner, the Maybridge collection includes target-specified libraries of drug-like compounds created by applying different computational methodologies based upon bioinformatics and structure-based drug discovery. These focused screening libraries of compounds are selected from available collections to facilitate the discovery of novel chemical entities exhibiting specific biological activities or to screen against certain molecular or biological targets. They have demonstrated high hit rates.
Maybridge focused screening libraries
Other focused libraries (such as libraries of compounds that screen against inhibitors of histone deacetylase or HDAC) are available upon request.
Properties and profile of the Maybridge Screening Collection
The Maybridge Screening Collection is widely recognized for its market-leading diversity and quality. In compound screening, the size of your library is not as important as its diversity—inclusion of representative compounds across the “drug-like” space. A highly diverse library of compounds with the right chemical properties is more likely to discover “hits” than larger, less diverse libraries with less suitable chemical properties.
The Maybridge collection comprises structures with complex and diverse properties. In medicinal chemistry, Lipinski’s Rule of 5 (Ro5) is widely considered to be a preliminary measure of a compound’s likelihood to be orally active in humans.1 The Ro5 proposes the parameters (all multiples of 5) listed below as crucial for consideration during drug discovery. Here, we provide the calculated distribution of our Maybridge Screening Collection for each parameter.
The industry-leading library for fragment-based drug screening
Maybridge Screening Collection profile on Lipinski’s Rule of 5 for drug-like properties
Molecular weight
The ideal molecular weight of a drug compound is less than 500 Daltons. The Maybridge screening collection is composed primarily of compounds that fall within this ideal range.
H-bond acceptors
Compounds containing a maximum of 10 H-bond acceptors are more likely to be orally active in humans. Every structure in our screening collection has M H-bond acceptors or fewer.
H-bond donors
Compounds containing a maximum of 5 H-bond donors are more likely to be orally active in humans. Each structure in our screening collection has 5 H-bond donors or fewer.
Partition coefficient (cLogP)
The calculated partition coefficient (cLogP) provides an insight to the solubility of a compound. This is a critical consideration as drugs must navigate both hydrophobic and hydrophilic environments to reach their targets. Compounds should have cLogP values less than 5, and our Maybridge screening collection is composed primarily of structures that fall within the ideal range.
Rotatable bonds
Compounds containing a maximum of 10 rotatable bonds are more likely to be orally active in humans. Over 90% of our screening collection contain 10 or fewer rotatable bonds.
Partition coefficient (cLogP)
All calculations were performed at Maybridge using the Tsar GD version G.G from Accelrys (salt data was ignored). Definitions from the Tsar Reference Guide are provided below. 
  • H-bond acceptors are defined as accepting a lone pair of electrons to form a hydrogen bond: –O–, C=O, –CN, –N=, 3° amines (excluding sp2), SH, or C=S.
  • H-bond donors are defined as O-H, N-H, and S-H bonds where atoms donate a lone pair of electrons to form a hydrogen bond.
  • cLogP is defined as the sum of the atomic partial log P values. (6)
  • Rotatable bonds are defined as any single order, non-terminal, non-ring, and non-amide bond.
Properties of Maybridge Screening Collection compounds
Availability in quantity
Typically, about 95% of the compounds in our collection are available in >5 mg stock quantities, over 90% in >50 mg stock quantities, and a large proportion in gram quantities. This means we can ensure a very high level of re-supply of originally tested compounds. We provide most major brands of plates and vials but are happy to use those supplied by customers on request.
1Lipinski CA, Lombardo F, Dominy BW, Feeney PJ. Experimental and computational approaches to estimate solubility and permeability in drug discovery and development settings. Adv Drug Deliv Rev. 2001; 46: 3–26. PubMed
2Viswanadhan VN, Ghose AK, Revankar GR, Robins RK. Atomic physiochemical parameters for three dimensional structure directed quantitative structure-activity relationships. 4. Additional parameters for hydrophobic and dispersive interactions and their application for an automated superposition of certain naturally occurring nucleoside antibiotics. J Chem Inf Comput. Sci. 1989; 29: 163–172. Abstract
3McGregor MJ, Pallai PV. Clustering of large databases of compounds: Using the MDL “keys” as structural descriptors. J Chem Inf Comput. Sci. 1997; 37: 443–448. Abstract
4Zhao YH, Le J, Abraham MH, et al. Evaluation of human intestinal absorption data and subsequent derivation of a quantitative structure-activity relationship (QSAR) with the Abraham descriptors. J Pharm Sci. 2001; 90: 749–784. PubMed
5Platts HA, Abraham MH, Zhao YH, Hersey A, Ijaz L, Butina D. Correlation and prediction of a large blood-brain distribution data set—an LFER study. Eur J Med Chem. 2001; 36: 719–730. PubMed
6Crivori P, Cruciani G, Carrupt PA, Testa B. Predicting blood-brain barrier permeation from three-dimensional molecular structure. J Med Chem. 2000; 43: 2204–2216. PubMed
Success stories for Maybridge HTS libraries
The Maybridge Screening Collection has been the source of many successful fragment screening projects.
The industry-leading library for fragment-based drug screening
Olaparib and its Maybridge collection precursor molecule. Olaparib (right, trade name Lynparza), FDA-approved in 2014 for treatment of for BRCA-mutant ovarian cancer, was developed from a compound from our Maybridge Screening Collection (left), one of several such drugs that have received regulatory approval. Olaparib is an inhibitor of poly ADP ribose polymerase (PARP), an enzyme involved in DNA repair. It acts against cancers in people with hereditary BRCA1 or BRCA2 mutations, which includes many ovarian, breast, and prostate cancers. Lynparza is now also a “first-line” maintenance treatment for BRCA-mutated metastatic pancreatic cancer, reducing the risk of disease progression or death in some patients.
The industry-leading library for fragment-based drug screening
Selected hits obtained from the Maybridge Screening Collection that have led to further drug development
For Research Use Only. Not for use in diagnostic procedures.
1x1 image pixel for data collection 1x1 image pixel for data collection