High-quality labware and equipment for chemical research applications.
Many chemical reactions require use of specialty labware. Specialty vessels and labware made from ceramics, glassy carbon, precious metals, non-precious metals, PTFE, quartz, or other materials help to maintain purity and limit corrosion or oxidation of chemical intermediates or products. Specialty labware also supports high-temperature syntheses and analyses. 
Thermo Scientific offers a selection of high-quality labware products suitable for laboratory applications.
Our diverse range of products includes:
  • High-purity, corrosion- and oxidation-resistant oxide ceramics for high-temperature applications
  • Platinum metal labware for analytical applications
  • Glassy carbon products—including crucibles suitable for use in high-temperature differential thermal/thermal gravimetric analyses, and specimen holders and cuvettes for atomic absorption and atomic emission spectroscopy and multi-element analyses
  • Non-precious metal standard crucibles available in zirconium, nickel, molybdenum, and other metals and alloys
  • General and specialty laboratory equipment
Featured products
Ceramic labware
Oxide ceramics are manufactured with materials of pure metal oxides without the admixtures of silicates. Our products are comprised of high-purity alumina (Al₂O₃), zirconia (ZrO₂), or magnesia (MgO).
Glassy carbon labware
High-temperature treatment imparts special material properties to our glassy carbon products, including significantly improved corrosion resistance, strength, good electrical conductivity, impermeability to gases and liquids, and more.
Laboratory equipment
Thermo Scientific offers various types of laboratory equipment necessary for a wide range of research and industry applications, including gas purification, spectroscopy, and more.
Non-precious metal labware
Our high precision refractory metal crucibles come in virtually unlimited shapes and sizes that offers advantages in cost, efficiency, and length of service.
PTFE labware
Our complete range of PTFE laboratory products are uniquely suitable for many laboratory applications because of their almost total chemical inertness and wide range of working temperatures, with an upper limit of about 290°C. 
Quartz labware
Fused quartz products are manufactured by fusing naturally occurring crystalline silica, imparting many desirable properties such as excellent electrical insulation, high melting point, chemical purity, and corrosion resistance.
Platinum labware
High-performance platinum and precious metal labware for crystal growth and analytical research applications
Platinum resists the action of almost all acids and is unaffected by a wide variety of corrosive substances. It has a high melting point, a low heat capacity, and can be heated in air without any weight loss due to oxidation. These properties make our platinum labware ideal for use in many analytical research applications.
Structure & Element Search Tools
Structure Search Tool
Search our chemical database for an exact or substructure match on your molecule of interest. Or use the canvas to construct, import or edit molecules.
Element Search Tool
Find pure elements, alloys, and compounds in a range of formats.
For Research Use Only. Not for use in diagnostic procedures.
Structure Search Tool
Search our chemical database for an exact or substructure match on your molecule of interest. Or use the canvas to construct, import or edit molecules.
Element Search Tool
Find pure elements, alloys, and compounds in a range of formats.
1x1 image pixel for data collection