More Filters
View all, or filter by any of the categories below to find resources that are relevant to you
Product Name
Description
The Gibco CTS DynaCellect Magnetic Separation System is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal. The instrument utilizes an integrated magnet-rocker and fluidics management for target-cell separation and subsequent bead removal in a closed system. The key design feature is the user programmable software, which enables design flexibility, performance precision, and scalability from process development to commercial manufacturing. Together with the fit-for-purpose, sterile, single-use consumables, it delivers high cell purity, recovery, and viability.
CTS DynaCellect bead removal kit Copy link
The Gibco CTS DynaCellect Bead Removal Kit is a single-use, fit-for-purpose kit designed for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal. This kit is used for depletion and magnetic bead removal.
CTS DynaCellect cell isolation kit Copy link
The Gibco CTS DynaCellect Cell Isolation Kit is a single-use, fit-for-purpose kit designed for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal. This kit is used for isolation and activation of immune cells in cell processing applications.
Gibco CTS Xenon Electroporation System Copy link
The Gibco CTS Xenon Electroporation System is a closed, modular, large-scale platform that offers full control over electroporation parameters for optimal performance. The CTS Xenon Electroporation System delivers high-performance nonviral transfection by enabling electroporation of up to 2.5 billion T cells in 25 mL per run for process development and manufacturing work in clinical cell therapy.
CTS Xenon Electroporation Buffer Copy link
Gibco CTS Xenon Electroporation Buffer is designed specifically for use with the Xenon Electroporation System. It is a universal buffer that supports the transfection of a variety human primary cells and payloads (e.g., plasmid DNA, mRNA, miRNA, or siRNA) for gene up-regulation or down-regulation applications.
CTS Xenon Genome Editing Buffer Copy link
Gibco CTS Xenon Genome Editing Buffer is designed specifically for use with the Xenon Electroporation System. It has been designed to improve performance with gene editing specific payloads (e.g., CRISPR Cas9) for knock-out- or knock-in-based applications in a variety of human primary cells.
CTS Xenon MultiShot Cartridge Copy link
The Gibco CTS Xenon MultiShot cartridge, designed for clinical manufacturing, enables consistent electroporation of volumes up to 25 mL in under 25 minutes in a closed, GMP-compliant system.
CTS Xenon SingleShot Cartridge Copy link
The Gibco CTS Xenon SingleShot Electroporation Chamber is designed for rapid electroporation of up to 1 mL of cells for cell therapy process development applications.
This versatile system applies the proven counterflow centrifugation method to a broad range of cell processing applications, such as CAR-T therapy, stem cell therapy, and PBMC isolation.
CTS Rotea System Single Use Kit Copy link
The CTS Rotea closed system single-use kits, media, and reagents offer process flexibility and can be customized to support commercial scale manufacture.