For Research Use Only. Not for use in diagnostic procedures.
Detect and differentiate from four to over 40 respiratory pathogens
Including bacteria, viruses, and fungi, across multiple sample types 
Testing methods
Looking to expand your molecular research menu for broader pathogens?
Targeted-Panels
Targeted panels
Targets: 2 to 9
Example: SARS-COV-2, Flu A, Flu B, RSV
Throughput Options
Syndromic panels
Targets: 10 to 30
Example: Viral and bacterial targets
For Research Use Only. Not for use in diagnostic procedures.
Need help finding the right solution?
Learn more about PCR testing formats like single tube and plates ›
Find out more about PCR molecular testing research products. Expand your molecular testing research menu using the PCR instrument you have.
Respiratory pathogens
Applied Biosystems real-time PCR respiratory research products
Real-time PCR products with high analytical sensitivity are needed to support laboratory research to study outbreaks and determine disease etiology where pathogens are similar.
Applied Biosystems real-time PCR respiratory research products are used to analyze a range of respiratory pathogens including viruses, bacteria, and fungi, with high analytical sensitivity. 
These solutions are efficient, generate informative results, and solve many of the challenges associated with the use of immunoassay-based, culture-based, and other molecular research methods.
Applied Biosystems TrueMark real-time PCR solutions are part of a sample-to-results workflow. Benefits include the ability to:
  • Detect and differentiate multiple pathogens
  • Obtain reliable real-time PCR results—Accurate, sensitive, and specific
  • Select from multiple format options to suit your testing capacity—Choose from single tube assays, 96- or 384-well plates, cards, or OpenArray plates
  • Order individual assays, predesigned panels, or customize your own—Select from a catalogue of >600 assays targeting pathogens or resistance genes
  • Reduce time-to-implementation—Accelerate and streamline your validation process with Analytical Validation Consulting Services
Introducing the TrueMark SARS-COV-2, Flu A, Flu B, RSV Select Panel
  • Research testing for the most common upper respiratory viruses co-circulating during the winter season
    • SARS-COV-2
    • Flu A
    • Flu B 
    • RSV
  • Enables detection of co-infections of two or more of the viral targets
  • Multi-target assay design to compensate for emerging mutations and variants and provide increased confidence in results 
  • Combo-kit ready: Includes assay, master mix, and recommended controls 
Flyer:TrueMark SARS-CoV-2, Flu A, Flu B, RSV ›
For Research Use Only. Not for use in diagnostic procedures.
Research solutions
Targeted panels 
Applied Biosystems TrueMark SARS-CoV-2, Flu A, Flu B, RSV Select Panel
Format: Single-tube multiplex
  • Enables detection of co-infections of two or more of the viral targets
  • Multi-target assay design to compensate for emerging mutations and variants and provide increased confidence in results
  • Combo-kit ready: Includes assay, master mix, and recommended controls
Targets
Viral
SARS-COV-2
Influenza A
Influenza B
Respiratory syncytial virus (RSV)
Applied Biosystems TrueMark Respiratory I Plus Panel
Format: Pre-spotted 96-well plates
  • Panel with eight targets offer an intermediate approach to testing 
  • Focus on viral-only or bacterial-only panels
  • Duplex reaction with internal RNase P control contributes to confidence in results
  • Expand menu with existing 96-well equipment
Targets
Viral
TrueMark Respiratory I Plus Panel
Adenovirus (AV)Influenza B
Enterovirus (EV)Respiratory syncytial virus A/B (RSV)
Human Metapneumovirus (hMPV)Rhinovirus (RV)
Influenza A (Pan)SARS-CoV-2
Kit size 
120 samples/10 pack
Throughput
12 samples/plate
Control(s)
RNase P internal control and available positive control
Sample type(s)
Nasopharyngeal swab
Turnaround time
~4 hours
Instrument compatibility
•QuantStudio 5 Real-time PCR System
•QuantStudio 7 Pro Real-time PCR System
•Other Applied Biosystems real-time PCR instruments on request
Software
Design & Analysis Software 
Catalog number
•TrueMark Respiratory I Plus Panel A56284C
•TrueMark Custom Panel Request a quote or consultation
Syndromic panels
Applied Biosystems TrueMark Respiratory Panel 2.0, TaqMan Array Card
Format: TaqMan Array Card
  • Fast, easy setup
  • Low risk of user error or contamination
  • Only requires eight samples to fill a card 
Targets
Bacteria
Bordetella (Pan)Haemophilus influenzaeMoraxella catarrhalisStreptococcus pneumoniae
Bordetella pertussisKlebsiella pneumoniae complexMycoplasma pneumoniae 
Chlamydia pneumoniaeLegionella pneumophilaStaphylococcus aureus 
FungalPneumocystis jirovecii   
Viral
AdenovirusHerpesvirus 6 (HHV6)Influenza BParechovirus
BocavirusHHV3 (varicella zoster virus)Measles virusRespiratory Syncytial Virus A (RSV A)
Coronavirus 229EHHV4 (Epstein-Barr virus)MERS-CoVRespiratory Syncytial Virus B (RSV B)
Coronavirus HKU1HHV5 (cytomegalovirus)Mumps virusRhinovirus
Coronavirus NL63Human Metapneumovirus (hMPV)Parainfluenza virus 1SARS-CoV (SARS)
Coronavirus OC43Influenza A (Pan)Parainfluenza virus 2SARS-CoV-2
Enterovirus D68Influenza A/H1-2009Parainfluenza virus 3 
Enterovirus PanInfluenza A/H3Parainfluenza virus 4 
Kit size 
80 samples/10 pack
Throughput
8 samples/card
Control(s)
B. atrophaeus, 18S, Xeno, RNase P, and available positive control
Sample type(s)
Nasopharyngeal swab, nasopharyngeal aspirate, and bronchoalveolar lavage (BAL) 
Turnaround time
~5 hours total
Instrument compatibility 
•QuantStudio 7 Real-Time PCR System
•QuantStudio 12K Flex Real-Time PCR System 
Software
Relative Quantification Application
Catalog number
A49047
Applied Biosystems TrueMark Respiratory Panel 2.0, OpenArray Plate
Format: OpenArray Plate
  • Lowest cost per pathogen for broad pathogen detection studies
  • Scalable, allowing for up to 288 samples per day 
  • Allows for simultaneous testing of multiple panel types by running up to four Applied Biosystems OpenArray plates per PCR run
Targets
Bacteria
Bordetella (Pan)Haemophilus influenzaeLegionella pneumophilaStaphylococcus aureus
Bordetella pertussisKlebsiella pneumoniae complexMycoplasma pneumoniaeStreptococcus pneumoniae
Chlamydia pneumoniae   
Viral
AdenovirusHerpesvirus 6 (HHV6)Influenza A/H3Respiratory Syncytial Virus A (RSV A)
Coronavirus 229EHHV4 (Epstein-Barr virus)Influenza BRespiratory Syncytial Virus B (RSV B)
Coronavirus HKU1HHV5 (cytomegalovirus)Parainfluenza virus 1Rhinovirus
Coronavirus NL63Human Metapneumovirus (hMPV)Parainfluenza virus 2SARS-CoV-2
Coronavirus OC43Influenza A (Pan)Parainfluenza virus 3 
Enterovirus D68Influenza A/H1-2009Parainfluenza virus 4 
Kit size
480 samples/10 pack
Throughput
24 samples/plate
Control(s)
B. atrophaeus, Xeno, RNase P, and available positive control
Sample type(s)
Bronchoalveolar lavage, nasopharyngeal swab, or nasopharyngeal aspirate samples
Turnaround time
~5 hours
Instrument compatibility
QuantStudio 12K Flex Real-time PCR System
Software
Relative Quantification Application
Catalog number
A49044
Custom solutions
Whether you want to modify from an existing design or start from scratch, we're here to walk you through the process.
Ask about our curated, flexible content panels
  • Targeted panels: 4–9 targets
  • Wet-lab tested with semi-inventoried content
  • Duplex reaction with process and amplification controls
  • Arrives pre-spotted and dried down in plate to reduce hands on time and user error
  • Available on 96- or 384-well plate formats
  Custom TrueMark Respiratory II Panel (A55017)
  • Viral targets: Coronavirus 229E (CE), Coronavirus HKU1 (CH), Coronavirus NL63 (CN), Coronavirus OC43 (CO), Parainfluenza 1 (P1), Parainfluenza 2 (P2), Parainfluenza 3 (P3), Parainfluenza 4 (P4)
  • Control type: RNase P internal control
Request a consultation or place an order ›
Custom TrueMark Respiratory III Panel (A55017)
  • Bacterial targets: C. pneumoniae, Moraxella catarrhalis, Klebsiella pneumonia, Staphylococcus aureus (SA), Haemophilus Influenzae (HI), Legionella pneumophila (LP), Mycoplasma pneumoniae (MP), Streptococcus pneumoniae (SP)
  • Control type: RNase P internal control
Request a consultation or place an order ›
Don't see what you're looking for?
We offer customization across all formats so you can create a tailored solution that meets your laboratory's throughput and business needs.
Learn more ›
Request consultation ›
PCR testing format comparison
Single tube multiplex
96-well plates
384-well plates
384-well microfluidic cards
OpenArray nanofluidic plates
Format
Single tube assay
Duplex (target and internal control) TaqMan assays, pre-spotted
Singleplex TaqMan assays, pre-spotted
Number of assays
1 to 5
4 and 8
12 and 24
24 or 48
18, 37, or 56
Number of samples
1–384
12–24
16–32
1–8
12–48
Reaction volume
10 μL, 20 μL
10 μL, 20 μL
10 μL
1 μL
33 nL
Instrument compatibility
Applied Biosystems 7500 Real-time PCR System
Any QuantStudio instrument
QuantStudio 5 Real-Time PCR System
QuantStudio 12K Flex Real-Time PCR System
QuantStudio 7 Pro Real-Time PCR System
Other instruments available on request
Features and benefits
Flexibility in sample throughput; load wells only as needed
Panels with eight targets offer an intermediate approach to testing
Fast, easy setup
Lowest cost per pathogen for broad pathogen detection studies
Complete your infectious disease workflow with a single kit for DNA and RNA extraction
Maximize your nucleic acid yield for greater assay sensitivity from a range of biological specimens for infectious disease applications with the MagMAX Prime Viral/Pathogen NA Isolation Kit.
Learn more ›
MagMAX Prime Viral/Pathogen NA Isolation Kit