Samco Transfer Pipettes
The Thermo Scientific Samco Transfer Pipette is a single use microliter volume liquid collection tool with an innovative design that utilizes capillary action to aspirate to a specific, preset volume, facilitating one-handed operation. Transfer pipettes provide higher inter-day reproducibility for both single and multiple users and complete dispensing of the sample.
Select from a range of transfer pipettes to fit your application
TypeOptionsApplicationsRecommended UseCatalog
General Purpose
Bulb draw (mL):
0.3, 0.9, 1.8, 2.2, 2.4, 2.8, 3.0, 4.7, 6.2, 7.4
Rapid transfers,  controlled drop size, calibrated transfersLarge aperature for creams and gels, oils, industrial slurries, paints, perfumesBuy
Graduated 
Bulb draw (mL):
0.8, 1.8, 2.1, 3.2, 3.5, 4.3, 7.4
Remove and separate liquids, dispense small dropsIdeal for measured transfers, drop by drop work, aliqoutingBuy
Narrow Stem
Stem diameter (mm):
2.5, 3.0, 8.0
Genomics and potemoics companies, research labs–reagent prep, drop-wise additions to culture platesLoading gels, PCR work, DNA sample removal, feed hybridomas, lipid extractions, atomic absorption, spectroscopy Buy
Fine Tip
Stem diameter (mm):
1.2, 3.0, 4.6, 6.1
Sample preparation, layering in columns, transfers in columns, loading gelsLoading gels, PCR, lipid extractions, DNA samples removal, protein assaysBuy
Special Purpose
Bulb draw (mL):
0.2, 3.3
Assay development, research labsGreat for viscous fluids, paddle for mixingBuy
Exra Long
Length:
9in (22.5cm), 12in  (30.0cm)
Assay development, research labsSub-culturing and harvesting cells and tissues; Transferring from large bottles; Removal of supernatant fluids from centrifugationBuy
Exact Volume*
Dispense volume (μL):
20, 40, 50, 60, 80, 100, 150, 200, 250, 300, 400 µL 
Assay development, research in microbiology, agriculture, veterinary, food
Exact volume dispensing, analytical testing, pathogen testingBuy
Capillary 
Volume (µL):
25, 50 
Liquid transfer, rapid tests, lateral flow assay developmentLiquid transfer, point of care tests, rapid tests, lateral flow assay developmentBuy
*Note: Exact Volume is the name of this group of transfer pipettes. These conveniently dispense a precise volume within a tolerance.
Learn more about transfer pipettes by watching videos
Thermo Scientific Samco Pipettes Drop Size Matters
Prevent cross-contamination with Thermo Scientific™ Samco™ General-Purpose Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving.
Video Player is loading.
Current Time 0:00
Duration 1:00
Loaded: 0%
Stream Type LIVE
Remaining Time 1:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextThermo Scientific Samco Pipettes Drop Size Matters
        Prevent cross-contamination with Thermo Scientific™ Samco™ General-Purpose Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving.
      2. Now Playing
        Up NextThermo Scientific Samco Exact Volume Transfer Pipettes
        Perform precise dispensing of volumes from 20 to 400μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene.
      3. Now Playing
        Up NextThermo Scientific Samco Extra Long Transfer Pipettes
        Perform safe and easy sampling from large containers using Thermo Scientific™ Samco™ Extra-Long Transfer Pipettes. Perfect for tissue culture applications.
      4. Now Playing
        Up NextThermo Scientific Samco Fine Tip Transfer Pipettes
        Perform precise dispensing of small drops into small-volume containers using Thermo Scientific™ Samco™ Fine-Tip Transfer Pipettes. These fine-tip pipettes are ideal for removing cell layers and separating blood samples.
      5. Now Playing
        Up NextThermo Scientific Samco Narrow Stem Transfer Pipettes
        Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Narrow Stem Transfer Pipettes. These polyethylene pipettes prevent cross-contamination and are used for blood bank applications such as sedimentation procedures.
      6. Now Playing
        Up NextSpecial Purpose Thermo Scientific Padl-Pet Pipettes
        Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles.
      7. Now Playing
        Up NextThermo Scientific Samco Special Purpose Slurry Pipettes
        Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles.
      8. Now Playing
        Up NextThermo Scientific Samco Graduated Transfer Pipettes
        Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, autoclaving while preventing cross-contamination
      9. Now Playing
        Up NextThermo Scientific Samco Transfer Pipettes
        Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.