TCAutomation Laboratory Automation System
Efficiency and consistency in sample handling
Expandable automation to streamline lab processes
Improved efficiency and safety of sample management together with short and consistent turnaround times are the key benefits of laboratory automation. By automating the key pre- and post-analytical steps, such as decapping, centrifugation, aliquoting, sorting, recapping, storage and retrieval, Thermo Scientific™ TCAutomation™ helps to manage the increasing workloads and improve quality in clinical laboratories.
Contact us to speak with a distributor
Flexible, open and scalable system
TCAutomation system provides a solution for increased throughput needs and for integration of new analyzing technologies.
Each installation is planned for its individual optimal efficiency delivering improved sample management.
  • TCAutomation systems fit into various floor plans.
  • The modular concepts are scalable to meet the changing needs.
  • TCAutomation system can grow from a small automation to high volume total laboratory automation solution.
TCAutomation system supports analyzer interfaces from various different manufactures. Workcell solutions may include two or more system combination increasing the analyzer capacity and decreasing the manual work.
  • Single entry point without pre-sorting.
  • One centralized user interface offers integrated functionality for managing the whole workcell.
  • Easy addition of pre- and post-analytical functionality.
Watch TCAutomation system in action
TCAutomation BES Flex - Fully automated sample input and sorter system
TCAutomation BES Flex is a fully automated sample input and sorter system for batches of securely closed sample tubes.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextTCAutomation BES Flex - Fully automated sample input and sorter system
          TCAutomation BES Flex is a fully automated sample input and sorter system for batches of securely closed sample tubes.
        2. Now Playing
          Up NextTailor-made TCAutomation Systems manufactured to fit into even the most challenging laboratories
          Every ordered TCAutomation System undergoes several test procedures to ensure reliability. At customer laboratory this ensures short installation time.
        3. Now Playing
          Up NextThermo Scientific TCAutomation - Open and Expandable Laboratory Automation Solution
          TCAutomation solution manages clinical laboratories workloads by automating pre- and post-analytical steps and integrating various manufactures analyzers.
        4. Now Playing
          Up NextTCAutomation Conveyors
          Dual lane conveyors in different lengths for sample transport. Each system includes at least two U-Turns. L-, T-, Z- and internal U-Turns also available.
        5. Now Playing
          Up NextTCAutomation Cold Storage System
          Cold Storage is a refrigerated storage system from which samples can be automatically retrieved back to the track for re-testing or discharged according to the programmable timing. Storage capacity for up to 14 800 tubes.
        6. Now Playing
          Up NextTCAutomation Centrifuge Module
          Module for automatic and continuous centrifugation of samples. Automates loading, balancing and unloading of centrifuge racks.
        7. Now Playing
          Up NextTCAutomation Decapper Flex Module
          Module for automatic cap removal from different types and sizes of sample tubes.
        8. Now Playing
          Up NextTCAutomation Recapper Module
          Module for automatic capping of different types and sizes of primary and TCAutomation Aliquoter secondary tubes with conical caps.
        9. Now Playing
          Up NextTCAutomation Aliquoter & Labeler Module
          Module for automatic sample aliquoting to secondary tubes using disposable tips. Automatic barcode labeling of secondary tubes.
        10. Now Playing
          Up NextTCAutomation ES Flex Module
          ES Flex is a module for automatic sample loading and unloading. Module has automatic retrieval for rerun and excellent sample sorting capabilities.
      Labor-intensive tasks can be automated in different combinations
      TCAutomation ES Flex, Entry and Exit Modules
      ES Flex, Entry and Exit Modules - Modules for automatic or manual sample loading and unloading in user-specified racks. In ES Flex automatic retrieval for rerun and excellent sample sorting capabilities.
      TCAutomation Aliquoter/Labeler Module
      Aliquoter/Labeler Module – Automatic sample aliquoting to secondary tubes using disposable tips. Automatic barcode labeling of secondary tubes.
      TCAutomation Centrifuge Module
      Centrifuge Module – Automatic and continuous centrifugation of samples.
      TCAutomation Decapper Flex Module
      Decapper Flex Module – Automatic cap removal from different types and sizes of sample tubes.
      TCAutomation Recapper Module
      Recapper Module – Automatic capping of different types and sizes of primary and TCAutomation Aliquoter secondary tubes with conical caps.
      TCAutomation Buffer Module
      Buffer Module – Area for samples waiting for validation and for storing empty carriers.
      TCAutomation Cold Storage Module
      Cold Storage Module – Refrigerated storage module from which samples can be automatically retrieved back to the track for re-testing or discharged.
      TCAutomation Perpendicular and Parallel Bypass Modules
      Perpendicular and Parallel Bypass Modules – Analyzer- specific interface modules for point-of-space system connections.
      TCAutomation Robotic Analyzer Interfaces
      Robotic Analyzer Interfaces – Please contact Thermo Fisher Scientific for more information.
      Fully featured solutions for optimized workflow
      Effective sample processing
      Samples are transported within the TCAutomation system in single tube carriers that accommodate various tube types and sizes.
      • Sample tube carriers with an embedded RFID microchip enable faster and more reliable sample identification.
      • Excellent real-time sample tracking possibilities.
      Centralized operation
      TCAutomation controller, based on Windows™ software, offers in a single screen an overview of the entire system.
      • Controller allows users an easy way to monitor large automation configurations.
      • Route definitions are simple to manage and provide flexibility when optimizing the workflow.
      Expandability step by step
      Depending on the floor plan and efficiency requirements, TCAutomation systems can be expanded step by step by adding modules.
      • Expand functionality with additional modules
      • Increase throughput by multiple modules
      • Extend analytical integration by adding analyzer interfaces