Next-generation laboratory software for increased productivity and compliance across your business 
Thermo Scientific SampleManager LIMS Software version 21.3 is a powerful laboratory software designed to streamline workflows and eliminate manual tasks. It offers a range of features and improvements to enhance your organization's productivity and efficiency. The solution provides a safer, more secure software system that supports your scientific work, optimizes laboratory processes, enhances data analysis, and improves collaboration between different software systems.
Macbook displaying SampleManager LIMS software surrounded by yellow circle with grey background.
Dashboard shown is available in The Data Analytics Solution for SampleManager LIMS Software
SampleManager LIMS Software version 21.3 brings integration improvements for better efficiency through updated features, architecture, and product health.
Key updates include enhanced integration capabilities with support for SAP remote function calls, an upgraded Sweeper for handling complex files, and streamlined resource management for seamless task assignment. The software also offers improved data security and architectural enhancements for robust and reliable platform performance.
SampleManager LIMS Software version 21.3 capabilities
Advanced integration for efficient data management
  • Enhanced SMIDI: Now supports SAP remote function calls, enabling smooth and efficient interfacing with the SAP QM module and other SAP systems. 
  • Sweeper enhancement: Capable of picking up complex files like PDFs and binary files, ensuring comprehensive file integration and improving data processing. 
  • Resource management: Allows seamless creation and assignment of tasks, integrating personnel, location, and instrument management for streamlined workflows.
  • SampleManager - Chromeleon Link: Unifies data across the lab by integrating SampleManager LIMS with Thermo Scientific Chromeleon CDS Software, removing duplicated work and errors for seamless data flow, faster turnaround, improved accuracy, and regulatory compliance.
Scientist standing at a computer, using SampleManager 21.3 LIMS software.
Robust architecture and enhanced security
  • Enhanced architecture: Built on the reliable Microsoft.NET framework, continuous improvements ensure a stable and powerful system architecture. 
  • Improved security: Features such as token authentication and hardened session handling for the REST API provide robust data protection and secure integrations.
Female scientist sitting down using a computer with two scientists working in the background.
Improved workflow and data handling
  • Handling unrelated samples: Offers the ability to select and manage data based on custom criteria, enabling workflows to process diverse data sources effectively and access contextual information quickly. 
  • Attachments integration: Simplifies data import by integrating and extracting data directly from attachments, such as calibrated instrument data, eliminating multiple steps. 
  • Language deployer: Enhances localization by allowing the export and import of language kits across system instances, making interface customization straightforward. 
  • AQC enhancements: Creates a robust quality control process as set out in support of ASTM standard D6299.
Scientist pointing at a computer screen whilst a colleague looks on holding a tablet.
SampleManager LIMS Software core features
SampleManager LIMS Software is a highly configurable solution for managing laboratory operations, data, and processes. With over three decades of experience and a strong global presence, it is trusted by customers worldwide. Developed in an ISO 9001 compliant environment, the software drives reliability and quality. Its key strength lies in its adaptability, allowing it to be easily tailored to meet the specific needs of your organization. This eliminates the need for costly and time-consuming customization.
Resources
For Laboratory Use.
1x1 image pixel for data collection1x1 image pixel for data collection