‹ Back to learning center
View our on-demand webinars below.
Video Player is loading.
Current Time 0:00
Duration 40:03
Loaded: 0.41%
Stream Type LIVE
Remaining Time 40:03
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Studying small viral glycoproteins using an integrative structural biology approach
    Recent advances in single-particle cryo-electron microscopy (cryo-EM) have enabled structure determination of small (<200 kDa) biological complexes, allowing researchers to study proteins that resist crystallization. Dr. Joost Snijder, Assistant Professor, Utrecht University, and Dr. Ieva Drulyte, Cryo-EM Application Specialist, Thermo Fisher Scientific, present the cryo-EM structure of the ~80 kDa heavily glycosylated human coronavirus HKU1 hemagglutinin esterase (HE) at a global resolution of 3.4 Å.
    Video Player is loading.
    Current Time 0:00
    Duration 54:58
    Loaded: 0.30%
    Stream Type LIVE
    Remaining Time 54:58
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Native mass spectrometry-based screening for optimal sample preparation in single particle cryo-EM
      In this webinar, Dr. Dom Olinares discusses recent advances in single particle cryogenic electron microscopy (cryo-EM) have enabled the structural determination of numerous flexible and conformationally heterogeneous protein assemblies at high resolution, yielding unprecedented insights into their function. However, despite its extraordinary capabilities, cryo-EM remains extremely resource-intensive and time-consuming. It is therefore beneficial to have a means for rapidly assessing and optimizing the quality of samples prior to lengthy cryo-EM analyses.
      Video Player is loading.
      Current Time 0:00
      Duration 10:59
      Loaded: 1.49%
      Stream Type LIVE
      Remaining Time 10:59
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Integrative Structural Biology Going Viral
        Learn about integrative structural biology for viral research by watching this 10 minute video. Albert Konijnenberg, PhD, Product Manager, cryo-EM Sample Preparation & Integrative Structural Biology, Thermo Fisher Scientific, discusses the role of cryo-electron microscopy (cryo-EM) in vaccine development and as an assay for mutants. Learn how native and charge detection mass spectrometry can be used for Adeno-associated virus (AAV) particle characterization. Find out how combining structures from cryo-EM with glycan characterization by mass spectrometry can help scientists understand the bodies’ immune response to SARS-CoV-2.
        Contact us
        Style Sheet for Support and Service footer
        Style Sheet for Fonts
        Style Sheet for Cards