Style Sheet for Fonts
Style Sheet for Instrument Cards Original
Style Sheet for Global Design System
Thermo Fisher Scientific Electron Microscope Upgrade Program
Regularly upgrading your microscope allows you to enhance the capabilities of current system and utilize the instrument in the most effective and productive way possible. Microscope upgrades can expand the range of applications your instrument can accommodate and helps you maintain your competitive edge. Investing in upgrades not only extend the equipment lifetime but also assures that your microscope will continue to serve your changing microscopic analysis needs reliably for years to come.
Upgrades for the Thermo Fisher Scientific TEM, SEM, and Dual Beam FIB SEM instruments, providing you with the latest developments and capabilities for your instrument.
Whether it is time consuming frequent alignment of current equipment, limitation on image resolution and image processing for advanced materials research or time-to-result, Our continuously evolving automation features, detection enhancement and microscope software upgrades will lead to a superior microcopy experience for every researcher.
Accessories for Electron Microscopes
Thermo Scientific Falcon 4i Electron Detector
Falcon 4i Detector
  • High throughput for more images per hour
  • Unsurpassed imaging quality with high DQE
  • Lossless data compression with EER
μHeater
  • Ultra-fast heating solution for in situ high resolution imaging
  • Fully integrated
  • Temperatures up to 1200 °C
μPolisher
  • Potential to enable large number of novel, unexplored applications
  • Very low energy milling
  • Small spot size for precise local surface treatment
Thermo Scientific Ceta-D camera for electron diffraction
Ceta-D Camera
  • Optimum performance at any high tension (20–300 kV)
  • Compatible with post-column filters and spectrometers
  • Movie acquisition for dynamic studies
Thermo Scientific Selectris and Selectris X cryo electron microscopy imaging filters
Selectris and Selectris X Imaging Filters
  • Designed for high stability and atomic resolution imaging
  • Straightforward operation
  • Paired with the latest generation Thermo Scientific Falcon 4i Direct Electron Detector
Accessories for Phenom Desktop Scanning Electron Microscopes
Holders
Charge Reduction Sample Holder
  • Up to 8 times higher magnification
  • Faster sample preparation
  • Non conductive samples can be imaged in their natural state
Electrical Feedthrough Sample Holder
  • Connect electrical probes to the sample for in situ measurements
  • Sample can be height adjusted from 0–25 mm manually
  • Measurements of probe currents
Eucentric Sample Holder
  • Eucentric tilting and compucentric rotation on a desktop SEM
  • Fast time-to-image with sample loading < 1 minute
  • Real-time 3D sample visualization module
Metallurgical Sample Holder
  • Designed to support resin-mounted samples
  • Preferred solution for metallurgy and when working with inserts
  • Sample size up to 32 mm diameter and 30 mm height
Motorized Tilt & Rotation Sample Holder
  • Tilt range -10° to +45°
  • Continuous 360° compucentric rotation
  • Controlled by dedicated Motion Control ProSuite application
Standard Sample Holder
  • Compact stage allowing analysis of samples of up to 100 mm x 100 mm
  • Can be extended with 3 types of resin or metallurgical mount inserts
  • Used with Phenom Desktop SEM
Micro Tool Sample Holder
  • Quick and fast clamping
  • Tilting and rotation allow for easy sample positioning
  • No extra tooling required for sample loading
Nebula Particle Disperser
  • Standard method for uniform dry powder dispersion
  • Avoids particle clusters
  • Used with Phenom Desktop SEM
Inserts
Filter Inserts
  • Filter residue analysis and asbestos analysis
  • Available in two models which support 47 mm (1.85 inch) and 25 mm (1 inch) filters
  • Use on Phenom Desktop SEM
Resin Mount Inserts
  • A unique sample holder concept
  • Available in 3 models for supporting standard sized samples of 25 mm (~1 inch), 32 mm (~1 ¼ inch) and 40 mm (~1 ½ inch) diameter
Sample Holder Inserts
  • Faster cross-sectional imaging of coatings and multilayer samples
  • Easy clamping without the need for screws or extra tools
  • Eliminates the need for screws and tools to clamp the sample
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards