Quickly and accurately understand your fibrous materials
Fibrous materials, such as carbon or glass fiber (CFRP, GFRP), fabric (multi-layer composites), fiber-reinforced concrete (FRC), and organic fibers are used in a wide range of applications and industries (automotive, aerospace, etc.). Imaging techniques such as microCT or SEM allow for analysis of those materials; for example, to detect manufacturing imperfections or to quantify microstructures to obtain insights on mechanical properties (strength, stiffness, etc.).
Thermo Scientific Avizo Software is an all-in-one image analysis platform that allows the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage such as matrix cracking, fiber/matrix debonding, or fiber breakage. Imperfections in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignments).
Imaging data
Workflow using Avizo Software
Processing
Workflow using Avizo Software
Visualization
Workflow using Avizo Software
Analysis
Workflow using Avizo Software
Customization
Confidence
Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to industrial and scientific problems.
Customization
Customization
Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
icon-improve_collaboration_64x64
Support
Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
Software_Automation_65x64
Automation
Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency. 
Use cases
Fiber orientation analysis of a 50% glass fiber-reinforced polymer (GFRP) based on a combination of semi-crystalline polyamide with partially aromatic copolyamide
Many industries face the challenge of designing lightweight materials. Sandwich-structured composites are a special class of composite materials with the typical features of low weight, high stiffness, and high strength. The fibers of such material enhance properties of the final part (strength and stiffness) while minimizing weight. Their distribution and direction are crucial factors for the mechanical properties of the final part.
Avizo Software enables analysis of the internal fiber structure to improve these properties and the product development process. Fiber length distribution, diameter distribution, and through thickness orientation variation can be computed.
Royal DSM uses Avizo Software for glass fiber length and orientation analysis of reinforced polymers (GFRP).
avizo fiber royaldsm
Courtesy of EMS Grivory
3D fiber reconstruction in fiber reinforced concrete
Fibers are used in concrete to increase its structural integrity. The characteristics of fiber-reinforced concrete change with varying fiber material geometries, distributions, orientations, and densities.
Avizo Software enables characterization of the tensile strength of the FRC by identifying the fibers and providing their average orientation. Further quantification is performed to analyze porosity distribution in the concrete, based on pore network modeling techniques.
Video Player is loading.
Current Time 0:00
Duration 1:02
Loaded: 15.79%
Stream Type LIVE
Remaining Time 1:02
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Courtesy of EMPA
    Manufacturing defect analysis of a bike frame carbon fiber reinforced polymer (CFRP)
    Voids concentrate stress points in materials and can initiate cracks in composite materials. Therefore, volume fraction information and other microstructural characteristics such as spatial distribution and aspect ratio are of high value.
    In this example, the correlation between matrix cracks and fiber density is analyzed. Fiber density is mapped on the segmented model. Density is low around matrix cracks (voids). Visual inspection allows for verification that voids are smaller than the gaps they create between fibers.
    Video Player is loading.
    Current Time 0:00
    Duration 0:09
    Loaded: 100.00%
    Stream Type LIVE
    Remaining Time 0:09
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Data courtesy of Rigaku Corporation
      Evolution of damage during the fatigue of 3D woven glass-fibre reinforced composites subjected to tension-tension loading observed by time-lapse X-ray tomography
      Evaluation of material performance is of high importance. The ability to predict where damage occurs and how it evolves is critical in many industries. The nature of damage evolution captured in this study helps guide the design of a 3D composite with more resistance to fatigue damage.
      Read the research paper 
      Avizo fiber composite
      Courtesy of Henry Moseley X-ray Imaging Facility, School of Materials, University of Manchester
      Fertilizer influence on root growth
      With the aim to prevent global food security issues, optimization of the design of fertilizer granules for plant breeding and soil quality enhancement is an important topic of research. MicroCT scans of the plant from germination to maturity allow for visualization and analysis of the interactions of root and fertilizer to inform the design process.
      However, segmenting roots from X-ray tomography data is highly challenging for plants such as wheat, due to their fine roots and a density range similar to pore-water and organic matter found in the surrounding soil. The powerful XFiber extension of Avizo Software enables correlation of roots based on the greyscale information and automatically classifies them. Root length and diameter can then be computed to assess mean growth root rate per fertilizer.
      Read the research paper 
      Learn more
      Video Player is loading.
      Current Time 0:00
      Duration 1:08
      Loaded: 14.30%
      Stream Type LIVE
      Remaining Time 1:08
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Courtesy of Sharif Ahmed, µ-VIS X-Ray Imaging Centre, Faculty of Engineering and the Environment University of Southampton, Southampton, UK
        Fiber orientation analysis of a 50% glass fiber-reinforced polymer (GFRP) based on a combination of semi-crystalline polyamide with partially aromatic copolyamide
        Many industries face the challenge of designing lightweight materials. Sandwich-structured composites are a special class of composite materials with the typical features of low weight, high stiffness, and high strength. The fibers of such material enhance properties of the final part (strength and stiffness) while minimizing weight. Their distribution and direction are crucial factors for the mechanical properties of the final part.
        Avizo Software enables analysis of the internal fiber structure to improve these properties and the product development process. Fiber length distribution, diameter distribution, and through thickness orientation variation can be computed.
        Royal DSM uses Avizo Software for glass fiber length and orientation analysis of reinforced polymers (GFRP).
        avizo fiber royaldsm
        Courtesy of EMS Grivory
        3D fiber reconstruction in fiber reinforced concrete
        Fibers are used in concrete to increase its structural integrity. The characteristics of fiber-reinforced concrete change with varying fiber material geometries, distributions, orientations, and densities.
        Avizo Software enables characterization of the tensile strength of the FRC by identifying the fibers and providing their average orientation. Further quantification is performed to analyze porosity distribution in the concrete, based on pore network modeling techniques.
        Video Player is loading.
        Current Time 0:00
        Duration 1:02
        Loaded: 15.79%
        Stream Type LIVE
        Remaining Time 1:02
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Courtesy of EMPA
          Manufacturing defect analysis of a bike frame carbon fiber reinforced polymer (CFRP)
          Voids concentrate stress points in materials and can initiate cracks in composite materials. Therefore, volume fraction information and other microstructural characteristics such as spatial distribution and aspect ratio are of high value.
          In this example, the correlation between matrix cracks and fiber density is analyzed. Fiber density is mapped on the segmented model. Density is low around matrix cracks (voids). Visual inspection allows for verification that voids are smaller than the gaps they create between fibers.
          Video Player is loading.
          Current Time 0:00
          Duration 0:09
          Loaded: 100.00%
          Stream Type LIVE
          Remaining Time 0:09
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            Data courtesy of Rigaku Corporation
            Evolution of damage during the fatigue of 3D woven glass-fibre reinforced composites subjected to tension-tension loading observed by time-lapse X-ray tomography
            Evaluation of material performance is of high importance. The ability to predict where damage occurs and how it evolves is critical in many industries. The nature of damage evolution captured in this study helps guide the design of a 3D composite with more resistance to fatigue damage.
            Read the research paper 
            Avizo fiber composite
            Courtesy of Henry Moseley X-ray Imaging Facility, School of Materials, University of Manchester
            Fertilizer influence on root growth
            With the aim to prevent global food security issues, optimization of the design of fertilizer granules for plant breeding and soil quality enhancement is an important topic of research. MicroCT scans of the plant from germination to maturity allow for visualization and analysis of the interactions of root and fertilizer to inform the design process.
            However, segmenting roots from X-ray tomography data is highly challenging for plants such as wheat, due to their fine roots and a density range similar to pore-water and organic matter found in the surrounding soil. The powerful XFiber extension of Avizo Software enables correlation of roots based on the greyscale information and automatically classifies them. Root length and diameter can then be computed to assess mean growth root rate per fertilizer.
            Read the research paper 
            Learn more
            Video Player is loading.
            Current Time 0:00
            Duration 1:08
            Loaded: 14.30%
            Stream Type LIVE
            Remaining Time 1:08
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Courtesy of Sharif Ahmed, µ-VIS X-Ray Imaging Centre, Faculty of Engineering and the Environment University of Southampton, Southampton, UK
              Services
              Amira-Avizo-Introductory-training_1160x600
              Introductory training
              Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
              The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
              Amira-Avizo-advanced-training_1160x600
              Advanced training
              Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
              The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
              Amira-Avizo-Custom-Dev_1160x600
              Custom development
              With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
              We can customize and expand our software solutions at various levels.
              Resources
              Video Player is loading.
              Current Time 0:00
              Duration 1:02
              Loaded: 15.79%
              Stream Type LIVE
              Remaining Time 1:02
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Avizo for Fiber Analysis - 3D fiber reconstruction in fiber reinforced concrete.
                Video Player is loading.
                Current Time 0:00
                Duration 2:05
                Loaded: 7.86%
                Stream Type LIVE
                Remaining Time 2:05
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  Study on CFRP composites used in the aviation industry.
                  Video Player is loading.
                  Current Time 0:00
                  Duration 0:05
                  Loaded: 100.00%
                  Stream Type LIVE
                  Remaining Time 0:05
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • default, selected
                    Fiber characterization and orientation analysis in CFRP with Avizo Software.
                    Video Player is loading.
                    Current Time 0:00
                    Duration 1:08
                    Loaded: 14.30%
                    Stream Type LIVE
                    Remaining Time 1:08
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      Avizo fiber fertilizer influence on root growth.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 1:07
                      Loaded: 14.53%
                      Stream Type LIVE
                      Remaining Time 1:07
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • en (Main), selected
                        Surgical mask fibers visualization and analysis with Avizo Software.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 1:02
                        Loaded: 15.79%
                        Stream Type LIVE
                        Remaining Time 1:02
                        1x
                          • Chapters
                          • descriptions off, selected
                          • captions off, selected
                          • en (Main), selected
                          Avizo for Fiber Analysis - 3D fiber reconstruction in fiber reinforced concrete.
                          Video Player is loading.
                          Current Time 0:00
                          Duration 2:05
                          Loaded: 7.92%
                          Stream Type LIVE
                          Remaining Time 2:05
                          1x
                            • Chapters
                            • descriptions off, selected
                            • captions off, selected
                            • en (Main), selected
                            Study on CFRP composites used in the aviation industry.
                            Video Player is loading.
                            Current Time 0:00
                            Duration 0:05
                            Loaded: 100.00%
                            Stream Type LIVE
                            Remaining Time 0:05
                            1x
                              • Chapters
                              • descriptions off, selected
                              • captions off, selected
                              • default, selected
                              Fiber characterization and orientation analysis in CFRP with Avizo Software.
                              Video Player is loading.
                              Current Time 0:00
                              Duration 1:08
                              Loaded: 14.30%
                              Stream Type LIVE
                              Remaining Time 1:08
                              1x
                                • Chapters
                                • descriptions off, selected
                                • captions off, selected
                                • en (Main), selected
                                Avizo fiber fertilizer influence on root growth.
                                Video Player is loading.
                                Current Time 0:00
                                Duration 1:07
                                Loaded: 14.53%
                                Stream Type LIVE
                                Remaining Time 1:07
                                1x
                                  • Chapters
                                  • descriptions off, selected
                                  • captions off, selected
                                  • en (Main), selected
                                  Surgical mask fibers visualization and analysis with Avizo Software.
                                  The solution of choice for fiber analysis
                                  Avizo Software provides dedicated tools for fiber analysis:
                                  • Automatic segmentation of individual high-resolution fibers
                                  • Major orientations of low-resolution fibers
                                  • Quantification such as contact point or distance to closest neighbor
                                  • Fiber shape statistics (diameter, length, tortuosity, area, volume fraction, cross section perimeter, and area)
                                  • Statistics relative to the fibers' density (volume fraction, area, and tensor per material subdomain)
                                  • 3D plot of fiber's orientation on a sphere or half-sphere with properties mapped as color and height
                                  • Advanced fiber filtering modules
                                  Style Sheet for Instrument Cards Original
                                  Contact us
                                  Style Sheet for Global Design System
                                  Style Sheet for Komodo Tabs
                                  Style Sheet for Support and Service footer
                                  Style Sheet for Fonts
                                  Style Sheet for Cards