c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Spectra 300 Transmission Electron Microscope
For scientists to advance their understanding of complex samples and develop innovative materials, they must have access to robust, precise instrumentation capable of correlating form and function, as well as resolving space, time and frequency.
Thermo Fisher Scientific introduces the Thermo Scientific Spectra 300 (S)TEM – the highest resolution, aberration corrected, scanning transmission electron microscope for all materials science applications.
Spectra 300 Transmission Electron Microscope advantages
All Spectra 300 (S)TEM's are delivered on new platforms designed to offer an unprecedented level of mechanical stability and highest imaging quality though passive and (optional) active vibration isolation.
The system is housed in a fully redesigned enclosure with a built-in on-screen display for convenient specimen loading and removal. For the first time, full modularity and upgradeability can be offered between uncorrected and single-corrected configurations with variable heights, allowing maximum flexibility for different room configurations.
Video Player is loading.
Current Time 0:00
Duration 0:58
Loaded: 17.23%
Stream Type LIVE
Remaining Time 0:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Spectra 300 (S)TEM: A redesigned base and enclosure deliver the highest imaging quality with integrated passive or active vibration isolation configuration options.
    Spectra 300 Transmission Electron Microscope features
    The Spectra 300 (S)TEM can be optionally equipped with either a high-energy-resolution extreme field emission gun (X-FEG)/Mono or an ultra-high-energy resolution X-FEG/UltiMono. The monochromators of both sources are automatically excited and tuned with single-click operation to achieve the highest energy resolution possible on each configuration by using OptiMono or OptiMono+ respectively (see video below).
    The X-FEG/Mono can be automatically tuned from 1 eV down to 0.2 eV, while the X-FEG/UltiMono can be automatically tuned from 1 eV down to <30 meV.
    Both sources can be operated from 30 – 300 kV to accommodate the widest range of specimens. Both can also be run in standard mode, with the monochromators switched off, to accommodate experiments that require high brightness, including STEM EDS mapping, ultra-high-resolution STEM, or high total current, such as TEM imaging, all with no compromise to the other specifications of the system. This flexibility gives the Spectra 300 (S)TEM the capability to function in settings where a large range of experiments are expected to be performed on one system.
    Video Player is loading.
    Current Time 0:00
    Duration 2:18
    Loaded: 7.20%
    Stream Type LIVE
    Remaining Time 2:18
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      OptiMono+ exciting an X-FEG/UltiMono from the monochromator off state (with 1 eV energy resolution) to the monochromator fully excited state (<30 meV) at 60 kV. 
      The Spectra 300 (S)TEM can optionally be powered by a new cold field emission gun (X-CFEG). The X-CFEG has extremely high brightness (>>1.0 x 108 A/m2/Sr/V*), low energy spread (<0.4 eV), and can operate from 30 – 300 kV. This provides simultaneously high-resolution STEM imaging with high probe currents for high throughput, fast acquisition STEM analytics in parallel with high-energy resolution. With the powerful combination of X-CFEG and the S-CORR probe aberration corrector, sub-Angstrom (<0.8 Å) STEM-imaging resolution with over 1000 pA of probe current can be routinely achieved.
      High-angle annular dark-field (HAADF) images of silicon.
      Si[110] HAADF images taken with the X-CFEG/S-CORR combination; probe currents range from 0.016 nA (left) up to 1 nA (right) while maintaining <76 pm STEM resolution.
      Further, probe currents can be flexibly tuned from <1 pA up to the nA range with fine control of the gun and condenser optics, all with minimum impact on the probe aberrations, so that the widest range of specimens and experiments can be accommodated.
      As with all cold field emission sources, the sharp tip requires a periodic regeneration (called flashing) to maintain the probe current. With the X-CFEG, the tip only requires flashing once per working day, a process takes less than a minute. There is no measurable impact on the probe aberrations even in the highest resolution imaging conditions, and the daily tip flashing process has no impact on the tip lifetime.
      Video Player is loading.
      Current Time 0:00
      Duration 0:39
      Loaded: 25.02%
      Stream Type LIVE
      Remaining Time 0:39
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Tip flashing on the X-CFEG: 60 pm resolution at 200 kV is maintained before and after tip flashing without adjustment of the optics. The process takes <1 min, is required only once per working day, and has no impact on the lifetime of the tip.
        This new generation X-CFEG also produces enough total beam current (>14 nA) to support standard TEM imaging experiments (e.g. in situ) with large parallel probes, making it a uniquely all-purpose, yet high performance, C-FEG.
        Adding to the flexible nature of the X-CFEG is the capability to adjust the energy resolution by varying the extraction voltage.
        In the example below, the energy resolution was adjusted between 0.39eV, with <500pA of probe current and 0.31eV, with >300pA of probe current. Maintaining high probe currents with high energy resolution allows for detailed analysis of Energy Loss Near Edge Structure (ELNES) analysis without the need for a monochromator on core loss edges. The spatial resolution, as demonstrated in the HAADF image of DyScO3, remains unaffected (in this case <63pA) which means that STEM EELS experiments with simultaneously high spatial resolution, energy resolution and signal to noise ratio can now be performed.
        The lifetime of the tip is unaffected by the extraction voltage chosen to perform the experiment.
        ultra-hight brightness X-CFEG
        The energy resolution of the ultra-high brightness X-CFEG can be adjusted using the extraction voltage. In the case above it was varied between 0.39eV (with <500pA of probe current) and 0.31eV (with >300pA of probe current). The spatial resolution, as demonstrated in the HAADF image of DyScO3, remains unaffected (in this case <63pA). Sample courtesy Professor L.F. Kourkoutis, Cornell University
        The combination of enhanced mechanical stability, the latest 5th-order probe aberration correction and the high-resolution (S-TWIN) wide-gap pole piece results in an instrument with the highest commercially-available STEM resolution specifications.
        The Spectra 300 (S)TEM, with X-FEG/Mono or X-FEG/UltiMono, offers STEM resolution specifications of 50 pm at 300 kV, 96 pm at 60 kV and 125 pm at 30 kV with 30 pA of probe current or 100 pA with the X-CFEG.
        High-angle annular dark-field (HAADF) images of GaN[212].
        Ultra-high resolution HAADF STEM imaging on a Spectra 300 (S)TEM. GaN[212] imaged at 300 kV with 40.5 pm Ga-Ga dumbells resolved as well as 39 pm resolution in the FFT. The image was collected on a wide gap S-TWIN pole piece with 30 pA of probe current.
        For a full list of specifications, please refer to the  Spectra 300 (S)TEM datasheet.
        STEM imaging on the Spectra (S)TEM has been reimagined with the Panther STEM detection system, which includes a new data acquisition architecture and two new, solid state, eight-segment ring and disk STEM detectors (16 segments in total). The new detector geometry offers access to advanced STEM imaging capability combined with the sensitivity to measure single electrons.
        Schematic representation of STEM detectors.
        The 16 segmented ring and disk detectors of the Panther STEM detection system allow for a range of STEM signals without the need for multiple detectors.
        The entire signal chain is optimized and tuned to provide unprecedented signal-to-noise imaging capability with extremely low doses to facilitate imaging of beam sensitive materials. Additionally, the completely redeveloped data acquisition infrastructure can combine different individual detector segments, with the future possibility of combining detector segments in arbitrary ways, generating new STEM imaging methods and revealing information that is not present in conventional STEM techniques. The architecture is also scalable and provides an interface to synchronize multiple STEM and spectroscopic signals.
        High-angle annular dark-field (HAADF) images of SrTiO₃.
        Comparison SrTiO₃ [001] HAADF images taken with the Panther STEM detection system with 3 pA, 1.3 pA and <1 pA of probe current. Even with probe currents <1 pA, the signal-to-noise ratio in the image allows automation routines like OptiSTEM+ to correct 1st and 2nd order aberrations in the probe forming optics, delivering sharp images.
        Scanning transmission electron microscopy image of a metal organic framework.
        Extreme low-dose imaging of the metal organic framework (MOF) UiO 66 on the Spectra 300 (S)TEM. A probe current of <0.5 pA was used in combination with iDPC and the Panther STEM detection system to image atomic level details in this highly dose-sensitive material with a spatial resolution of 1.4 Å. The image is a single shot with a frame time of 23.5 seconds (Specimen courtesy of Professor Y. Han, King Abdullah University of Science and Technology).
        The Spectra 300 (S)TEM can be configured with an electron microscope pixel array detector (EMPAD) or a Thermo Scientific Ceta Camera with speed enhancement to collect 4D STEM data sets.
        The EMPAD is capable of 30-300 kV and provides a high dynamic range (1:1,000,000 e- between pixels), high signal-to-noise ratio (1/140 e-), and high speed (1100 frames per second) on a 128 x 128 pixel array, which makes it the optimal detector for 4D STEM applications. (E.g. Applications where the details of the central and diffracted beams need to be analyzed simultaneously, as in the following ptychography image.)
        More details can be found in the  EMPAD datasheet.
        Electron microscope pixel array detector (EMPAD) image of MoS₂.
        The EMPAD detector can be used for a wide variety of applications. On the left, it is used to extend spatial resolution (0.39 Å) beyond the aperture limited resolution at low accelerating voltages (80 kV) in a bi-layer of the 2D material MoS₂ ( Jiang, Y. et al. Nature 559, 343–349, 2018). On the right, it is used to independently image dark field reflections, revealing the complex microstructure of the precipitates in a superalloy (Sample courtesy Professor G. Burke, University of Manchester).
        The Ceta Camera with speed enhancement offers an alternative for 4D STEM applications where a greater number of pixels is required and when EDS analysis needs to be combined with each point in the STEM scan. This solution provides higher resolution diffraction patterns (up to 512 x 512 pixel resolution), suited for applications such as strain measurement.
        From high-throughput, high signal-to-noise ratio elemental mapping in EDS and EELS to oxidation state and surface phonon probing with ultra-high-resolution EELS, the Spectra 300 (S)TEM offers the spectroscopic flexibility to accommodate the widest range of analytical requirements.
        The Spectra 300 (S)TEM can be configured with your choice of three different sources with varying energy resolution (X-FEG Mono, X-FEG UltiMono, and X-CFEG), two different EDS detector geometries (Super-X and Dual-X), and a range of Gatan Continuum spectrometers and energy filters, providing you with the freedom to configure the systems to suit your research needs.
        The Thermo Scientific EDS detector portfolio provides a choice of detector geometries to suit your experimental requirements and optimize EDS results. Both configurations have a symmetric design, producing quantifiable data.  Note that holder shadowing as a function of tilt is compensated in both detector configurations via built-in Thermo Scientific Velox Software functionality.
        The Spectra 300 (S)TEM can be configured with either Super-X (for spectrum cleanliness and quantification) or Dual-X (for the largest solid angle and high-throughput STEM EDS mapping).
        Video Player is loading.
        Current Time 0:00
        Duration 0:12
        Loaded: 99.73%
        Stream Type LIVE
        Remaining Time 0:12
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Video Player is loading.
          Current Time 0:00
          Duration 0:10
          Loaded: 100.00%
          Stream Type LIVE
          Remaining Time 0:10
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            The Super-X detector system provides a highly collimated solid angle of 0.7 Sr and a Fiori number greater than 4000. Super-X is designed for STEM EDS experiments, where spectral cleanliness and quantification are critical.
            The Dual-X detector system provides a solid angle of 1.76 Sr and a Fiori number greater than 2000. Dual-X is designed for high-throughput STEM EDS experiments, such as EDS tomography, or when signal yield is low and fast mapping is critical.
            A DyScO3 perovskite system is examined with the Dual-X detectors below. The ultra-high brightness (>>1.0 x 108 A/m2/Sr/V*) of the X-CFEG and the resolving power of the S-CORR probe corrector are used to deliver a probe to the specimen with 150 pA of current and size <80 pm. With these high brightness probe conditions, EDS mapping can be done rapidly with high sampling and a high signal to noise ratio, resulting in, for the first time, sub-Angstrom spatial information in a single elemental, raw, and unfiltered EDS map. A fast Fourier transform of the Sc map shows up to 90 pm resolution. Additionally, the built-in EDS quantification engine in the Velox Software makes STEM EDS on Spectra 300 (S)TEM fast, easy and quantifiable.
            DyScO₃ image obtained using the Dual-X detectors on a scanning transmission electron microscope.
            DyScO₃ specimen investigated with the powerful combination of ultra-high brightness X-CFEG, S-CORR and the large solid angle (1.76 Sr) of the Dual-X detectors, resulting in high signal-to-noise ratio, atomic resolution (up to 90 pm), unfiltered EDS maps (Sample courtesy Professor L.F. Kourkoutis, Cornell University).
            When the Spectra 300 (S)TEM is equipped with an X-FEG Mono it is optimized for high-throughput EELS elemental mapping and for probing the fine structure of core-loss edges, extracting sensitive chemical information. The energy resolution of an X-FEG Mono can be tuned between <0.2 eV and 1 eV.
            Plasmon position along a gold nanowire imaged on a scanning transmission electron microscope.
            Video Player is loading.
            Current Time 0:00
            Duration 0:30
            Loaded: 33.17%
            Stream Type LIVE
            Remaining Time 0:30
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Localized positions of plasmon excitations along a gold nanowire as a function of their excitation energy (between 0.18 – 1.2 eV), investigated with an electron probe of <0.2 eV energy resolution.
              A Spectra 300 configured with an X-FEG UltiMono provides the highest possible energy resolution. This configuration is capable of tuning the energy resolution between <0.025 eV and 1 eV.
              Electron probe image of a magnesium oxide crystal obtained on a scanning transmission electron microscope.
              Electron probe image of a magnesium oxide crystal obtained on a scanning transmission electron microscope.
              Map of two surface phonons split in energy by 7 meV on a MgO crystal, observed via an electron probe with energy resolution <0.025 eV. (Specimen and analysis courtesy of Isobel Bicket and Prof. Gianluigi Botton, The Canadian Centre for Electron Microscopy, McMaster University.)
              The Spectra 300 (S)TEM accepts a wide range of holders for in situ experiments with its all-in-one S-TWIN wide-gap pole piece. The Thermo Scientific NanoEx holder family can be seamlessly integrated with the microscope, enabling MEMS device-based heating for atomic imaging at elevated temperatures. Below, gold nanoparticles are heated to 700 degrees Celsius and the resulting motion is captured simultaneously with full frame 4k by 4k pixel resolution at a rate greater than 30 frames per second on a Thermo Scientific Ceta Camera with speed enhancement. The result is high spatial and temporal resolution of highly dynamic molecular behavior.
              Video Player is loading.
              Current Time 0:00
              Duration 2:02
              Loaded: 8.18%
              Stream Type LIVE
              Remaining Time 2:02
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • default, selected
                Video Player is loading.
                Current Time 0:00
                Duration 0:59
                Loaded: 16.80%
                Stream Type LIVE
                Remaining Time 0:59
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • default, selected
                  On the left is a high frame rate movie of gold nano-islands at high temperature, collected on a Ceta Camera with speed enhancement. On the right, the 4k x 4k sensor allows digital zoom while maintaining high resolution in the field of interest.
                  Specifications
                  Style Sheet for Products Table Specifications
                  Style Sheet for Komodo Tabs
                  Image corrector
                  • Energy spread: 0.2–0.3 eV
                  • Information limit: 60 pm
                  • STEM resolution: 136 pm
                  Probe corrector
                  • Energy spread: 0.2–0.3 eV
                  • Information limit: 100 pm
                  • STEM resolution: 50 pm (125 pm @ 30 kV)
                  Uncorrected
                  • Energy spread: 0.2–0.3 eV
                  • Information limit: 100 pm
                  • STEM resolution: 136 pm
                  X-FEG/monochromator double corrected (probe+image corrector
                  • Energy spread: 0.2–0.3 eV
                  • Information limit: 60 pm
                  • STEM resolution: 50 pm (125 pm @ 30 kV)
                  X-CFEG double-corrected (probe+image correction) 
                  • Energy spread: 0.4 eV
                  • Information limit: 70 pm
                  • STEM resolution: 50 pm (136 pm @ 30 kV)
                  Source
                  • X-FEG Mono: High-brightness Schottky field emitter gun and monochromator with a tunable energy resolution range between 1 eV and <0.2 eV
                  • X-FEG UltiMono: High-brightness Schottky field emitter gun with ultra-stable monochromator and accelerating voltage with a tunable energy resolution range between 1eV and <0.03 eV
                  • X-CFEG: Ultra-high brightness with an intrinsic energy resolution of <0.4 eV
                  • Flexible high-tension range from 30 – 300 kV
                  Partnering in sustainability — ACT label
                  Understand your lab's environmental impact with the Spectra 300 TEM's ACT label. Created by My Green Lab, the ACT label sets the standard for determining the environmental impact of laboratory products and helps labs make informed, sustainable purchasing decisions.
                  Learn more about the ACT label
                  Resources
                  Video Player is loading.
                  Current Time 0:00
                  Duration 2:12
                  Loaded: 7.40%
                  Stream Type LIVE
                  Remaining Time 2:12
                  1x
                    • Chapters
                    • descriptions off, selected
                    • en (Main), selected
                    Incredible catalysis
                    Video Player is loading.
                    Current Time 0:00
                    Duration 2:28
                    Loaded: 6.61%
                    Stream Type LIVE
                    Remaining Time 2:28
                    1x
                      • Chapters
                      • descriptions off, selected
                      • en (Main), selected
                       More answers with ETEM
                      Video Player is loading.
                      Current Time 0:00
                      Duration 0:58
                      Loaded: 17.23%
                      Stream Type LIVE
                      Remaining Time 0:58
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • default, selected
                        Spectra 300 (S)TEM: A redesigned base and enclosure deliver the highest imaging quality though passive and (optional) active vibration isolation.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 2:18
                        Loaded: 7.20%
                        Stream Type LIVE
                        Remaining Time 2:18
                        1x
                          • Chapters
                          • descriptions off, selected
                          • captions off, selected
                          • default, selected
                          OptiMono+ exciting an X-FEG/UltiMono from the monochromator off state (with 1 eV energy resolution) to the monochromator fully excited state (<30 meV) at 60 kV.
                          Video Player is loading.
                          Current Time 0:00
                          Duration 0:39
                          Loaded: 25.02%
                          Stream Type LIVE
                          Remaining Time 0:39
                          1x
                            • Chapters
                            • descriptions off, selected
                            • captions off, selected
                            • default, selected
                            Tip flashing on the X-CFEG: 60 pm resolution at 200 kV is maintained before and after tip flashing without adjustment of the optics. The process takes <1 min, is required only once per working day, and has no impact on the lifetime of the tip.
                            Video Player is loading.
                            Current Time 0:00
                            Duration 0:12
                            Loaded: 99.73%
                            Stream Type LIVE
                            Remaining Time 0:12
                            1x
                              • Chapters
                              • descriptions off, selected
                              • captions off, selected
                              • default, selected
                              Dual X detector animation.
                              Video Player is loading.
                              Current Time 0:00
                              Duration 0:10
                              Loaded: 100.00%
                              Stream Type LIVE
                              Remaining Time 0:10
                              1x
                                • Chapters
                                • descriptions off, selected
                                • captions off, selected
                                • default, selected
                                Super X detector animation.
                                Video Player is loading.
                                Current Time 0:00
                                Duration 0:30
                                Loaded: 33.17%
                                Stream Type LIVE
                                Remaining Time 0:30
                                1x
                                  • Chapters
                                  • descriptions off, selected
                                  • captions off, selected
                                  • en (Main), selected
                                  Measurement of an area of gold nanowire shows the localized position of plasmon excitations along nanowires as a function of their excitation energy in the range of 0.18 eV to 1.2 eV.
                                  Video Player is loading.
                                  Current Time 0:00
                                  Duration 2:02
                                  Loaded: 8.18%
                                  Stream Type LIVE
                                  Remaining Time 2:02
                                  1x
                                    • Chapters
                                    • descriptions off, selected
                                    • captions off, selected
                                    • default, selected
                                    High frame rate movie of gold nano-islands at high temperature, collected on a Ceta Camera with speed enhancement.
                                    Video Player is loading.
                                    Current Time 0:00
                                    Duration 0:59
                                    Loaded: 16.80%
                                    Stream Type LIVE
                                    Remaining Time 0:59
                                    1x
                                      • Chapters
                                      • descriptions off, selected
                                      • captions off, selected
                                      • default, selected
                                      The 4k x 4k sensor allows digital zoom while maintaining high resolution in the field of interest.
                                      Electron microscope pixel array detector (EMPAD) images of MoS₂.
                                      The EMPAD detector can be used for a wide variety of applications. On the left, it is used to extend spatial resolution (0.39 Å) beyond the aperture limited resolution at low accelerating voltages (80 kV) in a bi-layer of the 2D material MoS₂ (Jiang, Y. et al. Nature 559, 343–349, 2018). On the right, it is used to independently image dark field reflections, revealing the complex microstructure of the precipitates in a superalloy (Sample courtesy Professor G. Burke, University of Manchester).
                                      Ingredients for the best STEM EELS data acquisition
                                      Register below and watch this on-demand webinar to learn how to best optimize the Thermo Scientific Spectra (S)TEM, with its ultra-high-brightness cold-FEG source (X-CFEG), for EELS data collection, especially in the case of a very large collection angle and/or short camera length, where the optics quality becomes very important.
                                      Register now
                                      Introducing the Spectra 300 (S)TEM
                                      Introducing the Spectra 300 (S)TEM
                                      Thermo Fisher Scientific introduces the Spectra 300 (S)TEM – the highest-resolution, aberration-corrected, scanning transmission electron microscope for all materials science applications.
                                      Register below to watch our recorded webinar and learn more about the Spectra 300 (S)TEM; with its wide-gap pole piece and an accelerating voltage range of 30–300 kV, it serves as the tool of choice for the widest range of materials investigations.
                                      Register now
                                      Video Player is loading.
                                      Current Time 0:00
                                      Duration 2:12
                                      Loaded: 7.40%
                                      Stream Type LIVE
                                      Remaining Time 2:12
                                      1x
                                        • Chapters
                                        • descriptions off, selected
                                        • en (Main), selected
                                        Incredible catalysis
                                        Video Player is loading.
                                        Current Time 0:00
                                        Duration 2:28
                                        Loaded: 6.61%
                                        Stream Type LIVE
                                        Remaining Time 2:28
                                        1x
                                          • Chapters
                                          • descriptions off, selected
                                          • en (Main), selected
                                           More answers with ETEM
                                          Video Player is loading.
                                          Current Time 0:00
                                          Duration 0:58
                                          Loaded: 17.23%
                                          Stream Type LIVE
                                          Remaining Time 0:58
                                          1x
                                            • Chapters
                                            • descriptions off, selected
                                            • captions off, selected
                                            • default, selected
                                            Spectra 300 (S)TEM: A redesigned base and enclosure deliver the highest imaging quality though passive and (optional) active vibration isolation.
                                            Video Player is loading.
                                            Current Time 0:00
                                            Duration 2:18
                                            Loaded: 7.20%
                                            Stream Type LIVE
                                            Remaining Time 2:18
                                            1x
                                              • Chapters
                                              • descriptions off, selected
                                              • captions off, selected
                                              • default, selected
                                              OptiMono+ exciting an X-FEG/UltiMono from the monochromator off state (with 1 eV energy resolution) to the monochromator fully excited state (<30 meV) at 60 kV.
                                              Video Player is loading.
                                              Current Time 0:00
                                              Duration 0:39
                                              Loaded: 25.02%
                                              Stream Type LIVE
                                              Remaining Time 0:39
                                              1x
                                                • Chapters
                                                • descriptions off, selected
                                                • captions off, selected
                                                • default, selected
                                                Tip flashing on the X-CFEG: 60 pm resolution at 200 kV is maintained before and after tip flashing without adjustment of the optics. The process takes <1 min, is required only once per working day, and has no impact on the lifetime of the tip.
                                                Video Player is loading.
                                                Current Time 0:00
                                                Duration 0:12
                                                Loaded: 99.73%
                                                Stream Type LIVE
                                                Remaining Time 0:12
                                                1x
                                                  • Chapters
                                                  • descriptions off, selected
                                                  • captions off, selected
                                                  • default, selected
                                                  Dual X detector animation.
                                                  Video Player is loading.
                                                  Current Time 0:00
                                                  Duration 0:10
                                                  Loaded: 100.00%
                                                  Stream Type LIVE
                                                  Remaining Time 0:10
                                                  1x
                                                    • Chapters
                                                    • descriptions off, selected
                                                    • captions off, selected
                                                    • default, selected
                                                    Super X detector animation.
                                                    Video Player is loading.
                                                    Current Time 0:00
                                                    Duration 0:30
                                                    Loaded: 33.17%
                                                    Stream Type LIVE
                                                    Remaining Time 0:30
                                                    1x
                                                      • Chapters
                                                      • descriptions off, selected
                                                      • captions off, selected
                                                      • en (Main), selected
                                                      Measurement of an area of gold nanowire shows the localized position of plasmon excitations along nanowires as a function of their excitation energy in the range of 0.18 eV to 1.2 eV.
                                                      Video Player is loading.
                                                      Current Time 0:00
                                                      Duration 2:02
                                                      Loaded: 2.45%
                                                      Stream Type LIVE
                                                      Remaining Time 2:02
                                                      1x
                                                        • Chapters
                                                        • descriptions off, selected
                                                        • captions off, selected
                                                        • default, selected
                                                        High frame rate movie of gold nano-islands at high temperature, collected on a Ceta Camera with speed enhancement.
                                                        Video Player is loading.
                                                        Current Time 0:00
                                                        Duration 0:59
                                                        Loaded: 1.62%
                                                        Stream Type LIVE
                                                        Remaining Time 0:59
                                                        1x
                                                          • Chapters
                                                          • descriptions off, selected
                                                          • captions off, selected
                                                          • default, selected
                                                          The 4k x 4k sensor allows digital zoom while maintaining high resolution in the field of interest.
                                                          Electron microscope pixel array detector (EMPAD) images of MoS₂.
                                                          The EMPAD detector can be used for a wide variety of applications. On the left, it is used to extend spatial resolution (0.39 Å) beyond the aperture limited resolution at low accelerating voltages (80 kV) in a bi-layer of the 2D material MoS₂ (Jiang, Y. et al. Nature 559, 343–349, 2018). On the right, it is used to independently image dark field reflections, revealing the complex microstructure of the precipitates in a superalloy (Sample courtesy Professor G. Burke, University of Manchester).
                                                          Ingredients for the best STEM EELS data acquisition
                                                          Register below and watch this on-demand webinar to learn how to best optimize the Thermo Scientific Spectra (S)TEM, with its ultra-high-brightness cold-FEG source (X-CFEG), for EELS data collection, especially in the case of a very large collection angle and/or short camera length, where the optics quality becomes very important.
                                                          Register now
                                                          Introducing the Spectra 300 (S)TEM
                                                          Introducing the Spectra 300 (S)TEM
                                                          Thermo Fisher Scientific introduces the Spectra 300 (S)TEM – the highest-resolution, aberration-corrected, scanning transmission electron microscope for all materials science applications.
                                                          Register below to watch our recorded webinar and learn more about the Spectra 300 (S)TEM; with its wide-gap pole piece and an accelerating voltage range of 30–300 kV, it serves as the tool of choice for the widest range of materials investigations.
                                                          Register now
                                                          Applications
                                                          Process control using electron microscopy
                                                          Process control using electron microscopy
                                                          Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
                                                          Quality control and failure analysis using electron microscopy
                                                          Quality control and failure analysis
                                                          Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
                                                          Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
                                                          Fundamental Materials Research
                                                          Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
                                                          Semiconductor-RD_1920x1080
                                                          Semiconductor research and development
                                                          Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
                                                          Semiconductor Failure Analysis
                                                          Semiconductor Failure Analysis
                                                          Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
                                                          Semi-Materials-Character_1920x1080
                                                          Semiconductor materials characterization
                                                          Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
                                                          Display Module Failure Analysis
                                                          Semiconductor display technology
                                                          Display technologies are evolving to improve display quality and light conversion efficiency. Learn how metrology, failure analysis, and characterization solutions provide insights.
                                                          Techniques
                                                          Energy Dispersive Spectroscopy
                                                          Energy dispersive spectroscopy (EDS) collects detailed elemental information along with electron microscopy images, providing critical compositional context for EM observations. With EDS, chemical composition can be determined from quick, holistic surface scans down to individual atoms.
                                                          Learn more ›
                                                          3D EDS Tomography
                                                          Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
                                                          Learn more ›
                                                          Atomic-Scale Elemental Mapping with EDS
                                                          Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
                                                          Learn more ›
                                                          EDS Elemental Analysis
                                                          Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
                                                          Learn more ›
                                                          Electron Energy Loss Spectroscopy
                                                          Materials science research benefits from high-resolution EELS for a wide range of analytical applications. This includes high-throughput, high signal-to-noise-ratio elemental mapping, as well as probing of oxidation states and surface phonons.
                                                          Learn more ›
                                                          In Situ experimentation
                                                          Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
                                                          Learn more ›
                                                          Particle analysis
                                                          Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                                                          Learn more ›
                                                          Multi-scale analysis
                                                          Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
                                                          Learn more ›
                                                          Semiconductor TEM Imaging and Analysis
                                                          Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
                                                          Learn more ›
                                                          Automated Particle Workflow
                                                          The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
                                                          Learn more ›
                                                          Energy Dispersive Spectroscopy
                                                          Energy dispersive spectroscopy (EDS) collects detailed elemental information along with electron microscopy images, providing critical compositional context for EM observations. With EDS, chemical composition can be determined from quick, holistic surface scans down to individual atoms.
                                                          Learn more ›
                                                          3D EDS Tomography
                                                          Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
                                                          Learn more ›
                                                          Atomic-Scale Elemental Mapping with EDS
                                                          Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
                                                          Learn more ›
                                                          EDS Elemental Analysis
                                                          Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
                                                          Learn more ›
                                                          Electron Energy Loss Spectroscopy
                                                          Materials science research benefits from high-resolution EELS for a wide range of analytical applications. This includes high-throughput, high signal-to-noise-ratio elemental mapping, as well as probing of oxidation states and surface phonons.
                                                          Learn more ›
                                                          In Situ experimentation
                                                          Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
                                                          Learn more ›
                                                          Particle analysis
                                                          Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                                                          Learn more ›
                                                          Multi-scale analysis
                                                          Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
                                                          Learn more ›
                                                          Semiconductor TEM Imaging and Analysis
                                                          Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
                                                          Learn more ›
                                                          Automated Particle Workflow
                                                          The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
                                                          Learn more ›
                                                          Style Sheet to change H2 style to p with em-h2-header class
                                                          Contact us
                                                          Electron microscopy services
                                                          To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
                                                          Learn more ›
                                                          Style Sheet for Support and Service footer
                                                          Style Sheet for Fonts
                                                          Style Sheet for Cards