Belt scale product comparison table
Explore the Ramsey Series Belt Scale System specifications and capabilities to determine which system is best suited to your operation.  For custom configurations, questions or help in selecting a system, contact a solution specialist using the Contact Sales link above.
Compare Ramsey Series belt scale systems
Ramsey IDEA Belt Scale System
Ramsey Series 20 Belt Scale System
Ramsey Series 17 Belt Scale System
Ramsey Series 14 Belt Scale System
Ramsey Series 30 Belt Scale System
Accuracy
+/-1%
+/-0.5%
+/-0.25%
+/-0.125%
+/-1%
Number of idlers
1
1 or 2
2 or 4
3, 4, or 6
1
Number of load cells
1 or 2
1 or 2
2
4
1 or 2
Belt widths
10-101-R1 max 914 mm (36 in)
10-101-R2 max 1524 mm (60 in)
406-1981 mm (16–78 in)
457-2590 mm (18-102 in)
457-2590 mm (18-102 in)
HD version available for larger sizes
Single Load Cell version 381-914 mm (15-36 in)
Dual Load Cell version 635-1524 mm (25-60 in)
Belt speed range: Material must spend at least 0.5 seconds in the weigh area;
varies depending on # of idlers and idler spacing.
Belt loading
Model 10-101-R1: 7.5-114 kg (16.5–250 lb)
Model 10-101-R2: 13.6-340 kg (30–750 lb)
Model 10-20 1:  22.7-816 kg (50–1800 lb)
Model 10-20-CWT: 9-22.7 kg (20–50 lb)
Contact a representative for calculation
11.9-1116 kg/m (8–750 lbs/ft)
Single Load Cell version 15.8-227 kg (35-500 lb)
Dual Load cell version 29.5-907 kg (65-2000 lb)
Idler spacing
R1 - Larger than 533 mm (21.5 in)
R2 - larger than 571 mm (22.5 in)
812 mm (32 in) minimum
Standards 900 mm, 1000 mm and 1200 mm (36 in, 42 in, 48 in)
Standards 900 mm, 1000 mm and 1200 mm (36 in, 42 in, 48 in)
Standards 900 mm, 1000 mm and 1200 mm (36 in, 42 in, 48 in)
Typical applications
Cost-effective weighing for non-critical or lower value materials, modular installation
General in plant or in-process
High-accuracy weighing
High-value materials
High quality and high accuracy requirement
Certifiable or basis-of-payment
Cost-effective weighing for non-critical or general in plant or in-process
1x1 image pixel for data collection