The complex structures of biopharmaceutical therapeutic agents require greater selectivity and sensitivity, which drives the demand for powerful instrumentation and software for their data acquisition and analysis. Previously this required two software packages, one in Development and one in Quality Control (QC). Thermo Scientific Chromeleon 7.3.2 Chromatography Data System (CDS) comfortably operates in both areas and supports Mass Spectrometry (MS) instruments in a compliant enterprise environment, providing a seamless transition from Development to QC within a single software.
What are users saying?
"We use Chromeleon daily in my labs. Chromeleon report templates enable us to keep all the raw data, data manipulation, and lot recording in one file so submitting to our lab notebook software is super easy. All experiment details are in one place."
- Research Lab Manager, Genentech
Support for biopharmaceutical workflows
Support for biopharmaceutical workflows
Chromeleon CDS provides native control of MS and MS/MS instruments (for GC, HPLC, and IC front ends) with workflow support for targeted and non-targeted analysis, including lot release testing of intact and digested proteins, oligonucleotides analysis, and LC-MS/MS-based pharmaceutical bioanalysis.
  • Control for over 35 Thermo Scientific MS instruments including single and triple quadrupole and Orbitrap high resolution accurate mass (HRAM)
  • Workflow support including bioanalysis, multi-attribute method (MAM), intact protein deconvolution, oligonucleotides analysis, dioxin and PCB analysis, and SVOCs
  • Built-in report templates and views specific to the needs of conducting MAM, oligonucleotides analysis and bioanalysis experiments
  • Bi-directional connections for Thermo Scientific SampleManager and Watson LIMS Software for complete end-to-end support
Learn more about Chromeleon CDS features for mass spectrometry
Built for compliance: Leverage compliance tools for your MS
  • Chromeleon CDS is the recommended software to control Thermo Scientific MS instruments for compliant analysis. Compliant tuning for Thermo Scientific ISQ, TSQ and Exploris series MS instruments
  • Achieve compliance with a full suite of features to ensure adherence to regulations, data integrity guidelines, and electronic record requirements
  • Maintain compliance with user controls and comprehensive audit trails to track and version all actions
  • Demonstrate compliance with easy, regular audit trail reviews
    • Audit trail events and queries quickly and clearly present the info needed
    • Versioning shows how changes have affected results
Built for QC: Up to 33% increased efficiency for routine MS analyses
Built for QC: Up to 33% increased efficiency for routine MS analyses
Feel confident with fewer failed runs and out-of-specification results through:
  • Built-in troubleshooting and diagnostics for specific Thermo Scientific HPLC and UHPLC systems to minimize downtime
  • Pre-run sequence ready checks and runtime evaluation of System Suitability Tests (SSTs) with Intelligent Run Control for results-based reactions, deliver more "right-first-time" analyses
  • Enhanced universal eWorkflow procedures streamline and control sequence creation to simplify adherence to SOPs
  • Spreadsheet-based report templates to calculate the results inside the CDS
Built for MAM: Straight through to breakthrough
Built for MAM: Straight through to breakthrough
Chromeleon CDS is an integral part of the Thermo Scientific MAM 2.0 workflow. This powerful, high-resolution MS-based workflow delivers easy, compliance-ready characterization and monitoring of peptides and allows critical quality attributes to be monitored throughout every stage of the drug development process. The workflow leverages comprehensive compliance capabilities from data acquisition, to quality attribute quantitation, new peak detection, and customizable spreadsheet-based reporting.
Built for Bioanalysis: Large molecule bioanalysis and DMPK studies
Understanding your molecules is critical, and ensuring accuracy is paramount when it comes to DMPK studies. Chromeleon CDS provides a single software solution that is scalable,from a local standalone workstation to a multi-site enterprise system, with built-in features to simplify meeting compliance requirements. It provides robust and reliable control of the HPLC and MS with integrated MS/MS functionality for data acquisition and processing. A bi-directional gateway for the Watson LIMS provides complete end-to-end support.
Built for Oligonucleotides: Robust, accurate analysis of your oligonucleotide products
From instrument control and data acquisition to automated data processing to customizable reports to streamlined data integrity reviews, Chromeleon CDS facilitates compliant-ready high resolution LC-MS analysis of modified and unmodified oligonucleotides. High throughput accurate molecular weight confirmation is streamlined with automated intact deconvolution processing. Chromeleon CDS provides enhanced easy electronic data reviews and secure organized data storage with the benefits of a scalable enterprise system for global collaboration.
Compliance-Ready MS for Biopharma
Examine real-use cases of how a networked CDS is the best choice for data acquisition and processing for quantitative LC-MS analyses.
Download eBook ›
End-to-end MAM Solution to move Biopharma forward
Discover the compliance-ready, enterprise MAM workflow that connects development and manufacturing.
Solutions for Biopharma Characterization and Control
Be Confident. As your partner, we provide all the right products and solutions for your characterization and control challenges.
 Download interactive book ›
* Required field
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code