Components of an HPLC system
A fully operational HPLC system combines hardware, software, and consumables, each essential in successful analysis. Here is a breakdown of each component and the function:
HPLC mobile phase
Every HPLC separation begins with the high-pressure flow of the mobile phase at a specified flow rate, which drives the sample from the injector through the column to the detector.
Capillaries and fittings
The capillaries and fittings complete an HPLC system by connecting the individual hardware components so the mobile phase and sample mixture flow continuously at high pressure.
Chromatography data system
A chromatography data system (CDS) is software that controls all the components of an HPLC system, including operation, mobile phase mixing, pump flow, sample injection, and column temperature.
FAQ
An HPLC instrument has four basic parts which include a pump, autosampler, a column compartment and detector.
A functional HPLC or UHPLC system combines hardware, software, solvents, columns and other consumables.
* Required field
*
*
*
*
*
*
*
*
CMD Form Code v3.0
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code