Search Thermo Fisher Scientific
VUV detectors paired with the Thermo Scientific TRACE 1600 Series GC conquer the challenge of complex mixtures, co-eluting compounds, and isomer differentiation for accurate hydrocarbon speciation, impurity detection, and trace-level detection.
Gas Chromatography (GC) is evolving, and VUV (Vacuum Ultraviolet) technology is at the forefront of this transformation. Now available on the TRACE 1600 Series GC, VUV spectroscopy, through VUV Analytics VGA-100 detector and the LUMA detector, provides accuracy in hydrocarbon speciation, impurity detection, and trace-level analysis.
Greater accuracy, automation, and confidence in your GC analysis
Traditional GC detectors struggle with complex mixtures, co-eluting compounds, and isomer differentiation. VUV spectroscopy overcomes these limitations by using absorption-based detection, capturing unique molecular fingerprints for each compound.
VUV detection provides qualitative and quantitative spectral information for most gas phase compounds.
Broad-spectrum absorption: VUV detectors measure how molecules absorb light in the vacuum ultraviolet range (120–240 nm), revealing unique spectral signatures.
Automated compound ID: To reduce errors and improve analysis speed, VUV software automatically deconvolutes overlapping peaks. unlike traditional GC detectors.
Quantitative and qualitative insights: VUV enables compound identification and quantification in a single run by analyzing complete spectral data.
Elevate your gas chromatography detection capabilities with vacuum ultraviolet spectroscopy through dedicated GC-VUV analyzers for fuels. Experience superior excellent sensitivity, selectivity, and ease of use, tailored to meet the needs of fuels characterization. To analyze finished fuels and feedstocks using traditional methods requires multiple analytical techniques, complicated hardware set up, extensive analysis time, and prior knowledge of the samples being analyzed, leading to wasted time on re-analysis.
With unique selectivity for trace analysis, GC-VUV technology ensures accurate and reliable results for impurities detection at part per billion levels, i.e. nitrosamine, in complex matrices. GC-VUV technology is ideal for residual water analysis, as well as solvent impurities in one single analytical run, saving time and resources while maintaining high sensitivity and selectivity.
Hydrocarbon speciation
Trace impurity detection
Analysis and workload
VGA-100 and LUMA detectors integrate seamlessly with the TRACE 1600 Series GC, offering plug-and-play installation, intuitive software, and superior analytical performance.
|
||
Type | Universal mass-sensitive GC detector | VUV multichannel absorbance GC detector |
Use | Accurately identify and quantify compounds, including isomers, through deconvolution and spectra library matching. | Simplify trace-level detection of impurities in complex matrices for accurate speciation in industrial samples. |
Applications | Hydrocarbon analysis, including:
|
Trace-level impurity detection for:
|
Features |
|
|
GC-VUV for hydrocarbon analysis
See how Vacuum Ultraviolet (VUV) technology can transform GC analysis of complex mixtures and benefit lab operations.
Ready to experience the future of GC?
Upgrade your lab with VUV technology and experience faster and more accurate, automated GC analysis.
For Research Use Only. Not for use in diagnostic procedures.