Dionex Aquion IC System: ion chromatography for water analysis
Perform reliable, basic anion and cation analyses with the Thermo Scientific Dionex Aquion ion chromatography system. This simple, compact ion chromatography (IC) instrument, commonly used for water analysis, is designed for ease of use and straightforward operation for budget-limited labs. It supports electrolytic suppression and includes a vacuum degasser for inline degassing of eluents. It also offers fast start-up times and reliable, stable performance for demanding applications in environmental, food safety, and academic laboratories.
Highlights
Conductivity detection
The inert, nonmetallic PEEK flow path of the Dionex Aquion IC system is designed and optimized for applications using conductivity detection. It ensures compatibility with corrosive eluents and IC analyses free from metal contamination. The high-performance conductivity cell is temperature controlled to protect against ambient temperature variations. Digital control of the cell offers a wide, dynamic range for detection.
Suppressor compatibility
The Dionex Aquion IC system supports both chemical and electrolytic suppression technologies. Suppressors remove conductive ions from the eluent to increase analyte signal while decreasing background noise. This enables the highest possible sensitivity and lowest detection limits for conductivity detection.
Optional column heater
Get increased precision and run applications at elevated temperatures with an optional column heater. The column heater preheats the eluent prior to entering the column and maintains the column temperature set by the user. Heater temperature can be set between 30°C and 60°C.
Eluent degasser
Eluent degassing reduces baseline noise, ensuring reproducibility and protection of eluents from contamination and decomposition. An optional built-in vacuum degas provides in-line degassing of eluents, and operation can be automated to sense when degassing is required.
CDS Software
The Dionex Aquion IC system comes with the easy-to-use Thermo Scientific Chromeleon Chromatography Data System (CDS). This software platform provides powerful control and data processing for IC and offers eWorkflows that make all our essential applications easily accessible.
Convenient bundles
Dionex Aquion IC bundles for anion or cation analysis include all hardware, software, and consumables to get started, with optional extended warranties available for peace of mind.
Videos
Video Player is loading.
Current Time 0:00
Duration 3:35
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:35
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    System specifications
    The table below provides an overview of the Dionex Aquion IC system specifications. Not all specifications are listed. For a full list of the specifications, download the Dionex Aquion Ion Chromatography system specifications sheet.
    Specification
    Dionex Aquion IC System
    Analytical pump and fluidics
    Type
    Serial dual-reciprocating pistons, microprocessor-controlled constant stroke, variable speed
    Construction
    Chemically inert, metal-free PEEK pump heads and flow paths compatible with aqueous eluents of pH 0–14 and reversed-phase solvents
    Pump operating pressure
    0–35 MPa (0–5000 psi)
    Flow rate range
    0.00–5.00 mL/min without changing pump heads
    Flow precision
    <0.1%, typically
    Flow accuracy
    <0.1%, typically
    Pressure ripple
    <1% at 13.8 MPa (2000 psi) and 1.0 mL/min
    Eluent on-off valve
    Standard
    Piston seal wash
    Dual-pump head, wash can be continuous when connected to rinse solution supply
    Pressure alarm limits
    Upper limit 0–35 MPa or 0–5000 psi in one unit (MPa or psi) increments; lower limit can be set up to one unit lower than upper limit
    Vacuum degas
    Yes, optional, automatic control
    Eluent bottles
    Polypropylene, up to 4L volume
    Eluent bottle pressure
    Not required
    Columns supported
    2, 3, 4, and 5 mm i.d.; maximum length 250 mm analytical column with 50 mm guard column
    Injection valve
    Type
    6-port, 2 position Rheodyne valve, electrically actuated
    Column heater (optional)
    Operating temperature range
    30 to 60 °C (86 to 140 °F); minimum 5 °C above ambient; settable range is equal to working range
    Temperature accuracy
    ±0.5 °C at sensor, at 40 °C
    Suppressors and control
    Chemical suppression
    2 mm and 4 mm anion and cation, membrane suppression bed types
    Displacement chemical suppression
    2 mm and 4 mm anion and cation, membrane suppression bed types
    Electrolytic suppression
    2 mm and 4 mm anion and cation, membrane suppression bed types available
    Electrolytic suppression with external water mode
    2 mm and 4 mm anion and cation, membrane suppression bed types available
    Current control range
    Thermo Scientific Dionex ERS 500 Electrolytically Regenerated Suppressor: 4 mm, 0–500 mA in 1 mA increments 2 mm, 0–150 mA in 1 mA increments
    Thermo Scientific Dionex ERD 500 Electrolytically Regenerated Desalter: 0–500 mA (4 mm) and 0–150 mA (2 mm) in 1 mA increments
    Thermo Scientific Dionex SRN Self-Regenerating Neutralizer: 0–500 mA in 1 mA increment
    Salt converter
    Available in 2 and 4 mm versions
    Carbonic acid removal for anions
    Dionex AERS 500 suppressor with Thermo Scientific Dionex CRD 200 Carbonate Removal Device for hydroxide eluents Dionex AERS 500 suppressor with Dionex CRD 300 device for carbonate eluents
    Non-suppressed chromatography
    Yes, supported
    Suppressor wear parts
    None; peristaltic pump and inline filters not required
    Conductivity detector electronics and flow cell
    Type
    Microprocessor-controlled digital signal processor
    Cell drive
    8 kHz square wave
    Detector linearity
    1% up to 1 mS
    Maximum cell pressure
    10 MPa (1500 psi)
    Resolution
    0.00238 nS/cm
    Full-scale output ranges
    Digital signal range 0–15000 μS/cm
    Analog signal range 0–15000 μS/cm
    Electronic noise
    ±0.1 nS when background conductivity is 0–150 µS/cm
    ±2 nS when background conductivity is 151–3200 μS/cm
    Filter
    Rise times from 0 to 10 s, user selectable
    Temperature compensation
    Fixed at 1.7% per 1 °C at cell temperature
    Temperature range
    Ambient +7 °C, 30 to 55 °C
    Cell electrodes
    Passivated 316 stainless steel; compatible with MSA
    Cell body
    Chemically inert polymeric material
    Cell volume
    <1 µL
    Heat exchanger
    Inert, tortuous path for low axial dispersion
    Autosampler
    Automation using autosampler
    For more information about available autosamplers, visit the IC Autosamplers page
    System software
    Chromeleon CDS, requires Microsoft® Windows® 10
    Physical specifications
    Dimensions (h × w × d)
    56.1 cm × 22.4 cm × 53.3 cm (22.1 in × 8.8 in × 21 in)
    Weight
    24.5 kg (54 lb)
    Power requirements
    100 to 240 V AC, 50 to 60 Hz, autoranging
    Operating temperature
    4–40 °C (40–104 °F); cold-room-compatible (4 °C) as long as system power remains on
    Operating humidity range
    5–95% relative, noncondensing
    Control modes
    Full control through Chromeleon CDS software; alternative control through TTL or relay closures; two relay outputs, two TTL outputs, four programmable inputs
    USB communication protocol
    One USB input; one built-in two-output USB hub
    Leak detection
    Built-in, optical sensor
    For Research Use Only. Not for use in diagnostic procedures.