Overview
Size exclusion chromatography (SEC) columns are the ideal choice for monitoring aggregates.
Protein aggregation is a critical quality attribute (CQA) for monoclonal antibodies (mAbs) as well as adeno-associated viruses (AAVs). It plays a significant role in determining the overall quality of these biologics. To detect and quantify aggregates, size-exclusion chromatography (SEC) is the preferred method.
SEC operates on the steric-exclusion principle, where larger species are effectively excluded from the pores of the packing material and elute faster than smaller species. This principle allows for accurate determination of aggregate levels in a sample.
SEC HPLC Columns for Aggregate Analysis
SurePac Bio 550 SEC MDi columns
MAbPac SEC-1 HPLC columns
✓ Our most recent addition for fast, high-resolution separation of AAV monomers and aggregates
✓ For high-resolution separation of mAbs from monomers, aggregates, and Fab & Fc fragments resulting from proteolysis
Order now
Order now
3 ways to search
  1. Find an equivalent column to the one you’re using now
  1. Find the best column for your USP method
  1. Find the right column for your new method
Find your HPLC column with ease ›
Related resources
* Required field
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code