hdi-athlete-bio-passport-testing-230x195
Monitor select doping biomarkers
While the approach of screening for prohibited compounds or their metabolites in an athlete’s blood or urine sample is an effective approach to monitor athletes, it has its limitations. With new, modified or designer drug substances reportedly misused by athletes, anti-doping agencies and laboratories seek new detection strategies to combat emerging threats.
Thermo Fisher Scientific has delivered leading technology for steroid analysis for more than a quarter century.
The WADA guidelines introduced a series of blood- or urine-based test modules starting in 2009, which include the Athlete Hematological Passport (blood doping) and the Athlete Steroidological Passport, a longitudinal steroid profile to detect the abuse of testosterone; i.e.endogenous steroids administered exogenously. The principle of the Athlete Biological Passport (ABP) is to monitor select biological parameters (not drugs compounds) over time, allowing agencies to generate profiles for an athlete.
Contact us  Read the blog
Featured categories
Delivering a new standard in routine GC analysis that increases productivity and reduces cost of ownership. Choose from advanced sampling handling solutions, modern GC systems, quality columns and state-of-the-art reporting, reagents and accessories.
To ensure athletes play true, our market leading solutions help you profile athletes and confirm steroid use with precision and sensitivity expected.
Redefining HPLC and UHPLC to give you more. Streamline your applications with our highly integrated solutions including systems, standard and bio-LC columns, autosamplers, detectors, fittings, software and more.
Offering solid phase extraction (SPE), solid supported liquid extraction (SLE), micro extraction by packed sorbent (MEPS), and manifolds and dispensers you can use in the process. Don’t overlook the syringe filters, protein precipitation and protein digest products.
Confirm the presence of synthetic forms of endogenous anabolic androgenic steroids (EAAS) in urine samples.
Support in and out of competition testing with solutions to screen large sets of compounds in a single methods while staying ahead of future banned substances.
Typical passport of suspicious steroid profile confirmation workflow
Athlete biological passport sample preparation
For athlete samples testing positive for prohibited compounds or noted as suspicious as a result of steroid quantitation and confirmation testing, it is important to ensure samples are prepared consistently for isotopic characterization. Following hydrolysis, most labs perform additional sample preparation. Thermo Fisher Scientific offers the solutions to help maximize sample clean up and improve your analytical sensitivity.
We offer a variety of solutions to fit your laboratory workflow needs, including  the next generation SOLA solid phase extraction fritless  SPE , which provides higher levels of reproducibility and robustness compared to conventional options.  Learn more about the variety of options available, view our application handbook or try our selector guide.
Isotopic confirmation
The misuse of endogenous anabolic steroids can be detected by measuring the ∂13C values of the steroid itself or its metabolites which show depleted isotopic composition while the endogenous reference compounds are not affected. Samples showing suspicious steroid profile parameters are isotopically characterized in order to prove the origin of steroids.
Confirm your results with a single analysis
Confirm your results with a single analysis
general-1-spot-40x40
general-2-spot-40x40
general-3-spot-40x40
general-4-spot-40x40
general-5-spot-40x40
Metabolite identification
Comprehensive studies on the metabolism of therapeutic agents and drugs with misuse potential are required to identify metabolites. Direct couple your IRMS to a GC-MS system to identify unknown metabolites and provide structural elucidation and confirmation.
Choose Thermo Scientific™ ISQ™ series quadrupole GC-MS Systems for time-proven performance and productivity. Ideal for high-throughput laboratories that want to achieve uninterrupted productivity
Athlete biological passport sample preparation
For athlete samples testing positive for prohibited compounds or noted as suspicious as a result of steroid quantitation and confirmation testing, it is important to ensure samples are prepared consistently for isotopic characterization. Following hydrolysis, most labs perform additional sample preparation. Thermo Fisher Scientific offers the solutions to help maximize sample clean up and improve your analytical sensitivity.
We offer a variety of solutions to fit your laboratory workflow needs, including  the next generation SOLA solid phase extraction fritless  SPE , which provides higher levels of reproducibility and robustness compared to conventional options.  Learn more about the variety of options available, view our application handbook or try our selector guide.
Isotopic confirmation
The misuse of endogenous anabolic steroids can be detected by measuring the ∂13C values of the steroid itself or its metabolites which show depleted isotopic composition while the endogenous reference compounds are not affected. Samples showing suspicious steroid profile parameters are isotopically characterized in order to prove the origin of steroids.
Confirm your results with a single analysis
Confirm your results with a single analysis
general-1-spot-40x40
general-2-spot-40x40
general-3-spot-40x40
general-4-spot-40x40
general-5-spot-40x40
Metabolite identification
Comprehensive studies on the metabolism of therapeutic agents and drugs with misuse potential are required to identify metabolites. Direct couple your IRMS to a GC-MS system to identify unknown metabolites and provide structural elucidation and confirmation.
Choose Thermo Scientific™ ISQ™ series quadrupole GC-MS Systems for time-proven performance and productivity. Ideal for high-throughput laboratories that want to achieve uninterrupted productivity
Removing Doubt in Forensic Science
In this eBook overview of the current state of forensic chemistry and the latest mass spectrometry technology for staying ahead of your casework, we discuss:
  • Changes in regulations
  • Drugged driving cases
  • Polydrug use - blood alcohol, marijuana, cocaine,
  • Novel psychoactive substances - cathinones, cannabinoids, fentanyl, designer drugs
Register to download
Featured video
Exclusive Interview with World Cup Anti-doping Director Dr. Martial Saugy, Director, LAD Take a tour of the WADA-accredited Laboratorie Suisse d’Analyse du Dopage (LAD) and learn how the team handled and tested anti-doping specimens from the 2014 FIFA World Cup in Brazil.
Featured webinar
In this webinar Dr. Alessandro Casilli, Brazilian Laboratory for Doping Control, recent advances in high resolution accurate mass (HRAM-MS) Orbitrap technology for measuring different ionization modes during the same run, and improving specificity and detection limits in sports doping control analyses.
Register to view webinar