DataTaker DT90 Series Data Loggers
Meet the compact and low power dataTaker® DT90 Series of data loggers.
Designed for companies in need of universal and low power data logging solutions, this compact data logger is cost-effective, yet packs all the necessary features for logging your data.
The DT90 Series has been designed with an emphasis on low power, small size, and ease of deployment. This data logger is ideal for your remote monitoring environmental and industrial applications. There are two models available–the DT90L and DT90N. The DT90N is perfect for setting up small stations that require multiparameter sensors that only have a digital output. The DT90L is designed for multiparameter applications and can interface with both analog and digital sensors. Both options in this series share the same kernel board and core functionality.
Request quote    Download brochure
Compact, all-in-one monitoring
The DT90 series of data loggers are compact, all-in-one monitoring modules with an integrated modem, specifically designed for remote applications. The rugged design and wide operating temperature range provides reliable operation in virtually any environment. These are low-power and cost-effective data loggers with a dedicated mounting handle for a simple and quick installation process.
Data retrieval and device configuration are available via USB interface for on-site communication. You can also use the automatic data delivery feature to send logged data to an FTP server at regular intervals. Alarm configurations can trigger SMS messages to multiple mobile phones.
DT90 programming software can easily take you through the setup of your logger and how you want the system to work, customized for you.
Main features and benefits:
  • Ease of deployment—Simple connectivity via USB on-site makes it easy to configure and test the deployed monitoring module.
  • Universal—This universal logger can connect to any type of sensor. Analogue*: voltage, current, RTD; Digital: SDI-12, MODBUS, pulses
  • Rechargeable lithium ion battery—With up to 300 days of runtime on one charge and the ability to connect directly to solar power, this logger is the best solution for your remote monitoring applications.
  • Robust—With its solid aluminum case and fully waterproof probe (IP68 rated), this logger is suitable for harsh environments.
  • Cost-effective—This is a low maintenance product. Minimal attention is needed after setup.
*For DT90L version
Compare DT90 models
DT90L (Lite)
DT90N (Nano)
DT90L (Lite) data logger
DT90N (Nano) data logger
This version uses a large 48 Wh integrated rechargeable lithium battery pack and features both analog and digital inputs, with the whole design being fitted in a miniature 160 x 100 x 80 mm black powder-coated aluminum cast enclosure.
The dataTaker nano option uses a smaller 16 Wh integrated rechargeable lithium battery pack when compared to the DT90L. It is all digital (no analog inputs), with the whole design being fitted in a miniature 100 x 70 x 38 mm black powder-coated aluminum cast enclosure.
Work time example
A DT90 logger powered from a 2W solar panel and sampling a rain gauge every minute, with hourly data transmission, has the ability to work all-year round
Analog channels
Up to 8 inputs with 16-bit resolution
-
Digital channels
Dedicated counter inputs, 3 pulse counters inputs
SDI12 input
One input for up to 2 sensors, 32 parameters per sensor
MODBUS Client
Read data from MODBUS sensors/devices on RS485 port
Internal battery
Lithium 48 Wh
Lithium 16 Wh
Dimensions
160 x 100 x 80 mm, 1 Kg
100 x 70 x 38 mm, 700 g
Voltage output
Switchable range: 0–50 mV, 0–2.5 V, 4–20 mA current loop
-
RTD–material supported
PT100
-
Sensor excitation outputs
5 V or 12 V
12 V
Scheduling of data acquisition
  • Number of channels: 26 (each has independent schedule setting) 
  • Schedule Rates: 1 second to 24 hours
Alarms
  • Conditions: high or low threshold
  • Number of conditions: 3
  • Action: send SMS, increase/decrease sampling rate or transmission rate
Data storage
  • Internal non-volatile capacity: 16 MB (min 500,000 data points)
Integrated cellular modem
  • Network selection 3G or 4G
Power supply
  • External power: 5-25 Vdc
  • Solar panel: 9 V or 12 V
Physical and environment
  • Construction: Black anodized cast aluminum
  • Working temperature: -40 °C to +70 °C
  • Humidity: 0–100% RH (when the lid is closed)
  • Ingress of Protection: IP67
Included accessories
  • Resource USB (software + user manual)
  • USB communication cable
DT90 configuration software
The DT90 configuration software is an intuitive graphical interface that allows you to configure your data logger, view real-time data in charts, and retrieve your historical data for analysis. This application runs directly from your PC and can be accessed via USB.
Features include:
  • Simple Interface for programming
  • Easy alarm setup
  • USB connection
  • Ability to download historical data
  • Display data in chart or table
  • Export data into a CSV file·
  • Firmware upgrade interface
dataTaker feature highlights
dataTaker live cloud service
Collect, Share, Process.
This powerful and versatile cloud service makes collecting, monitoring, analysing, sharing, and storing information from your dataTaker easier.
  • Monitor—Have real-time remote access to measurements from anywhere at any time, allowing configuration changes for dataTaker loggers from your PC, smartphone, or tablet.
  • Store and share—Synchronise and save data to your personal cloud account. The dataTaker Live dashboard lets you collaborate and share data.
  • Collect—Automatically upload data from sensors, loggers, or industrial gateways to your personal cloud service account.
  • Analyse—Perform data analysis with standard and custom scripts.
Learn more
Optional accessories for both models
Solar panel with brackets and 1 m cable
  • Rating: 2 W, 5 W, or 10 W
Power adaptor 230 V
  • Cable length: 1 meter or 5 meter
Mounting bracket
  • Type: pole mounting or DIN rail mounting
Antenna
  • Straight antenna with screw
  • Straight antenna with magnetic base
  • Dome antenna
Environmental (Hydrology/Meteorology)
Industrial
Agriculture applications for environmental logger
  • Surface water monitoring
  • Weather and climate monitoring
  • Air quality and pollution
  • Monitoring
  • Environmental research
  • Flood warning
  • Agriculture and soils research and monitoring
  • Water quality monitoring
  • Aquaculture
  • Energy monitoring
  • Temperature profiling
  • Material testing
  • Level/flow/monitoring
  • Design engineering
Scheduling of data acquisition
  • Number of channels: 26 (each has independent schedule setting) 
  • Schedule Rates: 1 second to 24 hours
Alarms
  • Conditions: high or low threshold
  • Number of conditions: 3
  • Action: send SMS, increase/decrease sampling rate or transmission rate
Data storage
  • Internal non-volatile capacity: 16 MB (min 500,000 data points)
Integrated cellular modem
  • Network selection 3G or 4G
Power supply
  • External power: 5-25 Vdc
  • Solar panel: 9 V or 12 V
Physical and environment
  • Construction: Black anodized cast aluminum
  • Working temperature: -40 °C to +70 °C
  • Humidity: 0–100% RH (when the lid is closed)
  • Ingress of Protection: IP67
Included accessories
  • Resource USB (software + user manual)
  • USB communication cable
DT90 configuration software
The DT90 configuration software is an intuitive graphical interface that allows you to configure your data logger, view real-time data in charts, and retrieve your historical data for analysis. This application runs directly from your PC and can be accessed via USB.
Features include:
  • Simple Interface for programming
  • Easy alarm setup
  • USB connection
  • Ability to download historical data
  • Display data in chart or table
  • Export data into a CSV file·
  • Firmware upgrade interface
dataTaker feature highlights
dataTaker live cloud service
Collect, Share, Process.
This powerful and versatile cloud service makes collecting, monitoring, analysing, sharing, and storing information from your dataTaker easier.
  • Monitor—Have real-time remote access to measurements from anywhere at any time, allowing configuration changes for dataTaker loggers from your PC, smartphone, or tablet.
  • Store and share—Synchronise and save data to your personal cloud account. The dataTaker Live dashboard lets you collaborate and share data.
  • Collect—Automatically upload data from sensors, loggers, or industrial gateways to your personal cloud service account.
  • Analyse—Perform data analysis with standard and custom scripts.
Learn more
Optional accessories for both models
Solar panel with brackets and 1 m cable
  • Rating: 2 W, 5 W, or 10 W
Power adaptor 230 V
  • Cable length: 1 meter or 5 meter
Mounting bracket
  • Type: pole mounting or DIN rail mounting
Antenna
  • Straight antenna with screw
  • Straight antenna with magnetic base
  • Dome antenna
Environmental (Hydrology/Meteorology)
Industrial
Agriculture applications for environmental logger
  • Surface water monitoring
  • Weather and climate monitoring
  • Air quality and pollution
  • Monitoring
  • Environmental research
  • Flood warning
  • Agriculture and soils research and monitoring
  • Water quality monitoring
  • Aquaculture
  • Energy monitoring
  • Temperature profiling
  • Material testing
  • Level/flow/monitoring
  • Design engineering
CMD SchemaApp code