Orbitrap Exploris MX, Perfect fit
Orbitrap Exploris MX
Fit for purpose mass detection
Accelerate biopharmaceutical late-stage development, commercialization, manufacturing, and quality control (QC) by easily and reliably obtaining compliance-ready, high-quality mass information.
The Thermo Scientific Orbitrap Exploris MX mass detector is the perfect fit for deploying routine mass monitoring.
In addition to high-resolution mass detection provided by Thermo Scientific Orbitrap mass analyzers, this fit for purpose system is simple to operate and compliance ready with Thermo Scientific Chromeleon chromatography data system (CDS) software. Consistent performance between the Thermo Scientific Orbitrap Exploris 240 mass spectrometer and Orbitrap Exploris MX detector ensures seamless method transfer from development to QC.
High-resolution accurate-mass (HRAM) data provide utmost confidence for:
  • Sequence confirmation
  • Peptide monitoring
  • Intact protein analysis (from small to large molecules, reduced mAbs, and intact mAbs under native and denaturing conditions)
  • Glycan profiling
  • Oligonucleotide and oligonucleotide impurity mass confirmation
Speak to a specialist   Take 3D Tour   Request a trade-in
On-demand Webinar: Toward a Next-Generation Multi-Attribute Method (MAM): An Evaluation of the Orbitrap Exploris MX Mass Detector
Presented by Andrew W. Dawdy, Ph.D., Principal Scientist,
BioTherapeutics Pharmaceutical Sciences
Pfizer, Inc.
Watch now
Take an in-depth look at the Orbitrap Exploris MX mass detector
Interact with this unique video to learn more about this system and how it can be the perfect fit for your laboratory.
Watch now
On this page
Highlights of Orbitrap Exploris MX mass detector
Simplicity for scientists and technicians
Simplify tasks and reduce errors for more ‘right first time’ analyses with Chromeleon CDS, with built in methods, control, data processing, and management. The suite of smart tools makes your work faster and easier, while ensuring reproducible, high-quality mass monitoring.
Accelerate productivity
High uptime boosts productivity. Rapid “set and forget” calibration procedures provide consistent mass stability for at least four weeks at prescribed conditions. Instrument status monitoring and optimized pressure control alert users when maintenance is needed, avoiding unnecessary downtime and repeat analyses.
Compliance for competitive advantage
Designed for data integrity, data security, and compliance, fully scalable Chromeleon CDS provides comprehensive technical controls to give you a competitive edge in meeting global regulatory requirements.
Consistent results deliver confidence
Robust and reliable performance from system to system and site to site ensure confidence in results used to make critical decisions. Industry-proven Orbitrap technology, now purposely designed for mass monitoring in future-proof lab operations.
Seamless method transfer to MAM
Easy transfer of sophisticated Multi-Attribute Method (MAM) assays from development to manufacturing bridges the gap between development and QC.
Dedicated, global support team
The dedicated, global Thermo Scientific MAM team maximizes your productivity and confidence with installation, training, and fast service and support. Standardized system performance evaluation test (SET) and installation qualification/operational qualification (IQ/OA) thoroughly assess system performance with comprehensive installation and post-maintenance acceptance criteria.
Accelerate MAM deployment
Seamless knowledge and method transfer
Benefit from the end-to-end Thermo Scientific MAM 2.0 workflow approach to comprehensive characterization and monitoring of biotherapeutics from development to commercialization. Orbitrap Exploris 240 mass spectrometer methods, established during development, seamlessly transfer to the Orbitrap Exploris MX detector in manufacturing or by contract partners. In addition to full integration into the high-resolution MAM 2.0 workflow, the usability and low cost of the Orbitrap Exploris MX detector make it the perfect fit for high-throughput QC and contract labs.
Users can identify and evaluate relevant product quality attributes (PQAs) via high-throughput analytics provided by Thermo Scientific BioPharma Finder software. Connected labs can access data from development through QC using MAM 2.0 powered by Thermo Scientific Ardia Platform. And, PQAs can be monitored throughout the entire drug development process using the MAM target peptide workbook built from the list of identified peptides upon incorporation into a Chromeleon processing method. The processing method along with the instrument method, injection sequence, view setting and report template can be stored into a MAM eWorkflow for easy and direct transfer to QC via Chromeleon CDS Enterprise software.
Video Player is loading.
Current Time 0:00
Duration 1:07
Loaded: 14.49%
Stream Type LIVE
Remaining Time 1:07
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    MAM 2.0 offers:
    • Fast decision making based on high-confidence PQAs obtained using industry-leading HRAM Orbitrap technology
    • Seamless knowledge and method transfer across instruments, departments, and sites using compliance-ready, Chromeleon CDS enterprise software, and Ardia Platform
    Thermo Scientific MAM 2.0
    The end-to-end MAM solution for a fully connected, compliance-ready platform. MAM 2.0 includes everything you need to bring your next biotherapeutic candidate straight through to breakthrough.
    Learn about MAM 2.0 ›
    Orbitrap Exploris 240
    Consistent performance between the Thermo Scientific Orbitrap Exploris 240 mass spectrometer and Orbitrap Exploris MX detector ensures seamless method transfer from research and development through to manufacturing and QC.
    Learn about Orbitrap Exploris 240 ›
    Applications performed by the Orbitrap Exploris MX detector
    Compliance-ready MAM monitoring of peptide quality attributes
    Post-translational modifications (PTMs) and process-induced modifications can occur during upstream and downstream bioprocessing, formulation, and storage. Any PTMs impacting safety or efficacy are critical quality attributes (CQAs) that must be monitored and controlled. Liquid chromatography mass spectrometry (LC-MS) MAM offers simultaneous, direct, site-specific identification and quantitation of a broad set of peptide CQAs. With the ability to replace multiple assays used during drug development and manufacturing, MAM can accelerate approvals, increase product and process knowledge, reduce development time, and reduce lot release assay cost and time.
    The Orbitrap Exploris MX detector adds unprecedented simplicity, robustness, and HRAM-enabled confidence to MAM workflows. In addition to operational productivity, integrated Chromeleon CDS provides superior MS data-processing tools and comprehensive controls for regulated labs.
    Why MAM?
    Embraced by industry and regulatory leaders, MAM is key to the future of high-quality biotherapeutics. MAM adoption across all stages of development and manufacturing enables efficient and comprehensive understanding of processes and products by replacing many assays with one. Thermo Fisher Scientific has developed a fit-for-purpose MAM solution—now drug developers have the partner they need to aid them in deploying MAM.
    Wide applicability
    The enhanced Orbitrap Exploris MX-based MAM workflow is applicable for monitoring PTMs such as glycosylation, deamidation, oxidation, peptides for sequence confirmation, and glycan profiling.
    High-sensitivity mass determination up to m/z 8000
    With optional mass range up to m/z 8000 and HRAM data quality, the Orbitrap Exploris MX detector is the ideal choice for rapid, high-confidence molecular weight profiling, even for complex mAb glycoforms at low concentrations. Confirming the molecular weight of large intact proteins under native and denaturing conditions is essential to biotherapeutic production and QC.
    Why analyze native proteins
    Analysis under non-denaturing conditions allows characterization of proteins in their near-native biological states. Previously, obtaining this information required multi-step labor intensive isolation and purification. Using native MS, quality attributes can be directly assessed with minimal need for sample preparation, rapidly providing mass information for all proteins in a sample.
    Chromeleon CDS streamlines molecular weight determination
    The automated parameter optimizer makes it easy to obtain trusted results. Complementary deconvolution algorithms optimized for isotopically resolved and unresolved data provide accurate molecular weights for intact proteins, in both denaturing and native conditions, and at low abundances.
    Intact mass monitoring of oligonucleotide sequences
    The Orbitrap Exploris MX detector is an ideal solution for sensitive and robust intact mass monitoring of oligonucleotide sequences and their low-level production related impurities. Compared to low-resolution instruments which only provide average molecular weight, HRAM data enables resolution of very closely related intact oligonucleotide sequences. Unlike software that relies on traditional peak picking algorithms, Xtract and ReSpect deconvolution algorithms implemented in BioPharma Finder software make use of the Sliding Window feature intact and provide mass deconvolution on a chromatographic time scale to identify failed oligonucleotide sequences and process related impurities, even at low levels or in poorly resolved chromatographic peaks.
    Complexity, diversity, and size of oligonucleotides and potential impurities makes analyses challenging
    HRAM LC-MS is the technique of choice due to its mass accuracy, reproducibility, robustness, sensitivity, and speed. HRAM capability is indispensable in verifying that an oligonucleotide has the expected molecular weight.
    BioPharma Finder software uniquely handles oligonucleotide nomenclature and custom modifications
    BioPharma Finder software provides an intuitive set of tools to support sequence modifications at each subunit of the oligonucleotide, including the nucleoside base, backbone linker, and 2’ ribose positions. Custom building blocks can easily be inserted into existing sequences or utilized for future sequence creation. Chemical modifications can also be created and searched to identify unexpected or variable modifications of the target sequence.
    Oligonucleotide intact mass confirmation analysis of a 38mer oligonucleotide
     Click image to enlargeOligonucleotide intact mass confirmation analysis of a 38mer oligonucleotide at a resolution setting of 120,000. (A) Full-MS spectrum with a zoom into the -8 charge state is shown (insert) as well as the (B) resulting spectrum after deconvolution with a zoom into the lower abundant signals (insert). (C) Exemplified, signals in the deconvoluted spectrum are assigned to the masses in the displayed BioPharma Finder result list (colored asterisks), showing the Full length product (FLP) (highest abundance; green asterisk) as well as identified low level impurities.
    Software solutions to streamline data analysis
    Chromeleon Chromatography Data System (CDS)
    Simplify your workflow and get superior insights into your data. Benefit from ease of use, comprehensive instrument control, automated data analysis, and flexible reporting.
    Learn about CDS ›
    BioPharma Finder software
    Complete biotherapeutic characterization includes intact mass analysis, peptide mapping, oligonucleotide analysis, top- and bottom-down analysis or multi-attribute method (MAM) workflows.
    Learn about BioPharma Finder software ›
    Orbitrap Exploris MX mass detector ordering guide
    Services