Thermo Scientific™ Veterinary Microbiology Solutions
More speed, more simplicity, more support for animal diagnostic and susceptibility testing
Designed with veterinary laboratories in mind, Thermo Scientific™ Veterinary Microbiology Solutions include a complete range of high quality culture media, identification and susceptibility testing products, and automated Johne’s Disease instrumentation and reagents.
From standard, veterinary-specific solutions to custom formularies and fully automated instrumentation, no one is more committed to bringing more to animal testing.
Request info
Flexible, customizable veterinary antimicrobial susceptibility testing (AST)
The Thermo Scientific™ Sensititre™ ID/AST System includes over 40 veterinary-specific antimicrobials, for susceptibility testing with true MIC results, and superior quality and reproducibility. Combined with modular equipment options, the Sensititre system allows you to create the ultimate test program tailored to your veterinary laboratory’s requirements.
Diagnostic Systems
As the system of choice for global surveillance programs including National Antimicrobial Resistance Monitoring System (NARMS) coordinated via FDA-CVM, USDA and the CDC, the Sensititre system provides standardized results for confident veterinary susceptibility testing.
See our complete offering of antimicrobials and standard MIC plates for veterinary AST, as well as our full range of AST solutions.
Customize AST to your unique needs
Choose from over 40 specific veterinary antimicrobials including Gamithromycin to design a Thermo Scientific™ Sensititre™ Custom Plate tailored to your formulary, dilution ranges and patient population. Design for results based on real-time resistance trends and local epidemiology, to improve clinical success and effectively manage resistance.
Four simple steps to design your custom AST plate
Meet CLSI recommendations for equine AST
Perform accurate susceptibility testing using the Thermo Scientific™ Sensititre™ Equine MIC Plate, designed specifically for equine patients.
Learn more 
Confidently meet requirements for AST surveillance
Confidently meet requirements for AST surveillance in accordance with the European Food Safety Authority (EFSA)
In a scientific report published by EFSA, the technical specifications of the harmonised monitoring and reporting of antimicrobial resistance in Salmonella, Campylobacter and indicator commensal Escherichia coli and Enterococcus spp. bacteria are addressed. Through the availability of specially designed Sensititre™ AST Plates, you can test for resistance in these organisms with confidence and assurance.
Read SmartNote
Simplify AST for avian and poultry samples
Simplify AST for avian and poultry samples
Utilize the antimicrobials and dilutions you need for avian and poultry samples with our species-specific Thermo Scientific™ Sensititre™ Avian MIC Plate.
Learn more
Bring efficiency through best practice to microbiology quality control testing
Bring efficiency through best practice to microbiology quality control testing
Safeguarding the purity and identity of quality control organisms through proper maintenance is critical to achieving accurate control results for culture media and reagents. Our guide highlights industry guidelines and best practices to ensure compliance to standards, as well as approaches to reduce the time spent maintaining frozen stock cultures.
Download Stock Culture Maintenance Protocol
Related products: