A more accurate and reliable presumptive test
Bring the accuracy and reliability of a lab-proven technology directly to the scene with the Thermo Scientific TruNarc Handheld Narcotics Analyzer. The TruNarc analyzer easily and quickly tests for over 530 substances, including narcotics, stimulants, depressants, hallucinogens and analgesics.
  • Library regularly updated to include emerging drug threats
  • Requires no direct contact with most substances
  • Delivers clear, real-time results for presumptive evidence
  • Provides automated, tamper-proof records with scan results, including time-and-date stamps to help expedite prosecution.
First Responder Fridays - Fox25 Boston - Thermo Fisher Scientific
FOX 25 segment for safety and security instruments. CAD has a release to use this externally.
Video Player is loading.
Current Time 0:00
Duration 1:59
Loaded: 0%
Stream Type LIVE
Remaining Time 1:59
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextFirst Responder Fridays - Fox25 Boston - Thermo Fisher Scientific
        FOX 25 segment for safety and security instruments. CAD has a release to use this externally.
      2. Now Playing
        Up NextThermo Fisher Scientific - Javier Pena - TruNarc
        Retired DEA Agent Javier Pena gives a testimonial regarding the Thermo Fisher Scientific TruNarc.
      3. Now Playing
        Up NextTruNarc - Stephen Murphy
        Testimonial about TruNarc from retired DEA agent Stephen Murphy.
      4. Now Playing
        Up NextTruNarc - Ernie Batista
        Testimonial about TruNarc from retired DEA agent Ernie Batista.
      5. Now Playing
        Up NextThermo Scientific TruNarc Product Video - 2018
        Product video detailing features of the Thermo Scientific TruNarc Handheld Narcotics Analyzer.
    TruNarc case studies
    TruNarc Handheld Narcotics Analyzer with two bags of powder displaying a scan result of fentanyl.
    Efficient, economical, safe
    Agencies across the United States that deploy the TruNarc Handheld Narcotics Analyzer are seeing immediate benefits:
    • Even little-known substances can be identified almost instantly in the field
    • The need for law enforcement officers to handle potentially lethal narcotics is greatly reduced
    • Valuable lab time is freed up for higher priority cases
    • Reduced demand for lab testing delivers significant cost savings
    TruNarc Handheld Narcotics Analyzer FAQs
    Police officer holding TruNarc Narcotics Analyzer
    Want more information about how the TruNarc Handheld Narcotics Analyzer uses Raman technology to identify multiple controlled substances including narcotics, synthetic drugs, cutting agents, and precursor chemicals in a single test, without direct contact for most samples?