Overview
The Thermo Scientific MAX-Bev CO₂ Purity Monitoring System is a fully automated system designed to analyze carbonation streams for all gaseous impurities using state-of-the-art FTIR technology.
The MAX-Bev CO₂ Purity Monitoring System is capable of simultaneous accurate measurement of CO₂ absolute purity and more than 20 impurities down to ppb levels, as specified by the International Society of Beverage Technologists (ISBT). The elegant design incorporates complete control of the input process gas streams from all points of the carbonation process (delivery tanker, pre- and post- filtration), along with zero gas and validation gas. 
Thermo Scientific MAX-Bev™ 2.0 CO2 Purity Monitoring System
MAX-Bev CO₂ Purity Monitoring System features
With the MAX-Bev CO₂ Purity Monitoring System, total software control is maintained via the large front panel touchscreen or keyboard. Common communication protocols for data logging are used for easy incorporation into existing plant designs, and impurities observed in the Bev-gas are quickly detected, speciated, and quantified. Also offering easy-to-generate Certificates of Analysis, the MAX-Bev CO₂ Purity Monitoring System is the ideal system whenever and wherever carbonation gases must be analyzed for impurities.
Video: See how Coca-Cola is using the MAX-Bev to create "the Perfect Sip"
For Research Use Only. Not for use in diagnostic procedures.