What are recombinant antibodies?
Unlike traditional monoclonal antibody manufacturing, recombinant antibodies are produced in vitro as an expression of a defined antibody sequence which assures equivalent bioactivity and specificity across time and lots. They provide a solution to concerns like cell-line drift, lot-to-lot variation, and specificity in complex biological systems.
Recombinant antibodies can be genetically manipulated to offer higher quality and better performing reagents. They are known for their versatility by being suitable for a wide range of applications and being utilized in large scale or long-term experiments. Recombinant antibodies are available in diverse formats such as conjugated and purified forms with multiple species backbones, isotypes, and sub-types.
Video Player is loading.
Current Time 0:00
Duration 1:28
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:28
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Why choose recombinant antibodies?
    We are addressing the challenges faced by researchers in proteomics, human health, and drug discovery by offering highly validated recombinant antibodies.
    • Specificity: Our antibodies undergo rigorous advanced verification through one or more different methods like relative expression, knockout analysis, and cell treatment. Currently, the portfolio houses thousands of advanced verified antibodies and we are in continuous pursuit to increase these numbers.
    • Exceptional performance and sensitivity: Recombinant antibodies are available in different engineered backbones, resulting in enhanced sensitivity and supporting wider application coverage, without compromising on specificity.
    • Versatility and scalability: We offer antibodies raised in multiple species, validated using a rigorous process to suit various research needs and a wide range of applications. Recombinant production methods allow for large scale lot manufacturing.
    • Reproducibility: Recombinant monoclonal antibodies exhibit exceptional lot-to-lot reproducibility because banked DNA does not exhibit a genetic drift—a phenomenon where hybridomas change expression patterns over time.
    Video Player is loading.
    Current Time 0:00
    Duration 0:59
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 0:59
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Antibody selection guide
      Recombinant Rabbit Monoclonal Antibodies Recombinant Rabbit Polyclonal AntibodiesSuperclonal Recombinant AntibodiesVHH AntibodiesConventional Monoclonal AntibodyConventional Polyclonal Antibody
      Type
      Primary antibodyPrimary antibodySecondary antibodyPrimary and secondary antibodyPrimary antibody Primary antibody
      DescriptionDerived from rabbit monoclonal antibody-producing cell lines.Derived from a mixture of recombinant rabbit antibodies.Derived from a pool of recombinant secondary antibodies.Retain the antigen binding region of the heavy chain domain of a conventional antibody, but do not have the other light variable chain domains.Derived from fusing host animal spleen cells with myeloma cells to produce genetically homogeneous antibodies against a single epitope.Derived from the inoculation of a suitable mammal, such as a mouse, rabbit, or goat.
      AdvantagesThese recombinant cloned antibodies are not susceptible to cell-line drift or lot-to-lot variation, allowing for peak specificity and performance.Like conventional polyclonal antibodies, the recombinant polyclonal offers enhanced sensitivity for low abundance targets, while ensuring consistency in chain populations across lots, eliminating biological variability.Provide the epitope-precision of monoclonal antibodies while achieving the multi-epitope coverage and sensitivity of polyclonal antibodies.Roughly 1/10th the size of conventional IgG antibodies, providing better tissue penetration, high labeling density and better image resolution.Offer specificity to a single epitope and ensure a stable, long-term supply.Enhanced sensitivity for low abundance targets with multi-epitope coverage.
      SensitivityHighHighestHighHighVariable, dependent on epitope availabilityVariable, dependent on production and validation methods
      SpecificityHighHighHighVariable, dependent on epitope distribution among various clones, production, and validation methodsVariable, dependent on epitope availability and validation methodsVariable, dependent on epitope distribution among various clones, production, and validation methods
      Lot-to-lot consistencyHighestHighHighVariable, animal variability and purification processAberrant transcripts can be acquired from myeloma fusionsVariable, animal variability and purification process 
      Epitope variabilityNoneNoneNoneNoneNone, but can be acquired over timeVariable across lots
      ProductionDeveloped by isolating and cloning the specific antibody heavy and light chain DNA sequences.Comprised of a mixture of recombinant antibodies co-expressed from a library of heavy and light chains.Produce and characterize numerous individual recombinant antibodies. Then, carefully screen, select, and pool specific antibodies.Produced via recombinant techniques in bacterial or alternative animal-free expression systems.Hybridoma generation via immunization methods and expression via tissue culture or ascites production.Immunization via a host animal (positive and/or negative selection) of antibodies from the serum.
      Video Player is loading.
      Current Time 0:00
      Duration 1:03
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 1:03
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Learn more about how recombinant antibodies are produced.
        Video Player is loading.
        Current Time 0:00
        Duration 1:12
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:12
        1x
          • Chapters
          • descriptions off, selected
          • en (Main), selected
          Explore our extensive portfolio of recombinant antibodies.
          Featured webinar
          Highly specific antibodies for Parkinson’s Disease focused on recombinant technologies
          Learn about the importance of highly validated and reproducible recombinant antibodies and will the recent testing of Parkin antibodies and how they relate to the study of Parkinson's Disease.
          Register on wiley to watch on demand
          Related products
          Advanced Verification
          Thermo Fisher Scientific is committed to adopting higher validation standards for the Invitrogen antibody portfolio. We have implemented additional specificity tests to help ensure high confidence levels in our products. You can identify the products that have already undergone this testing with the Advanced Verification badge, shown above. This badge can be found in antibody search results and at the top of product webpages. The data supporting the Advanced Verification status can be found in the product specific data galleries. To learn more about our testing standards, please visit Invitrogen Antibody Verification.
          * The use or any variation of the word “validation” refers only to research use antibodies that were subject to functional testing to confirm that the antibody can be used with the research techniques indicated. It does not ensure that the product(s) was validated for clinical or diagnostic use.
          For Research Use Only. Not for use in diagnostic procedures.
          Stylesheet for Classic Wide Template adjustments
          Hero banner dark gradient overlay
          For Research Use Only. Not for use in diagnostic procedures.