Nalgene Containment Solutions
Empowering bioprocessing excellence
With 75 years of trusted single-use bioprocessing containment solutions, the Nalgene portfolio is designed with precision and innovation. Our products provide excellent containment and protection for your valuable biopharmaceutical processes.
We offer a comprehensive product portfolio of single-use bioprocessing solutions, so you can choose the products that meet your specific needs and budget. Rest assured in our product features and the peace of mind that comes from using a single supplier with a global supply chain.
Nalgene bottles and carboys
Nalgene bottles and carboys
Nalgene production bottles and carboys offer leakproof* storage for sensitive liquids, buffers, media, and more. They are available in PC, PETG, PP, LDPE, HDPE, FEP, and PFA in volumes ranging from 5 mL to 50 L. Extensive regulatory documentation helps ensure that the products meet your demanding biotechnology requirements and can easily integrate into your processes.
Be particular in choosing the right solution
Application
Standard
Low particulate
Ultralow particulate
Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
Ideal for storage and/or transportation of pH-sensitive materials, such as biological drug substance
Key features
Sterile and non-sterile options
Manufactured from a high-quality resins
Leakproof closures*
Triple-layer packaging (two heat seals and one carton liner)
Produced in an ISO 7 clean room
Compliant with USP <788> 
Certified to 1/3 of the allowable USP <788> particulate limits 
Washed with WFI (water for injection)**
Inspected, assembled, and packaged in ISO 5 (Class 100) clean rooms with ISO 4 (Class 10) hoods and full gowning for operators
* Our guarantee for a leakproof seal is subject to our standard product warranty, as set forth in the Thermo Fisher Scientific Terms and Conditions of Sale. Our products are leakproof under use with our required specifications at ambient temperature and pressure when used with their corresponding closures, when handled properly and secured by the user properly. However, to ensure safe usage, customers are advised to test our containers and closures under conditions of their planned application.
** Not all Platinum Certified Clean ultralow particulate bottles products require WFI wash to meet particulate claims
Nalgene bioprocessing containment accessories
Whether you are looking for reliable closures to secure your containers or durable tubing for various applications, we have got you covered. Choose from a wide range of Thermo Scientific Nalgene closures and tubing.
Nalgene bottle and carboy assembly systems
Fluid transfer systems play an important role in bioprocessing but can easily cause inefficiencies; multiple components need to be sourced, purchased, inventoried, and assembled. Our fit-for-purpose Thermo Scientific Nalgene Bottle and Carboy Assembly Systems are designed to reduce the burden of managing individual components and to enabled in-house system assembly and validation. 
Available in a variety of resins and sizes 
  • Polycarbonate (PC) 
  • Polyethylene terephthalate glycol (PETG)
  • High-density polyethylene (HDPE)
  • 125 mL to 20 L
Closures and components to meet your needs
  • Configuration available with 2- or 3-port closure
  • Comprehensive list of components available for process-specific assembly
Supply assurance and risk mitigation
  • Global manufacturing network, with in-region manufacturing built to ensure supply chain resiliency, redundancy, and excellent service
Standard Single-Use Bottle Assembly Systems
  • Easy-to-order configured solutions include Nalgene bottles with preconfigured cap sets and are delivered gamma-irradiated for use right out of the package.
  • Available off the shelf < 1 week
Download datasheet >
Customized Single-Use Bottle Assembly Systems
  • Featuring the BioTitan Retention Device, designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point***
  • Available off the shelf < 6 weeks
Contact us to learn more about how a custom bottle assembly system can benefit your laboratory.
Download infographic >
Customized Single-Use Carboy Assembly Systems 
  • Featuring the BioTitan Retention Device
  • Available off the shelf < 6 weeks
  • Available in multiple sizes: 10 L, 15 L, 20 L, 25 L, 50 L 
  • Custom low particulate washing available to help secure your substances with ultralow particulate options
Ask a specialist about how a custom carboy assembly can meet your application needs. 
Contact a specialist > 
*** No leaks reported since 2021 launch as of the publishing this web page May 2024
Nalgene glassware and vials
Low particulate
  • Choose from a large range of standard particle-certified containers assembled to contain as few as 5 particles per milliliter >0.5 microns
  • Non-standard containers may be custom cleaned on request
Total Organic Carbon (TOC) Glassware
  • Simplify and reduce the cost of cleaning validations. Low background is perfect for preparation and storage of standards. 
  • TOC-certified containers are available in several sizes, including the popular 40 mL autosampler vials–cleaned, certified, and ready-to-use
  • TOC water and other specialty waters are packaged in precleaned amber glass containers and are ideal for dilution of TOC testing samples
Sterile vials
  • Put your trust in proven products developed by industry experts for performance, usability, and cost-effectiveness
  • Glass vials and other containers are offered in a variety of sizes that have been specially prepared to meet endotoxin levels of less than 0.06 EU/mL
Silanized glassware
  • Surface deactivation treatments such as silanization may play an important role in preserving the integrity of certain materials or extracts stored in glass containers
  • It also helps to avoid alkalinization of contents which may occur as carbonates leach from the glass with normal weathering
Low particulate washing services
We understand the critical importance of maintaining clean and contaminant-free containers and component parts in the biopharma and vaccine industries. Our cleaning service offers unmatched expertise and state-of-the-art facilities to ensure the highest standards of cleanliness and compliance.
  • Comprehensive cleaning solutions: Our cleaning service is designed to meet the unique needs of CMO/CDMO and biotech companies. We have the capability to clean glass or plastic containers, stoppers, closures, or other component parts up to 50 L in size.
  • Quality assurance: Our cleaning processes are backed by rigorous quality control measures to ensure that every cleaned item meets the highest standards of cleanliness and regulatory compliance.
  • Risk mitigation: Our cleaning service helps reduce the risk of contamination, thereby safeguarding the integrity and safety of your biopharma and vaccine products.
  • Increased efficiency: Outsourcing your cleaning needs to us enables you to focus on your core operations, improving overall efficiency and productivity.
Nalgene tanks and tank liners
Designed for easy installation and maintenance, our Nalgene tanks and tank liners are cost-effective solutions that provide long-lasting performance, helping you optimize your fluid storage processes and minimize operational risks.
Nalgene containment solutions selection guide
Looking for reliable containment to protect your high-quality products in storage, transport, and sampling? Made from high-quality resins, Nalgene production bottles and carboys offer leakproof* storage for sensitive liquids, buffers, media, and more. Nalgene containers help ensure your substances stay secure, with low and ultralow particulate options available, addressing particulate level (USP <788>), endotoxin level, and sterility requirements.
Additional bioprocessing resources
As a trusted partner, we are committed to supporting your journey toward innovative biotech solutions.
Accelerate your therapeutic monoclonal antibody development and manufacturing program.                             
Learn, evaluate, connect.   
Connect with a Bioprocessing representative or request more info.                                                               
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.