A scientist working with a HyPerforma single-use mixer (S.U.M.)
The Thermo Scientific HyPerforma Single-Use Mixer (S.U.M.) has been designed using a proven engineered mixer platform design and offers a more ergonomic and efficient user experience for upstream and downstream mixing applications. The complete line of HyPerforma S.U.M.s includes 50, 100, 200, 500, 1,000, and 2,000 L sizes that helps ensure consistent scalability while providing a number of application usages from powder media and buffer hydration to advanced downstream harvest and intermediate pooling steps.
The Touchscreen Console for HyPerforma S.U.M. provides superior integrated sensor monitoring, automated mixing recipes, and pump control for pH and saline titration and for automatic fill and harvest of the S.U.M.
In bioproduction, mixing technologies can be found completing vital tasks across the entire workflow, and thus mixing equipment should be selected to best meet the demands of an increasingly complex industry. Enhance your upstream and downstream processes with the Touchscreen Console for S.U.M. The Touchscreen Console provides the user with advanced sensor monitoring and user-defined automated mixing recipes. The Touchscreen Console will help improve yields, shorten process times, and optimize workflows.
Key advantages
Customized module user interface allows for superior process monitoring and automation. Modules include:
  • Agitation (mixing rpm)
  • Mass (utilization of load cell readings)
  • Temperature (1 or 2 RTDs)
  • Timer
  • pH (1 or 2 readings)
  • Conductivity (1 or 2 readings)
  • BPC pressure (automated BPC air fill)
  • Liquid pressure (pump control based on liquid pressure in lineset)
  • Pump control (up to four pumps, used for fill, harvest, and bolus control)
  • Automated and metered
  • Fill and harvest
  • Recipes (user defined process automation)
  • Auxiliary input
As an engineering to order product, the HyPerforma S.U.M. with touchscreen console may be fully integrated with either Thermo Scientific HyPerforma G3 Bioprocess Controllers, or controllers from other manufacturers, providing an open-architecture mixing solution configured to your unique requirements.
System highlights
  • Touchscreen Console—enhanced user interface provides monitoring and control of the mixing parameters
  • Unique proprietary mixing system that provides conventional overhead mixing while maintaining sterility and integrity of the S.U.B. BPC
  • Mixer vessel offered with non-jacketed (no heat transfer) or water jacket (vessel side and bottom) for integration with external temperature control unit
  • Single-use contact surfaces for mixing, venting, and temperature sensing
  • Conical vessel bottom for better drainage and less hold-up volume
  • Open cart frame for easier cleaning
  • Load cells weighing systems enable accurate batch weight monitoring
Key features
  • Adjustable powder hanger that fits 1 kg, 5 kg, and 25 kg Powdertainer BPCs
  • Cable management system provides greater organization for electrical cables, tubing, and linesets while making the unit easier to clean
  • Dedicated adjustable tool holder keeps necessary tools available for convenience
HyPerforma S.U.M.s of varying sizes, including 50, 100, 200, 500, 1,000, and 2,000 L
Critical upstream application steps:
  • Media preparation
  • Final formulation steps
  • Buffer preparation
  • Harvest vessels
  • Large-volume mixing (up to 2,000 L)
Critical downstream application steps:
  • Pooling and liquid transfer
  • Product suspension
  • Mixing and storing multiple batches
  • Buffer preparation
  • Viral inactivation
  • Acid/base titration
In bioproduction, mixing technologies can be found completing vital tasks across the entire workflow, and thus mixing equipment should be selected to best meet the demands of an increasingly complex industry. Enhance your upstream and downstream processes with the Touchscreen Console for S.U.M. The Touchscreen Console provides the user with advanced sensor monitoring and user-defined automated mixing recipes. The Touchscreen Console will help improve yields, shorten process times, and optimize workflows.
Key advantages
Customized module user interface allows for superior process monitoring and automation. Modules include:
  • Agitation (mixing rpm)
  • Mass (utilization of load cell readings)
  • Temperature (1 or 2 RTDs)
  • Timer
  • pH (1 or 2 readings)
  • Conductivity (1 or 2 readings)
  • BPC pressure (automated BPC air fill)
  • Liquid pressure (pump control based on liquid pressure in lineset)
  • Pump control (up to four pumps, used for fill, harvest, and bolus control)
  • Automated and metered
  • Fill and harvest
  • Recipes (user defined process automation)
  • Auxiliary input
As an engineering to order product, the HyPerforma S.U.M. with touchscreen console may be fully integrated with either Thermo Scientific HyPerforma G3 Bioprocess Controllers, or controllers from other manufacturers, providing an open-architecture mixing solution configured to your unique requirements.
System highlights
  • Touchscreen Console—enhanced user interface provides monitoring and control of the mixing parameters
  • Unique proprietary mixing system that provides conventional overhead mixing while maintaining sterility and integrity of the S.U.B. BPC
  • Mixer vessel offered with non-jacketed (no heat transfer) or water jacket (vessel side and bottom) for integration with external temperature control unit
  • Single-use contact surfaces for mixing, venting, and temperature sensing
  • Conical vessel bottom for better drainage and less hold-up volume
  • Open cart frame for easier cleaning
  • Load cells weighing systems enable accurate batch weight monitoring
Key features
  • Adjustable powder hanger that fits 1 kg, 5 kg, and 25 kg Powdertainer BPCs
  • Cable management system provides greater organization for electrical cables, tubing, and linesets while making the unit easier to clean
  • Dedicated adjustable tool holder keeps necessary tools available for convenience
HyPerforma S.U.M.s of varying sizes, including 50, 100, 200, 500, 1,000, and 2,000 L
Critical upstream application steps:
  • Media preparation
  • Final formulation steps
  • Buffer preparation
  • Harvest vessels
  • Large-volume mixing (up to 2,000 L)
Critical downstream application steps:
  • Pooling and liquid transfer
  • Product suspension
  • Mixing and storing multiple batches
  • Buffer preparation
  • Viral inactivation
  • Acid/base titration
Solutions utilizing the HyPerforma S.U.M. with Touchscreen Console
Overview HyPerforma SUM Touchscreen Console
Functionality and ease-of-use for consistent scalability across a range of applications from powder media and buffer hydration to downstream harvest and intermediate pooling steps.
Video Player is loading.
Current Time 0:00
Duration 4:34
Loaded: 0.00%
Stream Type LIVE
Remaining Time 4:34
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextOverview HyPerforma SUM Touchscreen Console
        Functionality and ease-of-use for consistent scalability across a range of applications from powder media and buffer hydration to downstream harvest and intermediate pooling steps.
      2. Now Playing
        Up NextUpstream HyPerforma SUM Touchscreen Console
        Functionality and ease-of-use for consistent scalability across a range of applications from powder media and buffer hydration to downstream harvest and intermediate pooling steps.
      3. Now Playing
        Up NextDownstream HyPerforma SUM Touchscreen Console
        Functionality and ease-of-use for consistent scalability across a range of applications from powder media and buffer hydration to downstream harvest and intermediate pooling steps.
    Related products
    Thermo Scientific TruBio, TruChrom, and TruPur automation software platforms are capable of controlling bioreactor systems from research to commercial manufacturing.
    Thermo Scientific HyPerforma G3 Bioprocess Controllers provide ease of data transferability and scalability throughout your process and incorporates innovative features that enhance productivity, quality, and reproducibility.
    Ordering information
    Hyperforma S.U.M. hardware
    Catalog # Name Size Price (USD) Qty
    SUM0050.9001 HyPerforma™ Single-Use Mixer, 50 L, non-jacketed, DC motor, touchscreen console, with load cells 50 L -
    SUM0050.9002 HyPerforma™ Single-Use Mixer, 50 L, jacketed, DC motor, touchscreen console, with load cells 50 L -
    SUM0100.9001 HyPerforma™ Single-Use Mixer, 100 L, non-jacketed, DC motor, touchscreen console, with load cells 100 L -
    SUM0100.9002 HyPerforma™ Single-Use Mixer, 100 L, jacketed, DC motor, touchscreen console, with load cells 100 L -
    SUM0200.9001 HyPerforma™ Single-Use Mixer, 200 L, non-jacketed, DC motor, touchscreen console, with load cells 200 L -
    SUM0200.9002 HyPerforma™ Single-Use Mixer, 200 L, jacketed, DC motor, touchscreen console, with load cells 200 L -
    SUM0500.9001 HyPerforma™ Single-Use Mixer, 500 L, non-jacketed, DC motor, touchscreen console, with load cells 500 L -
    SUM0500.9002 HyPerforma™ Single-Use Mixer, 500 L, jacketed, DC motor, touchscreen console, with load cells 500 L -
    SUM1000.9001 HyPerforma™ Single-Use Mixer, 1000 L, non-jacketed, DC motor, touchscreen console, with load cells 1000 L -
    SUM1000.9002 HyPerforma™ Single-Use Mixer, 1000 L, jacketed, DC motor, touchscreen console, with load cells 1000 L -
    SUM2000.9003 HyPerforma™ Single-Use Mixer, 2000 L, non-jacketed, AC motor, touchscreen console, 240 VAC, with load cells 2000 L -
    SUM2000.9004 HyPerforma™ Single-Use Mixer, 2000 L, jacketed, AC motor, touchscreen console, 240 VAC, with load cells 2000 L -
    Bioprocessing resources
    Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: