Discover the exceptional performance of the Attune Flow Cytometer family—Attune NxT, Attune CytPix, and Attune Xenith—engineered to set a new standard in flow cytometry. At the heart of each Attune Flow Cytometer lies our revolutionary acoustic focusing technology, which utilizes sound waves to meticulously align cells within the flow cell. This innovative approach surpasses traditional hydrodynamic methods, resulting in faster processing speeds, exceptional data quality, and increased sample throughput.
The Attune NxT cytometer is the reliable workhorse instrument your lab can rely on for consistent and dependable performance. The Attune CytPix model features a high-speed brightfield camera and AI-driven image analysis software, with recent upgrades including machine learning tools and an intuitive user interface, enabling enhanced data precision, accuracy, and faster insights into sample biology. The Attune Xenith cytometer is a high-parameter spectral analyzer, providing both spectral unmixing and conventional flow capabilities for comprehensive and versatile cell analysis.
Leverage the power of advanced technology and innovation to accelerate your discoveries. Trust in the Attune Flow Cytometer family to deliver the reliability, speed, and accuracy that set your work apart.
Try our configurator tool to design an Attune NxT or CytPix system to fit your research needs.
New Invitrogen Attune Xenith Flow Cytometer
Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis
Attune flow cytometer selection tools
Considerations when purchasing a flow cytometer
Learn about key experimental concepts and flow cytometer components.
Workflow automation options
Pair your Attune flow cytometer with an autosampler for increased workflow efficiency, and add robotic integration for maximum throughput.
FREE—Flow Cytometry Panel Design Service and Panel Builder Tool
Our scientists will design panels for you as many times you need—free of charge, no product purchase needed.
Attune flow cytometer capabilities
Acoustic focusing cytometry
Sound waves enable precise cell alignment, resulting in exceptional data quality and ~10X faster sample flow rates.
Visualization of imaging-enhanced flow cytometry data measurement
Learn how the Edison-award winning Attune CytPix Flow Cytometer integrates imaging with flow cytometry to enhance data precision and accuracy.
High-speed spectral flow cytometry
Combine acoustic focusing technology with high-parameter analysis for flexible spectral unmixing and conventional workflows.
Flow cytometry educational resources
A monthly meetup to talk all things flow cytometry
Join us for 'Let's Talk Flow’—Your go-to series for mastering flow cytometry! Discover best practices shared in bite-sized webinars.
Educational videos and webinars
Learn more from researchers who use Attune Flow Cytometers in their labs.
Flow Cytometry Learning Center
Whether you’re a beginner or an expert, explore our resources to advance your research and expertise in flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
1x1 image pixel for data collection 1x1 image pixel for data collection