Cancer organoid drug screening webinar
Listen to Dr. Christophe Deben of Orbits Oncology share his experience with OncoPro medium in the recent webinar, "Advancing cancer organoid drug screening with novel media systems."
Watch it on demand now
Gibco OncoPro Tumoroid Culture Medium streamlines the culture of patient-derived cancer organoids (tumoroids) to help make physiologically relevant cancer models more accessible. This adaptable culture system is readily integrated into your workflow so you can upgrade your media and maintain your results. Overall, OncoPro medium offers several key benefits to cancer researchers:
Cancer icon
Optimized—OncoPro medium was developed to support expansion of tumoroids without overgrowth of non-malignant cells. As such, this culture system helps ensure preservation of donor-specific characteristics, including mutational profiles and gene expression patterns, in long-term culture.
 Gibco bottle icon
Streamlined—OncoPro medium is a modular media system, consisting of four components that can be supplemented with indication-specific growth factors as needed. Its suspension culture capability introduces a simplified tumoroid culture workflow, helping save time and resources.
Split arrow icon
Adaptable—OncoPro medium is compatible with a suspension or embedded tumoroid workflow, supports scale-up and automation, and sustains tumoroids across multiple cancer indications. OncoPro medium is also compatible with traditional applications and assays.
Customer stories
Erik Goka
"The Gibco OncoPro Tumoroid Culture Medium system allows us to seamlessly integrate tumor organoids into our research and drug discovery processes. With the tumoroid media system we can propagate all our organoid models in the same culture medium system irrespective of the tissue of origin. This media system has been instrumental in allowing us to bring tumor organoids into our lab for everyday use."
Erik Goka, PhD
Vice President of Biology
Revere Pharmaceuticals
Shawn Fahl
"The OncoPro media has greatly accelerated our development of long-term primary tumor culture systems. The media is easy to make, with a minimal amount of separate components compared to other systems, and the protocols are straightforward, allowing the transition from culture initiation to viable long-term cultures to be seamless and quick."
Shawn Fahl, PhD
V.P. Lab Operations, Cell Services & R&D,
BiospecimensDiscovery Life Sciences
Benefits and performance data of OncoPro medium
Discover how OncoPro medium was optimized for tumoroid culture
OncoPro Tumoroid Culture Medium is a novel media system that was specifically developed for the expansion of patient-derived tumoroids. Over 40 conditions were tested during the development of OncoPro to identify the combination of basal media, supplements, and growth factors that would best support long-term maintenance of donor-specific characteristics, including mutational profiles and gene expression patterns.
Figure 1. Design of experiment studies were used to fully optimize the OncoPro medium formulation for tumoroid growth. Combinations of medium components were tested in designed experiments to elucidate main effects and interactions of additives on tumoroid growth. In this example, two tumoroid lines were tested using 25 media recipes, with 1-2 replicate wells per condition. (A) Heat map of number of viable cells detected per media condition as measured by PrestoBlue HS assay. (B) Images of tumoroids in medium optimization experiments. Different conditions were tested on dissociated cells seeded from an identical initial cell pool, and conditions advantageous to tumoroid growth and retention of genetic and phenotypic characteristics were identified. (C) Design of experiment output for tumoroid line growth response to media optimization experimental conditions. Figure adapted from JMP 16 software output.
Figure 2. OncoPro medium selectively expands cancer cells. (A) Marker expression (flow cytometry) of cells freshly isolated from a colorectal tumor resection (never in culture) and tumoroid cells after line establishment (i.e., 7 passages in OncoPro medium). The panel included markers for epithelial-derived cells (EpCAM), colorectal cancer cells (CEACAM), immune cells (CD45), endothelial cells (CD31), and mesenchymal cells (vimentin). Data reveal preferential outgrowth of epithelial colorectal cancer cells over immune or endothelial cells. (B) Representative immunofluorescence image of EpCAM and actin (phalloidin) expression demonstrate that tumoroids are highly enriched for EpCAM-positive tumor cells.
Figure 3. OncoPro medium helps maintain mutational profiles of tumoroid lines over long-term culture. Correlation of germline and somatic single nucleotide variations (SNVs) between early and later-passage tumoroid cultures. Each dot represents the variant allelic frequency (VAF) for one genetic locus covered by the Oncomine Comprehensive Assay v3. Figure adapted from JMP 16 software output.
Understand how OncoPro medium streamlines media formulation and tumoroid culture
OncoPro medium affords researchers workflow versatility by letting users decide if they want to utilize established embedded culture methods or transition to suspension methods to help save both time and resources.
Scientific diagrams showing tumoroid culture methods
Figure 4. The OncoPro medium suspension culture method greatly simplifies tumoroid passaging, reducing hands-on time from 140 minutes for embedded culture to 60 minutes for suspension culture. (A) Schematic of culture method steps for passaging tumoroids in embedded culture. Boxes highlight the pain points of disrupting extracellular matrix (ECM) domes, losing cells trapped in ECM during washing, and pipetting and waiting for ECM domes to solidify, respectively. (B) Schematic of culture method steps for passaging tumoroids in suspension culture, which addresses these pain points due to reduced ECM requirement.
Figure created with BioRender.com.
Table comparing suspension vs embedded culture
Figure 5. Tumoroid cultures are readily scalable with the OncoPro medium suspension culture method. The number and format of culture vessels required to obtain increasing scale of tumoroid cultures are more user- and resource-friendly in suspension culture when compared to traditional embedded culture methods. (A) Suspension methods allow for ready expansion of 100 million cells within one passage, whereas the number of basement membrane extract (BME) domes required in embedded culture would be prohibitive for many users. (B) Average amount of BME required to harvest 100 million cells for 9 tumoroid lines using different culture methods. On average, suspension culture required less BME than embedded culture (mean ± SEM, not significant).
Figure 6. Patient-specific gene expression profiles are highly correlated between samples cultured using the streamlined suspension method and those cultured using traditional embedded methods. Ward hierarchical clustering and Spearman’s ρ correlations of gene expression for a panel of 1423 cancer-related genes across a variety of tumoroid lines representing four different cancer indications. Gene expression assayed by Ion AmpliSeq Transcriptome Human Gene Expression Panel, Chef-Ready Kit. Figures adapted from JMP 16 software output. With the exception of colorectal tumoroid line Z, established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Learn how OncoPro medium functions as an adaptable system
OncoPro Tumoroid Culture Medium Kit is a flexible system designed to address complex research needs. OncoPro medium accommodates multiple cancer indications using a single media kit and the scalable and automation-friendly suspension workflow facilitates smooth transitions to various downstream assays. The absence of Wnt, R-spondin, Noggin, and small molecule inhibitors make OncoPro well-suited for advanced biological studies and screening applications.
Figure 7. OncoPro medium is compatible with four different cancer indications, for both suspension and embedded culture. Morphology of NCI PDMR tumoroid lines expanded in NCI PDMR homebrew media (embedded culture), or in OncoPro medium embedded or suspension culture. Scale bar = 400 µm. Established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Figure 8. OncoPro medium is compatible with four different cancer indications, for both suspension and embedded culture. Cumulative population doublings of cells expanded in OncoPro medium suspension culture (purple), OncoPro medium embedded culture (red), or alternative media system (media system in which the line was derived prior to procurement) embedded culture (green). In general, tumoroids expanded in each culture condition with comparable growth rates. With the exception of colorectal tumoroid line Z, established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Figure 9. OncoPro medium helps maintain patient-specific gene expression profiles from lines derived in alternative media and embedded culture. Principal component analysis (PCA) plot comparing gene expression levels across tumoroid lines and culture conditions. Tumoroid lines were expanded in parallel in multiple media and culture conditions for 2-7 passages post-thaw prior to quantification of expression levels across over >20,000 human RefSeq genes using the Ion AmpliSeq Transcriptome Human Gene Expression Kit. Tumoroid lines tested were 782815-120-R-V1-organoid (Colorectal A), 919269-233-R3-V2-organoid (Colorectal B), 349418-098-R-V2-organoid (Lung), 549293-155-R-V1-organoid (Head and Neck), and 699152-130-R-V1-organoid (Pancreas). With the exception of colorectal tumoroid line Z, established tumoroid models were provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work. Figure adapted from TAC 4.0 software output.
Discover how OncoPro medium was optimized for tumoroid culture
OncoPro Tumoroid Culture Medium is a novel media system that was specifically developed for the expansion of patient-derived tumoroids. Over 40 conditions were tested during the development of OncoPro to identify the combination of basal media, supplements, and growth factors that would best support long-term maintenance of donor-specific characteristics, including mutational profiles and gene expression patterns.
Figure 1. Design of experiment studies were used to fully optimize the OncoPro medium formulation for tumoroid growth. Combinations of medium components were tested in designed experiments to elucidate main effects and interactions of additives on tumoroid growth. In this example, two tumoroid lines were tested using 25 media recipes, with 1-2 replicate wells per condition. (A) Heat map of number of viable cells detected per media condition as measured by PrestoBlue HS assay. (B) Images of tumoroids in medium optimization experiments. Different conditions were tested on dissociated cells seeded from an identical initial cell pool, and conditions advantageous to tumoroid growth and retention of genetic and phenotypic characteristics were identified. (C) Design of experiment output for tumoroid line growth response to media optimization experimental conditions. Figure adapted from JMP 16 software output.
Figure 2. OncoPro medium selectively expands cancer cells. (A) Marker expression (flow cytometry) of cells freshly isolated from a colorectal tumor resection (never in culture) and tumoroid cells after line establishment (i.e., 7 passages in OncoPro medium). The panel included markers for epithelial-derived cells (EpCAM), colorectal cancer cells (CEACAM), immune cells (CD45), endothelial cells (CD31), and mesenchymal cells (vimentin). Data reveal preferential outgrowth of epithelial colorectal cancer cells over immune or endothelial cells. (B) Representative immunofluorescence image of EpCAM and actin (phalloidin) expression demonstrate that tumoroids are highly enriched for EpCAM-positive tumor cells.
Figure 3. OncoPro medium helps maintain mutational profiles of tumoroid lines over long-term culture. Correlation of germline and somatic single nucleotide variations (SNVs) between early and later-passage tumoroid cultures. Each dot represents the variant allelic frequency (VAF) for one genetic locus covered by the Oncomine Comprehensive Assay v3. Figure adapted from JMP 16 software output.
Understand how OncoPro medium streamlines media formulation and tumoroid culture
OncoPro medium affords researchers workflow versatility by letting users decide if they want to utilize established embedded culture methods or transition to suspension methods to help save both time and resources.
Scientific diagrams showing tumoroid culture methods
Figure 4. The OncoPro medium suspension culture method greatly simplifies tumoroid passaging, reducing hands-on time from 140 minutes for embedded culture to 60 minutes for suspension culture. (A) Schematic of culture method steps for passaging tumoroids in embedded culture. Boxes highlight the pain points of disrupting extracellular matrix (ECM) domes, losing cells trapped in ECM during washing, and pipetting and waiting for ECM domes to solidify, respectively. (B) Schematic of culture method steps for passaging tumoroids in suspension culture, which addresses these pain points due to reduced ECM requirement.
Figure created with BioRender.com.
Table comparing suspension vs embedded culture
Figure 5. Tumoroid cultures are readily scalable with the OncoPro medium suspension culture method. The number and format of culture vessels required to obtain increasing scale of tumoroid cultures are more user- and resource-friendly in suspension culture when compared to traditional embedded culture methods. (A) Suspension methods allow for ready expansion of 100 million cells within one passage, whereas the number of basement membrane extract (BME) domes required in embedded culture would be prohibitive for many users. (B) Average amount of BME required to harvest 100 million cells for 9 tumoroid lines using different culture methods. On average, suspension culture required less BME than embedded culture (mean ± SEM, not significant).
Figure 6. Patient-specific gene expression profiles are highly correlated between samples cultured using the streamlined suspension method and those cultured using traditional embedded methods. Ward hierarchical clustering and Spearman’s ρ correlations of gene expression for a panel of 1423 cancer-related genes across a variety of tumoroid lines representing four different cancer indications. Gene expression assayed by Ion AmpliSeq Transcriptome Human Gene Expression Panel, Chef-Ready Kit. Figures adapted from JMP 16 software output. With the exception of colorectal tumoroid line Z, established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Learn how OncoPro medium functions as an adaptable system
OncoPro Tumoroid Culture Medium Kit is a flexible system designed to address complex research needs. OncoPro medium accommodates multiple cancer indications using a single media kit and the scalable and automation-friendly suspension workflow facilitates smooth transitions to various downstream assays. The absence of Wnt, R-spondin, Noggin, and small molecule inhibitors make OncoPro well-suited for advanced biological studies and screening applications.
Figure 7. OncoPro medium is compatible with four different cancer indications, for both suspension and embedded culture. Morphology of NCI PDMR tumoroid lines expanded in NCI PDMR homebrew media (embedded culture), or in OncoPro medium embedded or suspension culture. Scale bar = 400 µm. Established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Figure 8. OncoPro medium is compatible with four different cancer indications, for both suspension and embedded culture. Cumulative population doublings of cells expanded in OncoPro medium suspension culture (purple), OncoPro medium embedded culture (red), or alternative media system (media system in which the line was derived prior to procurement) embedded culture (green). In general, tumoroids expanded in each culture condition with comparable growth rates. With the exception of colorectal tumoroid line Z, established tumoroid models provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work.
Figure 9. OncoPro medium helps maintain patient-specific gene expression profiles from lines derived in alternative media and embedded culture. Principal component analysis (PCA) plot comparing gene expression levels across tumoroid lines and culture conditions. Tumoroid lines were expanded in parallel in multiple media and culture conditions for 2-7 passages post-thaw prior to quantification of expression levels across over >20,000 human RefSeq genes using the Ion AmpliSeq Transcriptome Human Gene Expression Kit. Tumoroid lines tested were 782815-120-R-V1-organoid (Colorectal A), 919269-233-R3-V2-organoid (Colorectal B), 349418-098-R-V2-organoid (Lung), 549293-155-R-V1-organoid (Head and Neck), and 699152-130-R-V1-organoid (Pancreas). With the exception of colorectal tumoroid line Z, established tumoroid models were provided by NCI PDMR. We thank the PDMR and their contributing institutions for their contributions to this work. Figure adapted from TAC 4.0 software output.
Looking for more information about OncoPro Tumoroid Culture Medium or how you can request a sample quote?
Contact us
Explore technical resources demonstrating the performance and application of OncoPro Medium
Application note titleDescription
Engineering patient-derived tumoroids to help advance cancer research
  • Explore advanced methods for overexpressing and downregulating genes in patient-derived colorectal cancer tumoroids using lipofection, electroporation, and lentiviral transduction.
  • Discover how to achieve optimal gene delivery with practical recommendations and troubleshooting tips to maximize payload uptake and maintain cell viability.
Tumoroid line derivation using OncoPro Tumoroid Culture Medium
  • Demonstrates that OncoPro Medium supports derivation of new tumoroid lines from clinical samples, such as tumor resections or patient-derived xenograft (PDX) tissues
  • Addresses factors affecting derivation success rates (e.g., tissue quality, fresh vs frozen samples, etc.)
  • Provides best practices for tissue handling, dissociation, and culture initiation when deriving new tumoroids
Culture of triple-negative breast cancer tumoroid lines in OncoPro Tumoroid Culture Medium
  • Demonstrates OncoPro Medium compatibility with triple-negative breast cancer (TNBC) tumoroids
  • Recommended indication specific growth factors: heat-stable FGF10 and beta-estradiol
  • Maintenance of patient-specific characteristics including morphology, growth rates, and mutational and transcriptional profiles
Enabling tumoroid-based compound screens with OncoPro Tumoroid Culture Medium
  • Highlights a) value of tumoroids over traditional immortalized cell lines for drug screening and b) how OncoPro suspension protocol enables high throughput workflows
  • Addresses practical considerations such as scaling up cultures, ensuring reproducible tumoroid plating, and designing multiplexed assay readouts.
Access five unique posters from our research and development scientists that highlight the key features of OncoPro medium and how it can be applied in certain tumoroid culture workflows such as drug screening and immuno-oncology. 
Poster TitleDescription
Derivation & long-term maintenance of patient-derived tumoroid lines in a defined, serum-free medium
  • Maintenance of growth rates, patient phenotype and genotype for ≥40 passages
  • Support for multiple cancer indications using tissue-specific growth factors
  • Scale up to >1 billion cancer organoid cells with easy-to-use suspension method
Expansion of established patient-derived tumoroids in a novel serum-free, Wnt agonist-free media system
  • Long-term maintenance of mutational status and gene expression levels in both embedded and suspension formats
  • Wnt-related signaling pathways are not differentially activated following culture in Wnt agonist-free OncoPro Tumoroid Culture Medium
How low can you go: Maintenance of tumoroid phenotype with a highly scalable and automation-compatible reduced-ECM suspension culture method
  • Suspension culture using OncoPro medium is compatible with existing tumoroid lines
  • Patient phenotype and genotype in suspension conditions using OncoPro medium are comparable with embedded method
Multiplexed plate-reader based drug screening of 3D-tumoroid models
  • Development and optimization of drug screening assay using tumoroids
  • Drug response comparison of tumoroids and immortalized cell lines 2D and 3D
  • Compound screening application using tumoroids
Engineering patient-derived tumoroids to enable high-throughput screening:Immuno-oncology workflows
  • Establishment of GFP-expressing tumoroid reporter pool
  • Demonstration of NK-cell mediated cytotoxicity against tumoroids
  • Comparison of primary, iPSC-derived, and NK lines cytotoxicity
Long-term maintenance of patient-specific characteristics in tumoroids from six cancer indications
The serum-free Gibco OncoPro Tumoroid Culture Medium enables establishment and maintenance of tumoroid lines from multiple cancer indications, retaining key tumor characteristics and supporting applications in cancer research, drug discovery, and personalized medicine.
Webinar: Engineering patient-derived tumoroids to enable high-throughput screening for immuno-oncology workflows
Patient-derived tumoroids, commonly referred to as cancer organoids, represent a promising avenue as cost-effective and biologically relevant cancer immunotherapy research models. In this study, we developed a proof-of-concept immunotherapy screening platform harnessing tumoroids to validate the cytotoxic activity of Natural Killer (NK) cells. Tumoroids were genetically engineered to create a GFP-expressing reporter pool that could be used to assess killing efficiency using live cell imaging platforms. We were able to demonstrate dose-dependent tumoroid killing with increasing effector-to-target ratios. This innovative approach holds the potential to streamline the engineering of patient-derived tumoroid models, facilitate the development of multiplex-killing assays, and provide an efficient tool for evaluating immunotherapies targeting solid tumors.
Speaker:
Garret Wong photo 
Garrett Wong, MS, Manager of Cell Biology Services, Thermo Fisher Scientific
Learning objectives:
  • Explore the applications of tumoroids in cancer immunotherapy workflows
  • Establish and engineer patient-derived reporter tumoroid models
  • Develop and optimize assays for immunotherapy screens using tumoroid co-cultures
Watch now
Webinar: Advancing cancer organoid drug screening with novel media systems
Explore the latest developments in cancer organoid (tumoroid) technology and the promising implications for drug screening during this two-part webinar. In the first half, Colin Paul discusses the capabilities of OncoPro medium, a novel system that simplifies the use of tumoroids for drug screening applications. This session reviews potential areas of optimization and highlights how OncoPro medium mitigates these challenges. In the second half, Christophe Deben from Orbits leads us through the cutting edge of imaging software. He demonstrates how their technology effectively resolves downstream bottlenecks in high-content image analysis, providing invaluable insights into the translation of tumoroid and cancer organoid technology for high-throughput applications. Watch these presentations and gain a deeper understanding of the current progress and future direction of tumoroid-based drug screening and image analysis techniques.
Speakers:
Dr. Christophe Deben photo
Dr. Christophe Deben,
Ph.D. Oncology and Cancer Biology, Professor Tumor Biology and Organoids, University of Antwerp
Dr. Colin Paul photo
Dr. Colin Paul,
Cell Biology R&D Staff Scientist, Thermo Fisher Scientific
Learning objectives:
  • Learn how cancer organoids can be adopted for high-throughput screening applications
  • Understand areas of optimization in drug screening workflows for improved consistency and reproducibility
  • Learn how image analysis can be improved to better correlate assay results with patient outcomes
Watch now
Explore four workflow presentations from our research and development scientists that provide guidance to further understand tumoroid derivation and tumoroid characterization methodologies.
PresentationDescription
 Migration and invasion assays using tumoroid models
  • Migration and invasion are crucial in physiological and pathological processes, especially in tumor metastasis
  • This presentation outlines methods for migration and invasion assays using tumoroids, and may be adaptable to other study formats
 Preparation of tumoroids for flow cytometry
  • Techniques like immunofluorescence, western blotting, and flow cytometry can characterize tumoroids, but their 3D structure and basement membrane extract (BME) presence can complicate sample preparation for analysis
  • This presentation provides guidance on preparing tumoroids for analysis by flow cytometry
 Preparation of tumoroids for immunofluorescence
  • Immunofluorescence, western blotting, and flow cytometry can characterize tumoroids, but their 3D structure and presence of BME can complicate sample preparation for analysis
  • This presentation helps provide guidance on adapting tumoroids for immunofluorescence imaging
 Tissue dissociation for tumoroid derivation
  • Derivation success rates are highly dependent on a number of factors, including cancer indication, cell proportion and viability, and processing time; not all samples can generate tumoroid lines
  • This presentation provides guidance on deriving tumoroid lines from fresh tumor tissue samples
Application note titleDescription
Engineering patient-derived tumoroids to help advance cancer research
  • Explore advanced methods for overexpressing and downregulating genes in patient-derived colorectal cancer tumoroids using lipofection, electroporation, and lentiviral transduction.
  • Discover how to achieve optimal gene delivery with practical recommendations and troubleshooting tips to maximize payload uptake and maintain cell viability.
Tumoroid line derivation using OncoPro Tumoroid Culture Medium
  • Demonstrates that OncoPro Medium supports derivation of new tumoroid lines from clinical samples, such as tumor resections or patient-derived xenograft (PDX) tissues
  • Addresses factors affecting derivation success rates (e.g., tissue quality, fresh vs frozen samples, etc.)
  • Provides best practices for tissue handling, dissociation, and culture initiation when deriving new tumoroids
Culture of triple-negative breast cancer tumoroid lines in OncoPro Tumoroid Culture Medium
  • Demonstrates OncoPro Medium compatibility with triple-negative breast cancer (TNBC) tumoroids
  • Recommended indication specific growth factors: heat-stable FGF10 and beta-estradiol
  • Maintenance of patient-specific characteristics including morphology, growth rates, and mutational and transcriptional profiles
Enabling tumoroid-based compound screens with OncoPro Tumoroid Culture Medium
  • Highlights a) value of tumoroids over traditional immortalized cell lines for drug screening and b) how OncoPro suspension protocol enables high throughput workflows
  • Addresses practical considerations such as scaling up cultures, ensuring reproducible tumoroid plating, and designing multiplexed assay readouts.
Access five unique posters from our research and development scientists that highlight the key features of OncoPro medium and how it can be applied in certain tumoroid culture workflows such as drug screening and immuno-oncology. 
Poster TitleDescription
Derivation & long-term maintenance of patient-derived tumoroid lines in a defined, serum-free medium
  • Maintenance of growth rates, patient phenotype and genotype for ≥40 passages
  • Support for multiple cancer indications using tissue-specific growth factors
  • Scale up to >1 billion cancer organoid cells with easy-to-use suspension method
Expansion of established patient-derived tumoroids in a novel serum-free, Wnt agonist-free media system
  • Long-term maintenance of mutational status and gene expression levels in both embedded and suspension formats
  • Wnt-related signaling pathways are not differentially activated following culture in Wnt agonist-free OncoPro Tumoroid Culture Medium
How low can you go: Maintenance of tumoroid phenotype with a highly scalable and automation-compatible reduced-ECM suspension culture method
  • Suspension culture using OncoPro medium is compatible with existing tumoroid lines
  • Patient phenotype and genotype in suspension conditions using OncoPro medium are comparable with embedded method
Multiplexed plate-reader based drug screening of 3D-tumoroid models
  • Development and optimization of drug screening assay using tumoroids
  • Drug response comparison of tumoroids and immortalized cell lines 2D and 3D
  • Compound screening application using tumoroids
Engineering patient-derived tumoroids to enable high-throughput screening:Immuno-oncology workflows
  • Establishment of GFP-expressing tumoroid reporter pool
  • Demonstration of NK-cell mediated cytotoxicity against tumoroids
  • Comparison of primary, iPSC-derived, and NK lines cytotoxicity
Long-term maintenance of patient-specific characteristics in tumoroids from six cancer indications
The serum-free Gibco OncoPro Tumoroid Culture Medium enables establishment and maintenance of tumoroid lines from multiple cancer indications, retaining key tumor characteristics and supporting applications in cancer research, drug discovery, and personalized medicine.
Webinar: Engineering patient-derived tumoroids to enable high-throughput screening for immuno-oncology workflows
Patient-derived tumoroids, commonly referred to as cancer organoids, represent a promising avenue as cost-effective and biologically relevant cancer immunotherapy research models. In this study, we developed a proof-of-concept immunotherapy screening platform harnessing tumoroids to validate the cytotoxic activity of Natural Killer (NK) cells. Tumoroids were genetically engineered to create a GFP-expressing reporter pool that could be used to assess killing efficiency using live cell imaging platforms. We were able to demonstrate dose-dependent tumoroid killing with increasing effector-to-target ratios. This innovative approach holds the potential to streamline the engineering of patient-derived tumoroid models, facilitate the development of multiplex-killing assays, and provide an efficient tool for evaluating immunotherapies targeting solid tumors.
Speaker:
Garret Wong photo 
Garrett Wong, MS, Manager of Cell Biology Services, Thermo Fisher Scientific
Learning objectives:
  • Explore the applications of tumoroids in cancer immunotherapy workflows
  • Establish and engineer patient-derived reporter tumoroid models
  • Develop and optimize assays for immunotherapy screens using tumoroid co-cultures
Watch now
Webinar: Advancing cancer organoid drug screening with novel media systems
Explore the latest developments in cancer organoid (tumoroid) technology and the promising implications for drug screening during this two-part webinar. In the first half, Colin Paul discusses the capabilities of OncoPro medium, a novel system that simplifies the use of tumoroids for drug screening applications. This session reviews potential areas of optimization and highlights how OncoPro medium mitigates these challenges. In the second half, Christophe Deben from Orbits leads us through the cutting edge of imaging software. He demonstrates how their technology effectively resolves downstream bottlenecks in high-content image analysis, providing invaluable insights into the translation of tumoroid and cancer organoid technology for high-throughput applications. Watch these presentations and gain a deeper understanding of the current progress and future direction of tumoroid-based drug screening and image analysis techniques.
Speakers:
Dr. Christophe Deben photo
Dr. Christophe Deben,
Ph.D. Oncology and Cancer Biology, Professor Tumor Biology and Organoids, University of Antwerp
Dr. Colin Paul photo
Dr. Colin Paul,
Cell Biology R&D Staff Scientist, Thermo Fisher Scientific
Learning objectives:
  • Learn how cancer organoids can be adopted for high-throughput screening applications
  • Understand areas of optimization in drug screening workflows for improved consistency and reproducibility
  • Learn how image analysis can be improved to better correlate assay results with patient outcomes
Watch now
Explore four workflow presentations from our research and development scientists that provide guidance to further understand tumoroid derivation and tumoroid characterization methodologies.
PresentationDescription
 Migration and invasion assays using tumoroid models
  • Migration and invasion are crucial in physiological and pathological processes, especially in tumor metastasis
  • This presentation outlines methods for migration and invasion assays using tumoroids, and may be adaptable to other study formats
 Preparation of tumoroids for flow cytometry
  • Techniques like immunofluorescence, western blotting, and flow cytometry can characterize tumoroids, but their 3D structure and basement membrane extract (BME) presence can complicate sample preparation for analysis
  • This presentation provides guidance on preparing tumoroids for analysis by flow cytometry
 Preparation of tumoroids for immunofluorescence
  • Immunofluorescence, western blotting, and flow cytometry can characterize tumoroids, but their 3D structure and presence of BME can complicate sample preparation for analysis
  • This presentation helps provide guidance on adapting tumoroids for immunofluorescence imaging
 Tissue dissociation for tumoroid derivation
  • Derivation success rates are highly dependent on a number of factors, including cancer indication, cell proportion and viability, and processing time; not all samples can generate tumoroid lines
  • This presentation provides guidance on deriving tumoroid lines from fresh tumor tissue samples
Order your OncoPro Tumoroid Culture Medium Kit
OncoPro product photo
Catalog # Name Size Price (TWD) Qty
A5701201 OncoPro™ Tumoroid Culture Medium Kit Each
21,520.00
Online offer
Ends: 30-Jun-2025
27,600.00
Save 6,080.00 (22%)
OncoPro Tumoroid Culture Medium recommended reagents
The reagents below are recommended for culturing cancer organoids using OncoPro Tumoroid Culture Medium Kit.
We have demonstrated the OncoPro Tumoroid Culture Medium Kit supports the culture of several epithelial-derived tumoroids, but additional growth factors may be required for some indications.
For other indications, please Contact our support team to learn how you may be able to use OncoPro medium.
The following recommended reagents can be purchased directly from the Fisher Scientific catalog: 
Recommended reagents from Thermo Fisher Scientific
Thermo Fisher Scientific also offers Gibco OncoPro Tumoroid Cell Lines for purchase. Utilize our general inquiry form below to get a quote and additional information.
Submit an inquiry
Blog articles
Engage with additional resources for tumoroid culture
Discover other products to support culture of tumoroids
Cell culture plastics family image
Nunc Cell Culture Plastics
Thermo Scientific Nunc cell culture plastics offer non-treated flasks to support tumoroid culture.
A bottle of HPLM with a graphic rendering of a cell
Gibco HPLM
Human plasma-like medium is a physiologically relevant medium useful for downstream assays.
For Research Use Only. Not for use in diagnostic procedures.