Conceptual art of a bottle of Gibco charcoal-stripped FBS over a cell
Charcoal-stripped fetal bovine serum is FBS that has been treated with activated carbon to remove non-polar/lipophilic components (i.e., steroids, growth factors, and some cytokines) regardless of molecular weight with little effect on salts, glucose, amino acids, etc. 
Charcoal-stripped FBS is used when studying the impact of steroids on cells in culture. With low levels of lipophilic components in the basal serum/media, androgens, fatty acids, or other molecules of interest can then be added at desired concentrations to study their effects.
Why researchers choose Gibco charcoal-stripped serum
Watch to discover why the removal of androgens and small molecules in charcoal-stripped FBS and dialyzed FBS are so important in cancer research.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Features of Gibco charcoal-stripped serum
      • Allows experimental control over concentrations of lipid-like components
      • Provides reduced lot-to-lot variability of serum hormone levels
      • Every lot measured for estradiol levels pre-and post-treatment (see  example COA)
      • Available in the convenient Gibco One Shot FBS 50 mL bottle format
      Computer generated model of an estrogen receptor
      Research areas using charcoal-stripped FBS*
      • Hormones or hormone receptors (androgens, estrogens, progesterone)
      • Cytotoxic drug response
      • Cellular signaling and reporter assays
      • Prostate and other hormone-responsive cancers
      * These results are based on a review of approximately 10,000 publications using query terms based on six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
      Order charcoal-stripped FBS
      Katalognr. Produkt Größe Preis (EUR) Menge
      12676029 Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL Each
      733,65
      Exklusiv online
      770,00
      Erparnis 36,35 (5%)
      A3382101 Fetal Bovine Serum, charcoal stripped, USDA-approved regions, One Shot™ format, 50 mL Each
      93,65
      Exklusiv online
      97,25
      Erparnis 3,60 (4%)
      Publications using Gibco charcoal-stripped FBS
      Resources
      Fetal Bovine Serum Basics
      Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
      Cell Culture Basics
      Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
      Gibco iMatch Sera Lot Matching Tool 
      Provide your previous Gibco FBS lot number or answer a few questions to find your ideal serum match.
      Related products
      Support
      Find your regional sales representative
      Connect with a regional FBS sales representative.
      Cell Culture & Transfection Support Center
      Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
      Need technical support?
      Contact our expert team for technical and application support of laboratory products.
      For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
      Stylesheet for Classic Wide Template adjustments