Cholesterol Lipid Concentrate (250X)
Product Image
Gibco™
Cholesterol Lipid Concentrate (250X)
250X Cholesterol Lipid Concentrate is a protein-free, chemically-defined supplement used for growing NSO and other lipid dependent cell lines toRead more
Have Questions?
Catalog number 12531018
Price (USD)/ 100 ml
278.00
-
Add to cart
Price (USD)/ 100 ml
278.00
Add to cart
Cholesterol Lipid Concentrate (250X)
Catalog number12531018
Price (USD)/ 100 ml
278.00
-
Add to cart
250X Cholesterol Lipid Concentrate is a protein-free, chemically-defined supplement used for growing NSO and other lipid dependent cell lines to high densities while supporting growth and protein expression without complicating downstream purification. The supplement can be added to 1X media such as CD Hybridoma and filtered with minimal loss of lipids. Custom formulations and packaging configurations as well as supplements used in feeding strategies are available through our Custom Products Services group.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration250 X
Culture TypeMammalian Cell Culture, Insect Cell Culture
FormLiquid
Product TypeCholesterol Lipid Concentrate
Shelf Life18 Months
SterilitySterile
Green FeaturesSustainable packaging
pH6 to 8
Quantity100 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit Size100 ml
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.