Precision control for elevated temperature experiments.
Precise temperature control and monitoring makes this unit ideal for clinical applications where elevated temperature stability is required. Controls are easy to use and do not require complicated programming, while under- and over-temperature alarms provide peace of mind. 
Key features
Enviro-Scan interface
The Thermo Scientific Enviro-Scan user interface operating modes include run, set point, calibration, and system configuration.
Remote alarm
Standard remote alarm contacts and available data outputs allow connection to an in-house monitor/alarm system to track chamber conditions, helping you meet internal and regulatory documentation requirements.
Options for applications
Many options available, including chart recorders and cell roller systems.
Specifications
Heat Only Reach-In Chamber
Size
29 cu. ft.
Exterior (W x H x D)
965 x 2032 x 833mm
38.00 x 80.00 x 33.00 in.
Interior (W x H x D)
787 x 1524 x 686mm
31.00 x 60.00 x 27.00 in.
Net weight
227kg / 500 lbs
Shipping weight
300kg / 660 lbs
Temperature
Temperature range
5°C above ambient to 60°C
Controller, sensor
Environ-Scan - PID, with RTD sensor
Display
Digital dual LED, 0.1°C steps readability & setability
Temperature Uniformity
≤ ±0.4°C @ 30°C
≤ ±0.3°C @ 37°C
≤ ±0.5°C @ 45°C
Temperature Stability
@ 30° to 45°C
Electrical
Voltage
115V, 50/60Hz, 9.0 Amps 3 Wire (including ground)
Plug
Nema 5-15P
Power switch
2-Pole
Interior Convenience receptacles
230W max., for standard EU CEE7/7US Nema 5-15 plug
Alarm Contacts
Temp. deviation and power failure; common, NO, NC; customer connections through RJ11 jack
Order Number
3960
Choose from options designed to support your application
Optional cell roll systems
An optional cell roll system allows extensive production of monolayer cell cultures in standard roller culture vessels. This helps improved oxygenation and exposure of the cells to the media growth area. Combined uniform temperature control and continuous, gentile rotation provided by the cell roll system create the ideal conditions to help increase culture yields.
The reach-in incubator chamber accommodates a cell roller up to 7 decks high with 5 positions per deck for a maximum total of 35 positions, or bottles. 
Incubator with Thermo Scientific Cell Roller System
Thermo Scientific Three-Tier Cell Roller System