Single use pipetting for precision and accuracy
Single use pipetting tools are essential in aseptic applications. Thermo Scientific Samco Transfer Pipettes allow for precise and accurate measurement and transfer of a small volume of liquid. They are ergonomic and easy to use, helping to enable efficient and controlled transfer of samples.
Key Features
Design with the end user in mind
Grips easily – no slipping
Clear, graduated markings
Fine-tip accuracy
Package materials reduces airborne mold spores from entering the testing area
Repeatable results
Inert, nontoxic polyethylene
Manufactured without mold-release agents that could interfere with test results
Reliability
Precision-cut tips deliver a uniform drop size
Controlled processes product a uniform bulb with uncompromised integrity`
Ability to design custom transfer pipettes
Custom design available Ability to design custom transfer pipettes
Select from a range of transfer pipettes to fit your application
*Exact volume is the name of this group of transfer pipettes.  These conveniently dispense a precise volume within a tolerance
Learn more about transfer pipettes
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.
1x1 image pixel for data collection