Digital PCR Community – CAR-T-Cell Monitoring
Exploring the benefits of dPCR to support CAR-T cell therapy research
Digital PCR (dPCR) is an excellent tool for rare target analysis applications because, unlike traditional PCR methods, dPCR divides samples into thousands of individual microreactions, then amplifies isolated molecules to endpoint. This helps to isolate even very small amounts of target for improved chances of detection and quantification. For applications such as CAR-T cell therapy research, where the frequencies of targets are often very low, dPCR is an extremely advantageous tool.
Dr. Raquel Munoz Garcia is an immunologist from the Hospital Universitario Virgen del Rocío in Seville, Spain. In this webinar, she discusses how dPCR was used to support CAR-T cell monitoring research.
Interested in hearing more? Listen to the episode of our Absolute Gene-ius podcast when we had the opportunity to talk with Dr. Munoz and learn more about what she thinks will be the future of cancer research.
Video Player is loading.
Current Time 0:00
Duration 31:59
Loaded: 0%
Stream Type LIVE
Remaining Time 31:59
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Digital PCR Content Library  
    Explore webinars & events, podcasts, publications, technical literature, and more.
    library-dpcr
    Digital PCR Education Center  
    Review dPCR basics and learn more about the technology and its applications.
    dPCR-vs-qPCR
    Talk to a specialist
    Have questions about implementing dPCR in your lab? Let us help you get started!
    Contact-Specialist-Card