dPCR enables quantitative detection of rare sequences in cancer and disease
Graphic image depicting cell-free DNA circulating in the blood stream
Digital PCR (dPCR) is a leading technology for detection and quantification of rare mutations and target sequences in cancer and disease. With the Applied Biosystems QuantStudio Absolute Q Digital PCR System, researchers can quickly and easily quantify relevant targets by leveraging a simple dPCR workflow backed by the trusted performance of TaqMan probe-based assays.
Benefits of digital PCR for rare mutation detection
dPCR can detect rare targets with mutation allele frequencies (MAFs) as low as 0.1%
Precise quantification of rare, single-nucleotide polymorphisms (SNPs) and other mutations over background requires highly sensitive analysis. Because dPCR effectively enriches low-level targets by partitioning samples into individual microchamber reactions, it is uniquely suited for applications requiring quantitative rare target detection, such as liquid biopsy analysis.
Technical note: Rare-target quantification on the the Absolute Q Digital PCR System
quantstudio-absolute-q-dPCR-rare-mutation
Figure 1. A demonstration of the ability of the QuantStudio Absolute Q dPCR System to detect mutant KRAS p.G12V alleles (purple dots) against a background of 99.9% wild type KRAS alleles (green dots) using the KRAS 520 assay and (orange dots) using the KRAS 520 assay. Green dots represent dPCR microchambers positive for both alleles.
dPCR increases sensitivity for rare target detection over conventional method
Absolute quantification using dPCR eliminates the need for a standard curve, improving both the accuracy and reproducibility of rare-target concentration measurement. dPCR also enables detection of ultra-rare target by leveraging pooling data to overcome limitations of sample input volume and concentration.
Poster: Precision genotyping for EGFR+ NSCLC using multiplexed dPCR on cell-free DNA
Technical Note: Improve sensitivity for rare targets by pooling digital PCR data
Graph depicting dPCR and qPCR EGFR mutation panel results
Figure 2. Correlation of EGFR Multiplex Panel results from the Absolute Q dPCR System to the duplex results gathered from qARMS-PCR. Each point represents a donor sample for which data was generated with both methods.
dPCR is an established technology for rare mutation analysis
With the ability to detect very low fractions of mutant targets in a background of abundant wild-type targets, dPCR is a technology of choice for cancer and genetic disease researchers.
Read QuantStudio Absolute Q dPCR publications ›
Video Player is loading.
Current Time 0:00
Duration 3:37
Loaded: 4.51%
Stream Type LIVE
Remaining Time 3:37
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Digital PCR for liquid biopsy analysis
    Liquid biopsy testing enables non-invasive biomarker testing to identify and track oncogenic mutations. Liquid biopsies detect cell-free DNA (cfDNA) released from apoptotic and necrotic cells. cfDNA that is released from cancer cells is referred to as circulating tumor DNA, or ctDNA.
    dPCR enables precise quantification of ctDNA from liquid biopsies
    Characterizing ctDNA helps researchers detect cancer early, measure therapeutic response, quantify residual tumor burden, and monitor emerging resistance to potential therapies. Because ctDNA fragments are typically short and very low in concentration, the sensitivity of dPCR makes it an ideal tool for ctDNA studies.
    Webinar: dPCR in liquid biopsy for management and detection of cancer
    Absolute Q dPCR assays for liquid biopsy
    Absolute Q Liquid Biopsy dPCR assays enable reproducible, specific detection of known somatic mutations. Each predesigned assay has been validated using controls to detect down to 0.1% variant allele frequency in genes relevant for cancer research.
    Absolute Q Liquid Biopsy dPCR assays are:
    • Simple—preformulated and verified assay in a single tube, enabling you to immediately start your research
    • Fast—minimal hands-on time; results in 90 minutes when used with the QuantStudio Absolute Q Digital PCR System
    • Highly Sensitive—detect down to 0.1% variant allele frequency
    • Robust—analyze your data with confidence using verified assays backed by a performance guarantee*
    Download flyer
    Flyer_Figure_AndromedaAssays
    Figure 3. Quantification of cancer mutations using Absolute Q Liquid Biopsy Digital PCR assays on as QuantStudio Absolute Q Digital PCR System.
    Absolute Q liquid biopsy assays ordering information
    To order Absolute Q Liquid Biopsy dPCR assays, indicate the desired quantity in the appropriate assay row, then scroll to the bottom of the table and click 'Add to cart'. Follow in-cart instructions to complete your purchase.
    Gene
    Amino Acid Mutation
    CDS Mutation
    COSM ID
    Assay ID
    Catalog #
    A53732
    700 rxn
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    BRAF
    p.V600K
    c.1798_1799GT>AA
    BRAF_473
    DGU62RA
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    BRAF
    p.V600E
    c.1799_1800TG>AA
    BRAF_475
    DGWCWA7
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_S752>A
    c.2237_2254del18
    EGFR_12367
    DGYMJF2
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_T751delLREAT
    c.2240_2254del15
    EGFR_12369
    DGZTDZY
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_P753>S
    c.2240_2257del18
    EGFR_12370
    DG2W7KW
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.D770_N771insG
    c.2310_2311insGGT
    EGFR_12378
    DG32Z6U
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_A750>P
    c.2239_2248TTAAGAGAAG>C
    EGFR_12382
    DG47VRR
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_T751>P
    c.2239_2251>C
    EGFR_12383
    DG7DPCN
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_S752>V
    c.2237_2255>T
    EGFR_12384
    DG9HHWK
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_P753delinsQ
    c.2239_2258delinsCA
    EGFR_12387
    DGAAAAC
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_T751>Q
    c.2238_2252>GCA
    EGFR_12419
    DGCE3U9
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_A750>P
    c.2238_2248>GC
    EGFR_12422
    DGDJXE6
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_T751>A
    c.2237_2251del15
    EGFR_12678
    DGEPRY3
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_T751delELREAT
    c.2236_2253del18
    EGFR_12728
    DGFVKJZ
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_T751>I
    c.2235_2252>AAT
    EGFR_13551
    DGGZE4X
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_T751delLREAT
    c.2238_2252del15
    EGFR_23571
    DGH49PV
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.S492R
    1476C>A
    EGFR_236670
    DGKA3AT
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.S492R
    1474A>C
    EGFR_236671
    DGMFWVP
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_T751>S
    c.2240_2251del12
    EGFR_6210
    DGNKRFM
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L861Q
    c.2582T>A
    EGFR_6213
    DGPRJZJ
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_S752>D
    c.2238_2255del18
    EGFR_6220
    DGRWEKG
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_A750delELREA
    c.2235_2249del15
    EGFR_6223
    DGTZ76E
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L858R
    c.2573T>G
    EGFR_6224
    DGU62RC
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.E746_A750delELREA
    c.2236_2250del15
    EGFR_6225
    DGWCWA9
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.G719S
    c.2155G>A
    EGFR_6252
    DGXGPV6
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.G719C
    c.2155G>T
    EGFR_6253
    DGYMJF3
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.L747_S752delLREATS
    c.2239_2256del18
    EGFR_6255
    DGZTDZZ
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    ERBB2
    p.L755S
    c.2264T>C
    ERBB2_14060
    DG2W7KX
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    ERBB2
    p.V777L
    c.2329G>T
    ERBB3_14062
    DG32Z6V
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    IDH1
    p.R132C
    c.394C>T
    IDH1_28747
    DG47VRT
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    JAK2
    p.V617F
    c.1849G>T
    JAK2_12600
    DG7DPCP
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KIT
    p.D816V
    c.2447A>T
    KIT_1314
    DG9HHWM
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KIT
    p.D816E
    c.2448C>G
    KIT_19285
    DGAAAAD
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61R
    c.182_183AA>GT
    KRAS_1168052
    DGCE3VA
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.A146T
    436G>T
    KRAS_19404
    DGDJXE7
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.A146V
    437C>T
    KRAS_19900
    DGEPRY4
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.A416P
    438G>C
    KRAS_19905
    DGFVKJ2
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.K117N
    351A>T
    KRAS_28519
    DGGZE4Y
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G12C
    c.34G>T
    KRAS_516
    DGH49PW
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G12S
    c.34G>A
    KRAS_517
    DGKA3AU
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G12R
    c.34G>C
    KRAS_518
    DGMFWVR
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G12V
    c.35G>T
    KRAS_520
    DGNKRFN
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G13C
    c.37G>T
    KRAS_527
    DGRWEKH
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G13S
    c.37G>A
    KRAS_528
    DGTZ76F
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G13R
    c.37G>C
    KRAS_529
    DGU62RD
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61K
    c.181C>A
    KRAS_549
    DGWCWCA
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61R
    c.182A>G
    KRAS_552
    DGXGPV7
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61L
    c.182A>T
    KRAS_553
    DGYMJF4
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61H
    c.183A>C
    KRAS_554
    DGZTDZ2
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.Q61H
    c.183A>T
    KRAS_555
    DG2W7KY
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    MLF2
    p.R158W
    c.472C>T
    MLF2_312886
    DG32Z6W
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NPM1
    p.W288fs*12
    c.863_864insTCTG
    NPM1_17559
    DG47VRU
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61L
    c.181_182CA>TT
    NRAS_12725
    DG7DPCR
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61K
    c.180_181AC>TA
    NRAS_12730
    DG9HHWN
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G12S
    c.34G>A
    NRAS_563
    DGAAAAE
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G12D
    c.35G>A
    NRAS_564
    DGCE3VC
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G12A
    c.35G>C
    NRAS_565
    DGDJXE9
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G12V
    c.35G>T
    NRAS_566
    DGEPRY6
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G13R
    c.37G>C
    NRAS_569
    DGFVKJ3
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G13C
    c.37G>T
    NRAS_570
    DGGZE4Z
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.G13G
    c.39T>C
    NRAS_576
    DGH49PX
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61K
    c.181C>A
    NRAS_580
    DGKA3AV
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61E
    c.181C>G
    NRAS_581
    DGMFWVT
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61P
    c.182A>C
    NRAS_582
    DGNKRFP
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61L
    c.182A>T
    NRAS_583
    DGPRJZM
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    NRAS
    p.Q61R
    c.182A>G
    NRAS_584
    DGRWEKJ
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3CA
    p.M1043V
    c.3127A>G
    PIK3CA_12591
    DGTZ76G
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3CA
    p.M1043I
    c.3129G>T
    PIK3CA_773
    DGU62RE
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3CA
    p.H1047R
    c.3140 A>G
    PIK3CA_775
    DGWCWCC
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3CA
    p.H1047L
    c.3140 A>T
    PIK3CA_776
    DGXGPV9
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3R1
    p.K567E
    c.1699A>G
    PIK3R1_1069611
    DGYMJF6
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R248W
    c.742C>T
    TP53_10656
    DGZTDZ3
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R273H
    c.818G>A
    TP53_10660
    DG2W7KZ
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R248Q
    c.743G>A
    TP53_10662
    DG32Z6X
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R282W
    c.844C>T
    TP53_10704
    DG47VRV
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R273L
    c.818G>T
    TP53_10779
    DG7DPCT
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R342*
    c.1024C>T
    TP53_11073
    DG9HHWP
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.G245C
    c.733G>T
    TP53_11081
    DGAAAAF
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.V173L
    c.517G>T
    TP53_43559
    DGCE3VD
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R248Q
    c.743_744GG>AA
    TP53_44908
    DGDJXFA
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R248W
    c.741_742CC>TT
    TP53_6545
    DGEPRY7
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.R248L
    c.743G>T
    TP53_6549
    DGFVKJ4
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    TP53
    p.G245S
    c.733G>A
    TP53_6932
    DGGZE42
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.T790M
    c.2369C>T
    EGFR_6240
    DGNKRFR
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    P.C797S
    c.2390G>C
    EGFR_5945664
    DGWCWCD
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    EGFR
    p.C797S
    c.2398T>A
    EGFR_6493937
    DGU62RF
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    PIK3CA
    p.E542K
    c.1624G>A
    PIK3CA_760
    DGXGPWA
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    KRAS
    p.G12D
    c.35G>A
    KRAS_521
    DG9HHWR
    Gene:
    Amino Acid Mutation:
    CDS Mutation:
    COSM ID:
    Assay ID:
    Catalog #
    A53732
    700 rxn:
    BRAF
    p.V600E
    c.1799T>A
    BRAF_476
    DG32Z6Y
    Other dPCR assays for rare target detection
    dpcr-assays-2
    Custom dPCR assays
    Can’t find the assay you’re looking for? Design a digital PCR assay with a self service tool or get help with our design services.
    Launch assay design hub ›
    dpcr-assays-1
    TaqMan Assays
    Adapt an existing TaqMan assay for use on the QuantStudio Absolute Q Digital PCR System.
    Explore assays ›
    Quick reference guide
    Resources
    Photo of the QuantStudio Absolute Q Digital PCR System over a graphical background
    QuantStudio Absolute Q Digital PCR System
    Explore our powerfully simple dPCR platform powered by microfluidic array plate (MAP) technology.
    Explore now ›
    Master mixes, microfluidic array plates, and other consumables for the QuantStudio Absolute Q Digital PCR system
    Digital PCR reagents & consumables
    Find master mixes and microfludic array plates and accessories for use with the QuantStudio Absolute Q dPCR System.
    See all products ›
    Scientist in lab preparing dPCR automated run
    Talk to a specialist
    Have questions about implementing dPCR in your lab? Let us help you get started.
    Contact us ›
     *Terms and conditions apply. For complete details of the guarantee, go to thermofisher.com/absoluteqassayguarantee
    For Research Use Only. Not for use in diagnostic procedures.
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs