dPCR enables quantitative detection of rare sequences in cancer and disease
Graphic image depicting cell-free DNA circulating in the blood stream
Digital PCR (dPCR) is a leading technology for detection and quantification of rare mutations and target sequences in cancer and disease. With the Applied Biosystems QuantStudio Absolute Q Digital PCR System, researchers can quickly and easily quantify relevant targets by leveraging a simple dPCR workflow backed by the trusted performance of TaqMan probe-based assays.
Benefits of digital PCR for rare mutation detection
dPCR can detect rare targets with mutation allele frequencies (MAFs) as low as 0.1%
Precise quantification of rare, single-nucleotide polymorphisms (SNPs) and other mutations over background requires highly sensitive analysis. Because dPCR effectively enriches low-level targets by partitioning samples into individual microchamber reactions, it is uniquely suited for applications requiring quantitative rare target detection, such as liquid biopsy analysis.
Technical note: Rare-target quantification on the the Absolute Q Digital PCR System
quantstudio-absolute-q-dPCR-rare-mutation
Figure 1. A demonstration of the ability of the QuantStudio Absolute Q dPCR System to detect mutant KRAS p.G12V alleles (purple dots) against a background of 99.9% wild type KRAS alleles (green dots) using the KRAS 520 assay and (orange dots) using the KRAS 520 assay. Green dots represent dPCR microchambers positive for both alleles.
dPCR increases sensitivity for rare target detection over conventional method
Absolute quantification using dPCR eliminates the need for a standard curve, improving both the accuracy and reproducibility of rare-target concentration measurement. dPCR also enables detection of ultra-rare target by leveraging pooling data to overcome limitations of sample input volume and concentration.
Poster: Precision genotyping for EGFR+ NSCLC using multiplexed dPCR on cell-free DNA
Technical Note: Improve sensitivity for rare targets by pooling digital PCR data
Graph depicting dPCR and qPCR EGFR mutation panel results
Figure 2. Correlation of EGFR Multiplex Panel results from the Absolute Q dPCR System to the duplex results gathered from qARMS-PCR. Each point represents a donor sample for which data was generated with both methods.
dPCR is an established technology for rare mutation analysis
With the ability to detect very low fractions of mutant targets in a background of abundant wild-type targets, dPCR is a technology of choice for cancer and genetic disease researchers.
Read QuantStudio Absolute Q dPCR publications ›
Video Player is loading.
Current Time 0:00
Duration 3:37
Loaded: 0%
Stream Type LIVE
Remaining Time 3:37
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Digital PCR for liquid biopsy analysis
    Liquid biopsy testing enables non-invasive biomarker testing to identify and track oncogenic mutations. Liquid biopsies detect cell-free DNA (cfDNA) released from apoptotic and necrotic cells. cfDNA that is released from cancer cells is referred to as circulating tumor DNA, or ctDNA.
    dPCR enables precise quantification of ctDNA from liquid biopsies
    Characterizing ctDNA helps researchers detect cancer early, measure therapeutic response, quantify residual tumor burden, and monitor emerging resistance to potential therapies. Because ctDNA fragments are typically short and very low in concentration, the sensitivity of dPCR makes it an ideal tool for ctDNA studies.
    Webinar: dPCR in liquid biopsy for management and detection of cancer
    Absolute Q dPCR assays for liquid biopsy
    Absolute Q Liquid Biopsy dPCR assays enable reproducible, specific detection of known somatic mutations. Each predesigned assay has been validated using controls to detect down to 0.1% variant allele frequency in genes relevant for cancer research.
    Absolute Q Liquid Biopsy dPCR assays are:
    • Simple—preformulated and verified assay in a single tube, enabling you to immediately start your research
    • Fast—minimal hands-on time; results in 90 minutes when used with the QuantStudio Absolute Q Digital PCR System
    • Highly Sensitive—detect down to 0.1% variant allele frequency
    • Robust—analyze your data with confidence using verified assays backed by a performance guarantee*
    Download flyer
    Flyer_Figure_AndromedaAssays
    Figure 3. Quantification of cancer mutations using Absolute Q Liquid Biopsy Digital PCR assays on as QuantStudio Absolute Q Digital PCR System.
    Absolute Q liquid biopsy assays ordering information
    To order Absolute Q Liquid Biopsy dPCR assays, indicate the desired quantity in the appropriate assay row, then scroll to the bottom of the table and click 'Add to cart'. Follow in-cart instructions to complete your purchase.
    Other dPCR assays for rare target detection
    dpcr-assays-2
    Custom dPCR assays
    Can’t find the assay you’re looking for? Design a digital PCR assay with a self service tool or get help with our design services.
    Launch assay design hub ›
    dpcr-assays-1
    TaqMan Assays
    Adapt an existing TaqMan assay for use on the QuantStudio Absolute Q Digital PCR System.
    Explore assays ›
    Quick reference guide
    Resources
    Photo of the QuantStudio Absolute Q Digital PCR System over a graphical background
    QuantStudio Absolute Q Digital PCR System
    Explore our powerfully simple dPCR platform powered by microfluidic array plate (MAP) technology.
    Explore now ›
    Master mixes, microfluidic array plates, and other consumables for the QuantStudio Absolute Q Digital PCR system
    Digital PCR reagents & consumables
    Find master mixes and microfludic array plates and accessories for use with the QuantStudio Absolute Q dPCR System.
    See all products ›
    Scientist in lab preparing dPCR automated run
    Talk to a specialist
    Have questions about implementing dPCR in your lab? Let us help you get started.
    Contact us ›
     *Terms and conditions apply. For complete details of the guarantee, go to thermofisher.com/absoluteqassayguarantee
    For Research Use Only. Not for use in diagnostic procedures.
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs